Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 16412986-16413494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNC2
Entrez Gene ID
54796
Gene Name
basonuclin 2
Gene Aliases
BSN2
UniGene
Hs.656581
Gene Chromosome Location
Chr.9: 16409503-16870838 on Build GRCh38
Transcripts
NM_001317939.1 NM_001317940.1 NM_017637.5 XM_011517924.1 XM_017014816.1 XM_017014817.1 XM_017014818.1 XM_017014820.1 XM_017014822.1 XM_017014823.1 XM_017014824.1 XM_017014825.1 XM_017014829.1

SNP Detail

SNP ID Chromosome Location
rs1000116425 Chr.9: 16413170-16413170 on Build GRCh38
rs367880769 Chr.9: 16413356-16413357 on Build GRCh38
rs200079950 Chr.9: 16413190-16413190 on Build GRCh38
rs977226831 Chr.9: 16413187-16413187 on Build GRCh38
rs59282202 Chr.9: 16413414-16413415 on Build GRCh38
rs770620752 Chr.9: 16413150-16413151 on Build GRCh38
rs968801535 Chr.9: 16413096-16413096 on Build GRCh38
rs956051811 Chr.9: 16413457-16413457 on Build GRCh38
rs574864912 Chr.9: 16413056-16413056 on Build GRCh38
rs985395009 Chr.9: 16413230-16413230 on Build GRCh38
rs1031692819 Chr.9: 16413439-16413439 on Build GRCh38
rs914688361 Chr.9: 16413067-16413067 on Build GRCh38
rs775036340 Chr.9: 16413017-16413017 on Build GRCh38
rs895974774 Chr.9: 16413135-16413135 on Build GRCh38
rs1057323905 Chr.9: 16413073-16413073 on Build GRCh38
rs1035687047 Chr.9: 16413040-16413040 on Build GRCh38
rs542880119 Chr.9: 16413089-16413089 on Build GRCh38
rs192464317 Chr.9: 16413407-16413407 on Build GRCh38
rs34575896 Chr.9: 16413200-16413200 on Build GRCh38
rs866658150 Chr.9: 16413171-16413171 on Build GRCh38
rs541019297 Chr.9: 16413260-16413260 on Build GRCh38
rs909563011 Chr.9: 16413019-16413019 on Build GRCh38
rs1056587557 Chr.9: 16413337-16413337 on Build GRCh38
rs373412079 Chr.9: 16413108-16413108 on Build GRCh38
rs531965876 Chr.9: 16413238-16413238 on Build GRCh38
rs940983821 Chr.9: 16413030-16413030 on Build GRCh38
rs1003696961 Chr.9: 16413065-16413065 on Build GRCh38
rs759629769 Chr.9: 16413418-16413419 on Build GRCh38
rs182451116 Chr.9: 16413112-16413112 on Build GRCh38
rs1029529034 Chr.9: 16413207-16413207 on Build GRCh38
rs1013133995 Chr.9: 16413141-16413141 on Build GRCh38
rs1040519249 Chr.9: 16413384-16413384 on Build GRCh38
rs918245095 Chr.9: 16413278-16413278 on Build GRCh38
rs148133806 Chr.9: 16413432-16413432 on Build GRCh38
rs939712352 Chr.9: 16413181-16413181 on Build GRCh38
rs1037192190 Chr.9: 16413047-16413047 on Build GRCh38
rs896005783 Chr.9: 16413351-16413351 on Build GRCh38
rs956273289 Chr.9: 16413045-16413045 on Build GRCh38
rs979234706 Chr.9: 16413100-16413100 on Build GRCh38
rs1044509812 Chr.9: 16413295-16413295 on Build GRCh38
rs71325969 Chr.9: 16413190-16413190 on Build GRCh38
rs939143098 Chr.9: 16413333-16413333 on Build GRCh38
rs764115308 Chr.9: 16413235-16413235 on Build GRCh38
rs953832290 Chr.9: 16413212-16413212 on Build GRCh38
rs545965856 Chr.9: 16413257-16413257 on Build GRCh38
rs398010371 Chr.9: 16413372-16413372 on Build GRCh38
rs201648557 Chr.9: 16413188-16413188 on Build GRCh38
rs970244596 Chr.9: 16413018-16413018 on Build GRCh38
rs189110399 Chr.9: 16413251-16413251 on Build GRCh38
rs948849009 Chr.9: 16413372-16413372 on Build GRCh38
rs5896689 Chr.9: 16413357-16413357 on Build GRCh38
rs1023149419 Chr.9: 16413157-16413157 on Build GRCh38
rs891781025 Chr.9: 16413450-16413450 on Build GRCh38
rs1044634383 Chr.9: 16413406-16413406 on Build GRCh38
rs192720739 Chr.9: 16413052-16413052 on Build GRCh38
rs113643183 Chr.9: 16413093-16413093 on Build GRCh38
rs74851347 Chr.9: 16413160-16413160 on Build GRCh38
rs532095133 Chr.9: 16413254-16413254 on Build GRCh38
rs943743 Chr.9: 16413178-16413178 on Build GRCh38

View Full Product Details