Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 16417243-16417738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNC2
Entrez Gene ID
54796
Gene Name
basonuclin 2
Gene Aliases
BSN2
UniGene
Hs.656581
Gene Chromosome Location
Chr.9: 16409503-16870838 on Build GRCh38
Transcripts
NM_001317939.1 NM_001317940.1 NM_017637.5 XM_011517924.1 XM_017014816.1 XM_017014817.1 XM_017014818.1 XM_017014819.1 XM_017014820.1 XM_017014821.1 XM_017014822.1 XM_017014823.1 XM_017014824.1 XM_017014825.1 XM_017014829.1

SNP Detail

SNP ID Chromosome Location
rs915472920 Chr.9: 16417281-16417281 on Build GRCh38
rs751893163 Chr.9: 16417335-16417335 on Build GRCh38
rs1002725657 Chr.9: 16417538-16417538 on Build GRCh38
rs145262491 Chr.9: 16417341-16417341 on Build GRCh38
rs1014753811 Chr.9: 16417305-16417305 on Build GRCh38
rs768070930 Chr.9: 16417650-16417650 on Build GRCh38
rs1021338056 Chr.9: 16417594-16417594 on Build GRCh38
rs1047156512 Chr.9: 16417323-16417324 on Build GRCh38
rs985286238 Chr.9: 16417707-16417707 on Build GRCh38
rs772271002 Chr.9: 16417333-16417333 on Build GRCh38
rs1011043356 Chr.9: 16417572-16417572 on Build GRCh38
rs1034006855 Chr.9: 16417542-16417542 on Build GRCh38
rs191036549 Chr.9: 16417321-16417321 on Build GRCh38
rs966308569 Chr.9: 16417694-16417694 on Build GRCh38
rs376913192 Chr.9: 16417512-16417513 on Build GRCh38
rs544033076 Chr.9: 16417310-16417310 on Build GRCh38
rs941451351 Chr.9: 16417697-16417697 on Build GRCh38
rs183346062 Chr.9: 16417576-16417576 on Build GRCh38
rs1022901502 Chr.9: 16417349-16417349 on Build GRCh38
rs77739492 Chr.9: 16417702-16417702 on Build GRCh38
rs989661673 Chr.9: 16417272-16417272 on Build GRCh38
rs7021128 Chr.9: 16417471-16417471 on Build GRCh38
rs976111888 Chr.9: 16417590-16417590 on Build GRCh38
rs191206526 Chr.9: 16417567-16417567 on Build GRCh38
rs548982810 Chr.9: 16417411-16417411 on Build GRCh38
rs556325063 Chr.9: 16417660-16417660 on Build GRCh38
rs539322873 Chr.9: 16417512-16417512 on Build GRCh38
rs75500294 Chr.9: 16417370-16417370 on Build GRCh38
rs576152178 Chr.9: 16417625-16417625 on Build GRCh38
rs554216034 Chr.9: 16417513-16417513 on Build GRCh38
rs953365276 Chr.9: 16417703-16417703 on Build GRCh38
rs1036144601 Chr.9: 16417488-16417488 on Build GRCh38
rs543479175 Chr.9: 16417645-16417645 on Build GRCh38
rs940498830 Chr.9: 16417477-16417477 on Build GRCh38
rs968630157 Chr.9: 16417623-16417623 on Build GRCh38
rs906462759 Chr.9: 16417718-16417718 on Build GRCh38
rs777218610 Chr.9: 16417437-16417437 on Build GRCh38
rs755667489 Chr.9: 16417686-16417686 on Build GRCh38
rs186687509 Chr.9: 16417442-16417442 on Build GRCh38
rs765763998 Chr.9: 16417445-16417445 on Build GRCh38
rs76817689 Chr.9: 16417306-16417306 on Build GRCh38
rs907166804 Chr.9: 16417534-16417534 on Build GRCh38
rs1005226993 Chr.9: 16417290-16417290 on Build GRCh38
rs750802107 Chr.9: 16417462-16417462 on Build GRCh38
rs536283381 Chr.9: 16417588-16417588 on Build GRCh38
rs886258795 Chr.9: 16417326-16417326 on Build GRCh38
rs554525205 Chr.9: 16417591-16417591 on Build GRCh38
rs550363923 Chr.9: 16417458-16417458 on Build GRCh38
rs577806819 Chr.9: 16417701-16417701 on Build GRCh38
rs956461716 Chr.9: 16417552-16417557 on Build GRCh38
rs186647725 Chr.9: 16417307-16417307 on Build GRCh38
rs1927627 Chr.9: 16417433-16417433 on Build GRCh38
rs182977194 Chr.9: 16417401-16417401 on Build GRCh38
rs999729078 Chr.9: 16417633-16417633 on Build GRCh38
rs1033972501 Chr.9: 16417308-16417308 on Build GRCh38
rs533265061 Chr.9: 16417331-16417331 on Build GRCh38

View Full Product Details