Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19305257-19305806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4C
Entrez Gene ID
55667
Gene Name
DENN domain containing 4C
Gene Aliases
C9orf55, C9orf55B, RAB10GEF, bA513M16.3
UniGene
Hs.249591
Gene Chromosome Location
Chr.9: 19230765-19374268 on Build GRCh38
Transcripts
NM_017925.6 NR_073201.3 XM_006716804.3 XM_006716805.3 XM_006716807.3 XM_011517959.2 XM_017014917.1 XM_017014918.1 XM_017014919.1 XM_017014920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3433053 c.754G>T p.D252Y Chr.9: 19305502-19305502 on Build GRCh38
4464207 c.614C>T p.P205L Chr.9: 19305362-19305362 on Build GRCh38
5919704 c.604-5C>T p.? Chr.9: 19305347-19305347 on Build GRCh38
3091439 c.643C>A p.L215I Chr.9: 19305391-19305391 on Build GRCh38
1674403 c.764C>T p.T255M Chr.9: 19305512-19305512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938677594 Chr.9: 19305672-19305672 on Build GRCh38
rs150851153 Chr.9: 19305523-19305523 on Build GRCh38
rs527294809 Chr.9: 19305706-19305706 on Build GRCh38
rs1038334164 Chr.9: 19305349-19305349 on Build GRCh38
rs202105741 Chr.9: 19305488-19305488 on Build GRCh38
rs746784015 Chr.9: 19305323-19305323 on Build GRCh38
rs138893671 Chr.9: 19305375-19305375 on Build GRCh38
rs534431134 Chr.9: 19305551-19305551 on Build GRCh38
rs947930056 Chr.9: 19305733-19305733 on Build GRCh38
rs747650091 Chr.9: 19305724-19305724 on Build GRCh38
rs541548793 Chr.9: 19305702-19305703 on Build GRCh38
rs769832872 Chr.9: 19305497-19305497 on Build GRCh38
rs201941170 Chr.9: 19305522-19305522 on Build GRCh38
rs562157504 Chr.9: 19305387-19305387 on Build GRCh38
rs779091934 Chr.9: 19305330-19305330 on Build GRCh38
rs757390408 Chr.9: 19305554-19305554 on Build GRCh38
rs142917193 Chr.9: 19305439-19305439 on Build GRCh38
rs1031062168 Chr.9: 19305728-19305728 on Build GRCh38
rs1030267833 Chr.9: 19305589-19305589 on Build GRCh38
rs747942042 Chr.9: 19305435-19305435 on Build GRCh38
rs772907784 Chr.9: 19305321-19305321 on Build GRCh38
rs781183320 Chr.9: 19305558-19305558 on Build GRCh38
rs75140950 Chr.9: 19305499-19305499 on Build GRCh38
rs747515678 Chr.9: 19305656-19305656 on Build GRCh38
rs766638485 Chr.9: 19305413-19305413 on Build GRCh38
rs147017743 Chr.9: 19305443-19305443 on Build GRCh38
rs776656069 Chr.9: 19305335-19305335 on Build GRCh38
rs76251519 Chr.9: 19305573-19305573 on Build GRCh38
rs558183996 Chr.9: 19305693-19305693 on Build GRCh38
rs560007225 Chr.9: 19305664-19305664 on Build GRCh38
rs1038215243 Chr.9: 19305735-19305735 on Build GRCh38
rs758719085 Chr.9: 19305610-19305610 on Build GRCh38
rs200445973 Chr.9: 19305517-19305517 on Build GRCh38
rs777705600 Chr.9: 19305547-19305547 on Build GRCh38
rs113068028 Chr.9: 19305540-19305540 on Build GRCh38
rs777987342 Chr.9: 19305466-19305466 on Build GRCh38
rs536133564 Chr.9: 19305624-19305624 on Build GRCh38
rs903580208 Chr.9: 19305717-19305717 on Build GRCh38
rs934047516 Chr.9: 19305307-19305307 on Build GRCh38
rs761363944 Chr.9: 19305519-19305519 on Build GRCh38
rs746471694 Chr.9: 19305719-19305719 on Build GRCh38
rs762458405 Chr.9: 19305504-19305504 on Build GRCh38
rs573148413 Chr.9: 19305320-19305320 on Build GRCh38
rs201749278 Chr.9: 19305416-19305416 on Build GRCh38
rs539652662 Chr.9: 19305668-19305668 on Build GRCh38
rs898298500 Chr.9: 19305777-19305777 on Build GRCh38
rs750758466 Chr.9: 19305410-19305410 on Build GRCh38
rs763762009 Chr.9: 19305403-19305403 on Build GRCh38
rs758582867 Chr.9: 19305458-19305458 on Build GRCh38
rs1056363871 Chr.9: 19305690-19305690 on Build GRCh38
rs761040561 Chr.9: 19305411-19305411 on Build GRCh38
rs760054923 Chr.9: 19305537-19305537 on Build GRCh38
rs752727787 Chr.9: 19305457-19305457 on Build GRCh38
rs758450700 Chr.9: 19305544-19305544 on Build GRCh38
rs79987248 Chr.9: 19305513-19305513 on Build GRCh38
rs1033276773 Chr.9: 19305566-19305566 on Build GRCh38
rs896336613 Chr.9: 19305703-19305703 on Build GRCh38
rs762563720 Chr.9: 19305390-19305390 on Build GRCh38
rs140468194 Chr.9: 19305306-19305306 on Build GRCh38
rs946106866 Chr.9: 19305378-19305378 on Build GRCh38
rs577366688 Chr.9: 19305296-19305296 on Build GRCh38
rs780308295 Chr.9: 19305638-19305638 on Build GRCh38
rs557707756 Chr.9: 19305666-19305666 on Build GRCh38
rs1025317871 Chr.9: 19305539-19305539 on Build GRCh38
rs941344182 Chr.9: 19305643-19305643 on Build GRCh38
rs575938607 Chr.9: 19305295-19305295 on Build GRCh38
rs746795622 Chr.9: 19305469-19305469 on Build GRCh38
rs569439261 Chr.9: 19305667-19305667 on Build GRCh38
rs1049822676 Chr.9: 19305311-19305311 on Build GRCh38
rs112734731 Chr.9: 19305637-19305637 on Build GRCh38
rs897024840 Chr.9: 19305363-19305363 on Build GRCh38
rs759534184 Chr.9: 19305373-19305373 on Build GRCh38
rs558768575 Chr.9: 19305287-19305287 on Build GRCh38
rs989013575 Chr.9: 19305588-19305588 on Build GRCh38
rs929776445 Chr.9: 19305779-19305779 on Build GRCh38
rs777430379 Chr.9: 19305621-19305621 on Build GRCh38
rs928685266 Chr.9: 19305670-19305670 on Build GRCh38
rs781131633 Chr.9: 19305483-19305483 on Build GRCh38
rs1044798584 Chr.9: 19305712-19305712 on Build GRCh38
rs751875472 Chr.9: 19305550-19305550 on Build GRCh38
rs770818546 Chr.9: 19305477-19305477 on Build GRCh38
rs139021247 Chr.9: 19305444-19305444 on Build GRCh38
rs764116784 Chr.9: 19305288-19305288 on Build GRCh38
rs1008520039 Chr.9: 19305541-19305541 on Build GRCh38

View Full Product Details