Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 23704811-23705316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL2
Entrez Gene ID
1993
Gene Name
ELAV like neuron-specific RNA binding protein 2
Gene Aliases
HEL-N1, HELN1, HUB
UniGene
Hs.166109
Gene Chromosome Location
Chr.9: 23690099-23850757 on Build GRCh38
Transcripts
NM_001171195.1 NM_001171197.1 NM_004432.3 XM_005251393.3 XM_005251394.3 XM_005251395.3 XM_006716734.3 XM_006716735.3 XM_006716736.3 XM_011517774.2 XM_011517776.2 XM_011517777.2 XM_011517778.2 XM_011517779.2 XM_011517780.2 XM_011517783.2 XM_011517784.2 XM_011517785.2 XM_011517786.2 XM_017014408.1 XM_017014409.1 XM_017014410.1 XM_017014411.1 XM_017014412.1 XM_017014413.1 XM_017014414.1 XM_017014415.1 XM_017014416.1 XM_017014417.1 XM_017014418.1 XM_017014419.1 XM_017014420.1 XM_017014421.1 XM_017014422.1 XM_017014423.1 XM_017014424.1 XM_017014425.1 XM_017014426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653161 c.492G>T p.M164I Chr.9: 23704997-23704997 on Build GRCh38
1578771 c.347G>A p.R116H Chr.9: 23705058-23705058 on Build GRCh38
3656668 c.339C>T p.S113S Chr.9: 23705066-23705066 on Build GRCh38
6653160 c.408G>T p.M136I Chr.9: 23704997-23704997 on Build GRCh38
3656656 c.558C>T p.V186V Chr.9: 23704931-23704931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767310033 Chr.9: 23704976-23704976 on Build GRCh38
rs958640409 Chr.9: 23704879-23704879 on Build GRCh38
rs768830241 Chr.9: 23705094-23705094 on Build GRCh38
rs897509045 Chr.9: 23705247-23705247 on Build GRCh38
rs80052308 Chr.9: 23705258-23705258 on Build GRCh38
rs1007743495 Chr.9: 23705128-23705128 on Build GRCh38
rs778552185 Chr.9: 23704919-23704919 on Build GRCh38
rs35923981 Chr.9: 23705256-23705257 on Build GRCh38
rs971335959 Chr.9: 23705273-23705273 on Build GRCh38
rs568494492 Chr.9: 23705104-23705104 on Build GRCh38
rs1045502459 Chr.9: 23704839-23704839 on Build GRCh38
rs770201356 Chr.9: 23704944-23704944 on Build GRCh38
rs10117078 Chr.9: 23705149-23705149 on Build GRCh38
rs373602728 Chr.9: 23705005-23705005 on Build GRCh38
rs202048762 Chr.9: 23704955-23704955 on Build GRCh38
rs755962046 Chr.9: 23704905-23704905 on Build GRCh38
rs1045855639 Chr.9: 23705077-23705077 on Build GRCh38
rs1046793464 Chr.9: 23704898-23704898 on Build GRCh38
rs961860090 Chr.9: 23704995-23704995 on Build GRCh38
rs762957101 Chr.9: 23704883-23704883 on Build GRCh38
rs750389844 Chr.9: 23704984-23704984 on Build GRCh38
rs890748568 Chr.9: 23705201-23705201 on Build GRCh38
rs867331367 Chr.9: 23704968-23704968 on Build GRCh38
rs746936739 Chr.9: 23704977-23704977 on Build GRCh38
rs774384105 Chr.9: 23704964-23704964 on Build GRCh38
rs949415668 Chr.9: 23704896-23704896 on Build GRCh38
rs932155323 Chr.9: 23705179-23705179 on Build GRCh38
rs1050720261 Chr.9: 23704979-23704979 on Build GRCh38
rs372183542 Chr.9: 23704985-23704985 on Build GRCh38
rs957210116 Chr.9: 23705229-23705229 on Build GRCh38
rs1014679711 Chr.9: 23704837-23704837 on Build GRCh38
rs372563684 Chr.9: 23705018-23705018 on Build GRCh38
rs369566672 Chr.9: 23705122-23705122 on Build GRCh38
rs528481339 Chr.9: 23704954-23704954 on Build GRCh38
rs1028270193 Chr.9: 23704865-23704865 on Build GRCh38
rs771308416 Chr.9: 23704926-23704926 on Build GRCh38
rs962065029 Chr.9: 23704843-23704843 on Build GRCh38
rs747705065 Chr.9: 23704922-23704922 on Build GRCh38
rs920424515 Chr.9: 23704864-23704864 on Build GRCh38
rs182816492 Chr.9: 23705103-23705103 on Build GRCh38
rs775787571 Chr.9: 23704946-23704946 on Build GRCh38
rs757945458 Chr.9: 23705022-23705022 on Build GRCh38
rs10966032 Chr.9: 23705110-23705110 on Build GRCh38
rs1036052027 Chr.9: 23705207-23705207 on Build GRCh38
rs991362587 Chr.9: 23705249-23705249 on Build GRCh38
rs537879287 Chr.9: 23704902-23704902 on Build GRCh38
rs749693675 Chr.9: 23705086-23705086 on Build GRCh38
rs367739377 Chr.9: 23705015-23705015 on Build GRCh38
rs1012788295 Chr.9: 23705290-23705290 on Build GRCh38
rs1049217224 Chr.9: 23705106-23705106 on Build GRCh38
rs535073558 Chr.9: 23705266-23705266 on Build GRCh38
rs781736838 Chr.9: 23704931-23704931 on Build GRCh38
rs577063937 Chr.9: 23705121-23705121 on Build GRCh38
rs144189536 Chr.9: 23704994-23704994 on Build GRCh38
rs766261753 Chr.9: 23705096-23705096 on Build GRCh38
rs918948305 Chr.9: 23705275-23705275 on Build GRCh38
rs34156148 Chr.9: 23704847-23704848 on Build GRCh38
rs552363422 Chr.9: 23704875-23704875 on Build GRCh38
rs770263915 Chr.9: 23705075-23705075 on Build GRCh38
rs781492568 Chr.9: 23705024-23705024 on Build GRCh38
rs371653269 Chr.9: 23704887-23704887 on Build GRCh38
rs878930072 Chr.9: 23704860-23704860 on Build GRCh38
rs764157725 Chr.9: 23704886-23704886 on Build GRCh38
rs796918651 Chr.9: 23705197-23705197 on Build GRCh38
rs781070371 Chr.9: 23705193-23705193 on Build GRCh38
rs1053814298 Chr.9: 23705148-23705148 on Build GRCh38
rs148503259 Chr.9: 23705048-23705048 on Build GRCh38
rs754743473 Chr.9: 23704910-23704910 on Build GRCh38
rs778744173 Chr.9: 23705007-23705007 on Build GRCh38
rs757169095 Chr.9: 23704888-23704888 on Build GRCh38
rs897947224 Chr.9: 23704845-23704845 on Build GRCh38
rs755889540 Chr.9: 23705287-23705287 on Build GRCh38
rs367997379 Chr.9: 23704882-23704882 on Build GRCh38
rs991280587 Chr.9: 23705063-23705063 on Build GRCh38
rs944950841 Chr.9: 23705205-23705205 on Build GRCh38
rs894195392 Chr.9: 23705274-23705274 on Build GRCh38
rs746168167 Chr.9: 23704940-23704940 on Build GRCh38
rs190640430 Chr.9: 23705013-23705013 on Build GRCh38
rs149957439 Chr.9: 23704918-23704918 on Build GRCh38
rs750146848 Chr.9: 23704900-23704900 on Build GRCh38
rs935449566 Chr.9: 23705120-23705120 on Build GRCh38
rs779512700 Chr.9: 23704908-23704908 on Build GRCh38
rs368330671 Chr.9: 23705002-23705002 on Build GRCh38
rs754746993 Chr.9: 23705134-23705134 on Build GRCh38
rs759981754 Chr.9: 23704871-23704871 on Build GRCh38
rs917180724 Chr.9: 23705078-23705078 on Build GRCh38
rs1032817040 Chr.9: 23705215-23705215 on Build GRCh38
rs568766646 Chr.9: 23704957-23704957 on Build GRCh38
rs996274130 Chr.9: 23704978-23704978 on Build GRCh38
rs559827484 Chr.9: 23704899-23704899 on Build GRCh38
rs199665367 Chr.9: 23704846-23704846 on Build GRCh38
rs1050997650 Chr.9: 23705190-23705190 on Build GRCh38
rs1053901943 Chr.9: 23704961-23704961 on Build GRCh38
rs1008572347 Chr.9: 23704986-23704986 on Build GRCh38
rs997520617 Chr.9: 23705178-23705178 on Build GRCh38

View Full Product Details