Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32457252-32457762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX58
Entrez Gene ID
23586
Gene Name
DEXD/H-box helicase 58
Gene Aliases
RIG-I, RIGI, RLR-1, SGMRT2
UniGene
Hs.190622
Gene Chromosome Location
Chr.9: 32455302-32526324 on Build GRCh38
Transcripts
NM_014314.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338082 c.2360A>G p.K787R Chr.9: 32457327-32457327 on Build GRCh38
6041732 c.2356G>T p.E786* Chr.9: 32457331-32457331 on Build GRCh38
6041731 c.2569G>T p.E857* Chr.9: 32457331-32457331 on Build GRCh38
6338081 c.2573A>G p.K858R Chr.9: 32457327-32457327 on Build GRCh38
3375225 c.2576G>A p.R859K Chr.9: 32457324-32457324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022236695 Chr.9: 32457499-32457499 on Build GRCh38
rs553017482 Chr.9: 32457576-32457576 on Build GRCh38
rs923328878 Chr.9: 32457679-32457679 on Build GRCh38
rs776072278 Chr.9: 32457368-32457368 on Build GRCh38
rs921056342 Chr.9: 32457671-32457671 on Build GRCh38
rs571056308 Chr.9: 32457484-32457484 on Build GRCh38
rs747835369 Chr.9: 32457494-32457495 on Build GRCh38
rs1036785614 Chr.9: 32457524-32457524 on Build GRCh38
rs142758049 Chr.9: 32457318-32457318 on Build GRCh38
rs758149047 Chr.9: 32457452-32457452 on Build GRCh38
rs774823967 Chr.9: 32457384-32457384 on Build GRCh38
rs747937039 Chr.9: 32457451-32457451 on Build GRCh38
rs1006675812 Chr.9: 32457318-32457319 on Build GRCh38
rs200263450 Chr.9: 32457440-32457440 on Build GRCh38
rs201050034 Chr.9: 32457557-32457558 on Build GRCh38
rs970246621 Chr.9: 32457503-32457503 on Build GRCh38
rs764349220 Chr.9: 32457377-32457377 on Build GRCh38
rs911080674 Chr.9: 32457626-32457626 on Build GRCh38
rs746468623 Chr.9: 32457361-32457361 on Build GRCh38
rs778542811 Chr.9: 32457450-32457450 on Build GRCh38
rs997479586 Chr.9: 32457447-32457447 on Build GRCh38
rs747735182 Chr.9: 32457336-32457336 on Build GRCh38
rs1035839826 Chr.9: 32457346-32457346 on Build GRCh38
rs573952640 Chr.9: 32457309-32457310 on Build GRCh38
rs558792146 Chr.9: 32457611-32457611 on Build GRCh38
rs567033543 Chr.9: 32457406-32457406 on Build GRCh38
rs942477816 Chr.9: 32457647-32457647 on Build GRCh38
rs767680439 Chr.9: 32457417-32457417 on Build GRCh38
rs756211210 Chr.9: 32457313-32457313 on Build GRCh38
rs779912375 Chr.9: 32457322-32457322 on Build GRCh38
rs545273610 Chr.9: 32457595-32457595 on Build GRCh38
rs369784399 Chr.9: 32457545-32457545 on Build GRCh38
rs367791334 Chr.9: 32457446-32457446 on Build GRCh38
rs750339953 Chr.9: 32457628-32457628 on Build GRCh38
rs561308296 Chr.9: 32457704-32457704 on Build GRCh38
rs75266121 Chr.9: 32457570-32457570 on Build GRCh38
rs770577853 Chr.9: 32457366-32457366 on Build GRCh38
rs751722546 Chr.9: 32457303-32457303 on Build GRCh38
rs1053560539 Chr.9: 32457702-32457702 on Build GRCh38
rs756123387 Chr.9: 32457435-32457435 on Build GRCh38
rs755939681 Chr.9: 32457498-32457499 on Build GRCh38
rs776336045 Chr.9: 32457461-32457461 on Build GRCh38
rs932465912 Chr.9: 32457682-32457682 on Build GRCh38
rs201726738 Chr.9: 32457340-32457340 on Build GRCh38
rs757298640 Chr.9: 32457578-32457578 on Build GRCh38
rs147393239 Chr.9: 32457325-32457325 on Build GRCh38
rs757554872 Chr.9: 32457304-32457304 on Build GRCh38
rs750661371 Chr.9: 32457423-32457423 on Build GRCh38
rs753706911 Chr.9: 32457444-32457444 on Build GRCh38
rs750464561 Chr.9: 32457310-32457310 on Build GRCh38
rs763580057 Chr.9: 32457369-32457369 on Build GRCh38
rs7865082 Chr.9: 32457560-32457560 on Build GRCh38
rs1001271677 Chr.9: 32457515-32457515 on Build GRCh38
rs1035766326 Chr.9: 32457560-32457560 on Build GRCh38
rs921077620 Chr.9: 32457472-32457472 on Build GRCh38
rs377457419 Chr.9: 32457301-32457301 on Build GRCh38
rs968409852 Chr.9: 32457371-32457371 on Build GRCh38
rs536750872 Chr.9: 32457500-32457500 on Build GRCh38
rs377205390 Chr.9: 32457483-32457484 on Build GRCh38
rs1043304266 Chr.9: 32457733-32457733 on Build GRCh38
rs779766104 Chr.9: 32457325-32457325 on Build GRCh38
rs939951361 Chr.9: 32457718-32457718 on Build GRCh38
rs748933699 Chr.9: 32457324-32457325 on Build GRCh38
rs551051664 Chr.9: 32457483-32457483 on Build GRCh38
rs947663704 Chr.9: 32457705-32457705 on Build GRCh38
rs75836526 Chr.9: 32457590-32457590 on Build GRCh38
rs79312808 Chr.9: 32457493-32457493 on Build GRCh38
rs911044020 Chr.9: 32457394-32457394 on Build GRCh38
rs137979887 Chr.9: 32457457-32457457 on Build GRCh38
rs768635697 Chr.9: 32457441-32457442 on Build GRCh38
rs5897507 Chr.9: 32457493-32457494 on Build GRCh38
rs5897506 Chr.9: 32457492-32457493 on Build GRCh38
rs991743179 Chr.9: 32457364-32457364 on Build GRCh38
rs371400863 Chr.9: 32457460-32457460 on Build GRCh38
rs773027801 Chr.9: 32457360-32457360 on Build GRCh38
rs544397798 Chr.9: 32457689-32457689 on Build GRCh38
rs536518031 Chr.9: 32457566-32457566 on Build GRCh38
rs754726367 Chr.9: 32457324-32457324 on Build GRCh38
rs749232403 Chr.9: 32457323-32457323 on Build GRCh38
rs188115153 Chr.9: 32457660-32457660 on Build GRCh38
rs958888973 Chr.9: 32457353-32457353 on Build GRCh38
rs58139813 Chr.9: 32457502-32457503 on Build GRCh38
rs530068055 Chr.9: 32457725-32457725 on Build GRCh38
rs777508556 Chr.9: 32457456-32457456 on Build GRCh38
rs17289116 Chr.9: 32457531-32457531 on Build GRCh38
rs976483071 Chr.9: 32457665-32457665 on Build GRCh38

View Full Product Details