Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32480093-32480635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX58
Entrez Gene ID
23586
Gene Name
DEXD/H-box helicase 58
Gene Aliases
RIG-I, RIGI, RLR-1, SGMRT2
UniGene
Hs.190622
Gene Chromosome Location
Chr.9: 32455302-32526324 on Build GRCh38
Transcripts
NM_014314.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
487360 c.1745T>A p.I582N Chr.9: 32480248-32480248 on Build GRCh38
3763881 c.1740T>A p.D580E Chr.9: 32480253-32480253 on Build GRCh38
6250340 c.1462C>A p.R488R Chr.9: 32480318-32480318 on Build GRCh38
1108169 c.1774G>A p.E592K Chr.9: 32480219-32480219 on Build GRCh38
4676885 c.1747G>T p.E583* Chr.9: 32480246-32480246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779421098 Chr.9: 32480289-32480289 on Build GRCh38
rs952790004 Chr.9: 32480457-32480457 on Build GRCh38
rs775528857 Chr.9: 32480177-32480185 on Build GRCh38
rs776620678 Chr.9: 32480232-32480232 on Build GRCh38
rs371517752 Chr.9: 32480155-32480155 on Build GRCh38
rs766439817 Chr.9: 32480229-32480229 on Build GRCh38
rs564803893 Chr.9: 32480437-32480437 on Build GRCh38
rs894790121 Chr.9: 32480140-32480140 on Build GRCh38
rs112437427 Chr.9: 32480293-32480293 on Build GRCh38
rs771023122 Chr.9: 32480327-32480327 on Build GRCh38
rs148194904 Chr.9: 32480226-32480226 on Build GRCh38
rs765214948 Chr.9: 32480358-32480358 on Build GRCh38
rs911198891 Chr.9: 32480504-32480504 on Build GRCh38
rs746918120 Chr.9: 32480171-32480172 on Build GRCh38
rs377675778 Chr.9: 32480316-32480316 on Build GRCh38
rs554089688 Chr.9: 32480425-32480425 on Build GRCh38
rs922740392 Chr.9: 32480287-32480287 on Build GRCh38
rs774494016 Chr.9: 32480394-32480395 on Build GRCh38
rs116612738 Chr.9: 32480256-32480256 on Build GRCh38
rs17217280 Chr.9: 32480253-32480253 on Build GRCh38
rs181063624 Chr.9: 32480431-32480431 on Build GRCh38
rs758161240 Chr.9: 32480192-32480192 on Build GRCh38
rs377366976 Chr.9: 32480424-32480424 on Build GRCh38
rs769958143 Chr.9: 32480172-32480173 on Build GRCh38
rs564860697 Chr.9: 32480464-32480464 on Build GRCh38
rs777598207 Chr.9: 32480195-32480195 on Build GRCh38
rs34804468 Chr.9: 32480594-32480594 on Build GRCh38
rs767021348 Chr.9: 32480398-32480398 on Build GRCh38
rs759443243 Chr.9: 32480242-32480242 on Build GRCh38
rs773422076 Chr.9: 32480318-32480318 on Build GRCh38
rs770358009 Chr.9: 32480210-32480210 on Build GRCh38
rs1055330519 Chr.9: 32480126-32480126 on Build GRCh38
rs554887373 Chr.9: 32480383-32480383 on Build GRCh38
rs902212250 Chr.9: 32480466-32480466 on Build GRCh38
rs755440232 Chr.9: 32480275-32480275 on Build GRCh38
rs749487693 Chr.9: 32480483-32480483 on Build GRCh38
rs763777619 Chr.9: 32480255-32480255 on Build GRCh38
rs752378672 Chr.9: 32480190-32480190 on Build GRCh38
rs371685816 Chr.9: 32480245-32480245 on Build GRCh38
rs754803009 Chr.9: 32480179-32480179 on Build GRCh38
rs981912638 Chr.9: 32480127-32480127 on Build GRCh38
rs775577620 Chr.9: 32480368-32480368 on Build GRCh38
rs746464794 Chr.9: 32480209-32480209 on Build GRCh38
rs780543649 Chr.9: 32480211-32480211 on Build GRCh38
rs77681738 Chr.9: 32480144-32480144 on Build GRCh38
rs764045756 Chr.9: 32480379-32480379 on Build GRCh38
rs61756274 Chr.9: 32480218-32480218 on Build GRCh38
rs902224032 Chr.9: 32480157-32480157 on Build GRCh38
rs752734310 Chr.9: 32480248-32480248 on Build GRCh38
rs373010189 Chr.9: 32480403-32480403 on Build GRCh38
rs952857720 Chr.9: 32480239-32480239 on Build GRCh38
rs759689916 Chr.9: 32480347-32480347 on Build GRCh38
rs916786841 Chr.9: 32480450-32480450 on Build GRCh38
rs1044695909 Chr.9: 32480392-32480392 on Build GRCh38
rs756908920 Chr.9: 32480391-32480391 on Build GRCh38
rs748454578 Chr.9: 32480294-32480294 on Build GRCh38
rs371404578 Chr.9: 32480270-32480270 on Build GRCh38
rs1003385622 Chr.9: 32480409-32480409 on Build GRCh38
rs55895980 Chr.9: 32480586-32480586 on Build GRCh38
rs143242678 Chr.9: 32480577-32480577 on Build GRCh38
rs747257290 Chr.9: 32480326-32480326 on Build GRCh38
rs762197606 Chr.9: 32480222-32480222 on Build GRCh38
rs143007332 Chr.9: 32480224-32480224 on Build GRCh38
rs369892125 Chr.9: 32480337-32480337 on Build GRCh38
rs769049565 Chr.9: 32480215-32480215 on Build GRCh38
rs139025513 Chr.9: 32480588-32480588 on Build GRCh38
rs367656115 Chr.9: 32480267-32480267 on Build GRCh38
rs367851160 Chr.9: 32480213-32480213 on Build GRCh38
rs534359728 Chr.9: 32480408-32480408 on Build GRCh38
rs977023667 Chr.9: 32480536-32480536 on Build GRCh38
rs768901800 Chr.9: 32480346-32480346 on Build GRCh38
rs185443227 Chr.9: 32480531-32480531 on Build GRCh38
rs773436516 Chr.9: 32480225-32480225 on Build GRCh38
rs778911920 Chr.9: 32480189-32480189 on Build GRCh38
rs762973812 Chr.9: 32480195-32480197 on Build GRCh38

View Full Product Details