Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33255148-33255650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG1
Entrez Gene ID
573
Gene Name
BCL2 associated athanogene 1
Gene Aliases
BAG-1, HAP, RAP46
UniGene
Hs.377484
Gene Chromosome Location
Chr.9: 33252471-33264761 on Build GRCh38
Transcripts
NM_001172415.1 NM_004323.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108296 c.1031C>T p.A344V Chr.9: 33255226-33255226 on Build GRCh38
1108298 c.983A>G p.N328S Chr.9: 33255274-33255274 on Build GRCh38
6609556 c.1032C>T p.A344A Chr.9: 33255225-33255225 on Build GRCh38
79140 c.991C>T p.Q331* Chr.9: 33255266-33255266 on Build GRCh38
7095242 c.1013C>T p.S338F Chr.9: 33255244-33255244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998720828 Chr.9: 33255363-33255364 on Build GRCh38
rs374335975 Chr.9: 33255183-33255183 on Build GRCh38
rs773498482 Chr.9: 33255327-33255327 on Build GRCh38
rs776334161 Chr.9: 33255198-33255198 on Build GRCh38
rs760937452 Chr.9: 33255337-33255337 on Build GRCh38
rs550679681 Chr.9: 33255493-33255493 on Build GRCh38
rs566624527 Chr.9: 33255619-33255619 on Build GRCh38
rs952752751 Chr.9: 33255239-33255239 on Build GRCh38
rs567371786 Chr.9: 33255507-33255507 on Build GRCh38
rs774359426 Chr.9: 33255398-33255399 on Build GRCh38
rs746655235 Chr.9: 33255285-33255285 on Build GRCh38
rs150092738 Chr.9: 33255273-33255273 on Build GRCh38
rs530127187 Chr.9: 33255562-33255562 on Build GRCh38
rs778754721 Chr.9: 33255264-33255264 on Build GRCh38
rs767796673 Chr.9: 33255230-33255230 on Build GRCh38
rs777614373 Chr.9: 33255360-33255360 on Build GRCh38
rs763258012 Chr.9: 33255508-33255508 on Build GRCh38
rs952762814 Chr.9: 33255584-33255584 on Build GRCh38
rs960510988 Chr.9: 33255468-33255468 on Build GRCh38
rs369437844 Chr.9: 33255324-33255324 on Build GRCh38
rs11551682 Chr.9: 33255286-33255286 on Build GRCh38
rs137872180 Chr.9: 33255241-33255241 on Build GRCh38
rs372748609 Chr.9: 33255258-33255258 on Build GRCh38
rs369195280 Chr.9: 33255354-33255354 on Build GRCh38
rs144676599 Chr.9: 33255419-33255419 on Build GRCh38
rs777456409 Chr.9: 33255283-33255283 on Build GRCh38
rs545605969 Chr.9: 33255346-33255346 on Build GRCh38
rs758160223 Chr.9: 33255279-33255279 on Build GRCh38
rs573756922 Chr.9: 33255202-33255202 on Build GRCh38
rs763545045 Chr.9: 33255218-33255218 on Build GRCh38
rs758304555 Chr.9: 33255359-33255359 on Build GRCh38
rs757368992 Chr.9: 33255285-33255286 on Build GRCh38
rs752636879 Chr.9: 33255357-33255357 on Build GRCh38
rs776279085 Chr.9: 33255297-33255297 on Build GRCh38
rs1802039 Chr.9: 33255191-33255191 on Build GRCh38
rs375910579 Chr.9: 33255323-33255323 on Build GRCh38
rs769443085 Chr.9: 33255319-33255319 on Build GRCh38
rs138422440 Chr.9: 33255296-33255296 on Build GRCh38
rs765823981 Chr.9: 33255347-33255347 on Build GRCh38
rs563328921 Chr.9: 33255178-33255178 on Build GRCh38
rs376594007 Chr.9: 33255583-33255583 on Build GRCh38
rs896672403 Chr.9: 33255540-33255554 on Build GRCh38
rs764767280 Chr.9: 33255224-33255224 on Build GRCh38
rs745315597 Chr.9: 33255307-33255307 on Build GRCh38
rs746658604 Chr.9: 33255181-33255181 on Build GRCh38
rs142325367 Chr.9: 33255254-33255254 on Build GRCh38
rs775006645 Chr.9: 33255225-33255225 on Build GRCh38
rs368992536 Chr.9: 33255180-33255180 on Build GRCh38
rs1035936351 Chr.9: 33255459-33255459 on Build GRCh38
rs200553287 Chr.9: 33255281-33255281 on Build GRCh38
rs770432019 Chr.9: 33255184-33255184 on Build GRCh38
rs374860968 Chr.9: 33255353-33255353 on Build GRCh38
rs778663104 Chr.9: 33255168-33255168 on Build GRCh38
rs771764541 Chr.9: 33255179-33255179 on Build GRCh38
rs897821433 Chr.9: 33255266-33255266 on Build GRCh38
rs750619571 Chr.9: 33255232-33255232 on Build GRCh38
rs11789000 Chr.9: 33255485-33255485 on Build GRCh38
rs371923631 Chr.9: 33255328-33255328 on Build GRCh38
rs1002166562 Chr.9: 33255372-33255372 on Build GRCh38
rs754903807 Chr.9: 33255259-33255259 on Build GRCh38
rs766652384 Chr.9: 33255342-33255342 on Build GRCh38
rs367820745 Chr.9: 33255226-33255226 on Build GRCh38

View Full Product Details