Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34126269-34126768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF12
Entrez Gene ID
25853
Gene Name
DDB1 and CUL4 associated factor 12
Gene Aliases
CT102, KIAA1892, TCC52, WDR40A
UniGene
Hs.493750
Gene Chromosome Location
Chr.9: 34086383-34126773 on Build GRCh38
Transcripts
NM_015397.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
608683 c.44C>T p.S15L Chr.9: 34126388-34126388 on Build GRCh38
3670024 c.67C>A p.Q23K Chr.9: 34126365-34126365 on Build GRCh38
1665430 c.47_49delCGG p.P16_G17>R Chr.9: 34126383-34126385 on Build GRCh38
6990528 c.61G>C p.D21H Chr.9: 34126371-34126371 on Build GRCh38
1108410 c.77A>G p.Q26R Chr.9: 34126355-34126355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971063781 Chr.9: 34126712-34126712 on Build GRCh38
rs925714265 Chr.9: 34126464-34126464 on Build GRCh38
rs1038618280 Chr.9: 34126738-34126738 on Build GRCh38
rs767652039 Chr.9: 34126391-34126391 on Build GRCh38
rs752372983 Chr.9: 34126483-34126483 on Build GRCh38
rs988268475 Chr.9: 34126396-34126396 on Build GRCh38
rs766263445 Chr.9: 34126410-34126410 on Build GRCh38
rs551305007 Chr.9: 34126729-34126729 on Build GRCh38
rs756059807 Chr.9: 34126408-34126408 on Build GRCh38
rs760099412 Chr.9: 34126648-34126648 on Build GRCh38
rs754848458 Chr.9: 34126417-34126417 on Build GRCh38
rs763341102 Chr.9: 34126369-34126369 on Build GRCh38
rs997672690 Chr.9: 34126683-34126683 on Build GRCh38
rs746464917 Chr.9: 34126438-34126438 on Build GRCh38
rs759384003 Chr.9: 34126474-34126474 on Build GRCh38
rs968221296 Chr.9: 34126614-34126614 on Build GRCh38
rs367969182 Chr.9: 34126454-34126454 on Build GRCh38
rs777431326 Chr.9: 34126436-34126436 on Build GRCh38
rs909621866 Chr.9: 34126289-34126289 on Build GRCh38
rs1038431036 Chr.9: 34126534-34126534 on Build GRCh38
rs760784395 Chr.9: 34126469-34126469 on Build GRCh38
rs949966672 Chr.9: 34126497-34126497 on Build GRCh38
rs998016156 Chr.9: 34126665-34126665 on Build GRCh38
rs531089936 Chr.9: 34126727-34126727 on Build GRCh38
rs908471927 Chr.9: 34126734-34126734 on Build GRCh38
rs771763243 Chr.9: 34126348-34126348 on Build GRCh38
rs767507199 Chr.9: 34126631-34126631 on Build GRCh38
rs950944511 Chr.9: 34126715-34126715 on Build GRCh38
rs867798236 Chr.9: 34126690-34126690 on Build GRCh38
rs772841289 Chr.9: 34126356-34126356 on Build GRCh38
rs746377084 Chr.9: 34126357-34126357 on Build GRCh38
rs772225634 Chr.9: 34126465-34126465 on Build GRCh38
rs558981323 Chr.9: 34126595-34126595 on Build GRCh38
rs761928345 Chr.9: 34126388-34126388 on Build GRCh38
rs941173359 Chr.9: 34126735-34126735 on Build GRCh38
rs753870584 Chr.9: 34126473-34126473 on Build GRCh38
rs1029670974 Chr.9: 34126583-34126583 on Build GRCh38
rs764337097 Chr.9: 34126379-34126379 on Build GRCh38
rs1007561931 Chr.9: 34126652-34126652 on Build GRCh38
rs564911076 Chr.9: 34126673-34126673 on Build GRCh38
rs1017697599 Chr.9: 34126472-34126472 on Build GRCh38
rs747897847 Chr.9: 34126425-34126425 on Build GRCh38
rs776109707 Chr.9: 34126449-34126449 on Build GRCh38
rs899811729 Chr.9: 34126358-34126358 on Build GRCh38
rs765083411 Chr.9: 34126475-34126475 on Build GRCh38
rs950975811 Chr.9: 34126576-34126577 on Build GRCh38
rs1047455098 Chr.9: 34126499-34126499 on Build GRCh38
rs1026510436 Chr.9: 34126692-34126692 on Build GRCh38
rs747732957 Chr.9: 34126347-34126347 on Build GRCh38
rs544649879 Chr.9: 34126600-34126600 on Build GRCh38
rs1032730458 Chr.9: 34126642-34126642 on Build GRCh38
rs371477186 Chr.9: 34126565-34126565 on Build GRCh38
rs756625241 Chr.9: 34126372-34126372 on Build GRCh38
rs761849168 Chr.9: 34126394-34126394 on Build GRCh38
rs573545210 Chr.9: 34126532-34126532 on Build GRCh38
rs774686751 Chr.9: 34126385-34126385 on Build GRCh38
rs766096633 Chr.9: 34126658-34126658 on Build GRCh38
rs968651486 Chr.9: 34126431-34126431 on Build GRCh38
rs770459144 Chr.9: 34126444-34126444 on Build GRCh38
rs769125170 Chr.9: 34126455-34126455 on Build GRCh38
rs982699957 Chr.9: 34126730-34126730 on Build GRCh38
rs750015423 Chr.9: 34126637-34126637 on Build GRCh38
rs778665963 Chr.9: 34126333-34126333 on Build GRCh38
rs1056254585 Chr.9: 34126627-34126627 on Build GRCh38
rs754618279 Chr.9: 34126328-34126328 on Build GRCh38
rs865841066 Chr.9: 34126624-34126624 on Build GRCh38
rs753411532 Chr.9: 34126688-34126688 on Build GRCh38
rs572605823 Chr.9: 34126599-34126599 on Build GRCh38
rs886930571 Chr.9: 34126507-34126507 on Build GRCh38
rs770363311 Chr.9: 34126362-34126362 on Build GRCh38
rs939158439 Chr.9: 34126620-34126620 on Build GRCh38
rs980888855 Chr.9: 34126610-34126610 on Build GRCh38
rs917127130 Chr.9: 34126496-34126496 on Build GRCh38
rs778557229 Chr.9: 34126420-34126420 on Build GRCh38
rs749266587 Chr.9: 34126482-34126483 on Build GRCh38
rs1017461090 Chr.9: 34126307-34126307 on Build GRCh38
rs758054870 Chr.9: 34126427-34126427 on Build GRCh38
rs1009810649 Chr.9: 34126596-34126596 on Build GRCh38
rs774576896 Chr.9: 34126457-34126457 on Build GRCh38
rs371492912 Chr.9: 34126470-34126470 on Build GRCh38
rs955615754 Chr.9: 34126386-34126386 on Build GRCh38
rs991240521 Chr.9: 34126686-34126686 on Build GRCh38
rs941212259 Chr.9: 34126513-34126513 on Build GRCh38
rs901940872 Chr.9: 34126678-34126678 on Build GRCh38
rs891166908 Chr.9: 34126588-34126588 on Build GRCh38
rs776023183 Chr.9: 34126367-34126367 on Build GRCh38
rs773311518 Chr.9: 34126468-34126468 on Build GRCh38
rs892395452 Chr.9: 34126415-34126415 on Build GRCh38
rs996815191 Chr.9: 34126582-34126582 on Build GRCh38

View Full Product Details