Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34458684-34459221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI1
Entrez Gene ID
27019
Gene Name
dynein axonemal intermediate chain 1
Gene Aliases
CILD1, DIC1, ICS1, PCD
UniGene
Hs.112667
Gene Chromosome Location
Chr.9: 34458752-34520989 on Build GRCh38
Transcripts
NM_001281428.1 NM_012144.3 XM_011517846.1 XM_011517847.2 XM_011517848.1 XM_011517849.1 XM_011517850.2 XM_017014625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4588795 c.48+1_48+2insT p.? Chr.9: 34459054-34459055 on Build GRCh38
1108457 c.18G>A p.A6A Chr.9: 34459023-34459023 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766124282 Chr.9: 34458923-34458923 on Build GRCh38
rs886063880 Chr.9: 34458861-34458861 on Build GRCh38
rs776744499 Chr.9: 34458971-34458971 on Build GRCh38
rs758881186 Chr.9: 34459105-34459105 on Build GRCh38
rs112930490 Chr.9: 34459018-34459018 on Build GRCh38
rs750115296 Chr.9: 34459094-34459094 on Build GRCh38
rs952361785 Chr.9: 34458783-34458783 on Build GRCh38
rs146501326 Chr.9: 34459045-34459045 on Build GRCh38
rs11547035 Chr.9: 34459027-34459027 on Build GRCh38
rs753360934 Chr.9: 34459102-34459102 on Build GRCh38
rs927687791 Chr.9: 34458834-34458834 on Build GRCh38
rs771114105 Chr.9: 34458967-34458967 on Build GRCh38
rs55750068 Chr.9: 34458836-34458836 on Build GRCh38
rs376040025 Chr.9: 34459083-34459083 on Build GRCh38
rs962155265 Chr.9: 34459059-34459059 on Build GRCh38
rs769873119 Chr.9: 34459066-34459066 on Build GRCh38
rs3802429 Chr.9: 34458828-34458828 on Build GRCh38
rs991100226 Chr.9: 34458764-34458764 on Build GRCh38
rs755692713 Chr.9: 34459032-34459032 on Build GRCh38
rs1018967138 Chr.9: 34458748-34458748 on Build GRCh38
rs569994109 Chr.9: 34459134-34459134 on Build GRCh38
rs144238090 Chr.9: 34459145-34459145 on Build GRCh38
rs765871660 Chr.9: 34459099-34459099 on Build GRCh38
rs775621604 Chr.9: 34459069-34459069 on Build GRCh38
rs746021545 Chr.9: 34459062-34459062 on Build GRCh38
rs759637740 Chr.9: 34458994-34458994 on Build GRCh38
rs993549177 Chr.9: 34458988-34458988 on Build GRCh38
rs148701985 Chr.9: 34459052-34459052 on Build GRCh38
rs916785214 Chr.9: 34458765-34458765 on Build GRCh38
rs760391161 Chr.9: 34459096-34459096 on Build GRCh38
rs62619982 Chr.9: 34458890-34458890 on Build GRCh38
rs367785401 Chr.9: 34458996-34458996 on Build GRCh38
rs139318318 Chr.9: 34458716-34458716 on Build GRCh38
rs1021908908 Chr.9: 34458744-34458744 on Build GRCh38
rs549925663 Chr.9: 34459126-34459126 on Build GRCh38
rs775290934 Chr.9: 34458998-34458998 on Build GRCh38
rs939753519 Chr.9: 34458822-34458822 on Build GRCh38
rs1012658655 Chr.9: 34458972-34458972 on Build GRCh38
rs150858875 Chr.9: 34458704-34458704 on Build GRCh38
rs762864487 Chr.9: 34459071-34459071 on Build GRCh38
rs576807362 Chr.9: 34458777-34458777 on Build GRCh38
rs372656672 Chr.9: 34459033-34459033 on Build GRCh38
rs751365548 Chr.9: 34459020-34459020 on Build GRCh38
rs991767798 Chr.9: 34458797-34458797 on Build GRCh38
rs889707507 Chr.9: 34458855-34458855 on Build GRCh38
rs762886409 Chr.9: 34459012-34459012 on Build GRCh38
rs902624536 Chr.9: 34458879-34458879 on Build GRCh38
rs1032811844 Chr.9: 34458910-34458910 on Build GRCh38
rs1008916139 Chr.9: 34458728-34458728 on Build GRCh38
rs1040888048 Chr.9: 34458868-34458868 on Build GRCh38
rs943887532 Chr.9: 34458861-34458861 on Build GRCh38
rs928415321 Chr.9: 34458819-34458819 on Build GRCh38
rs397515363 Chr.9: 34459055-34459056 on Build GRCh38
rs367641912 Chr.9: 34459151-34459151 on Build GRCh38
rs753202925 Chr.9: 34459037-34459037 on Build GRCh38
rs865853678 Chr.9: 34459041-34459041 on Build GRCh38
rs781426827 Chr.9: 34459058-34459058 on Build GRCh38
rs958256105 Chr.9: 34458726-34458726 on Build GRCh38
rs966777975 Chr.9: 34458710-34458710 on Build GRCh38
rs771238788 Chr.9: 34459056-34459056 on Build GRCh38
rs767356011 Chr.9: 34459091-34459091 on Build GRCh38
rs180838420 Chr.9: 34458826-34458826 on Build GRCh38
rs1052059710 Chr.9: 34458847-34458847 on Build GRCh38
rs760335957 Chr.9: 34458920-34458920 on Build GRCh38
rs770833457 Chr.9: 34459149-34459149 on Build GRCh38
rs1032327039 Chr.9: 34458722-34458722 on Build GRCh38
rs946387190 Chr.9: 34459162-34459162 on Build GRCh38
rs547949645 Chr.9: 34459050-34459050 on Build GRCh38
rs750045804 Chr.9: 34459030-34459030 on Build GRCh38
rs930937761 Chr.9: 34458831-34458831 on Build GRCh38
rs371754078 Chr.9: 34458980-34458980 on Build GRCh38
rs970908850 Chr.9: 34458773-34458773 on Build GRCh38
rs34040638 Chr.9: 34458944-34458944 on Build GRCh38
rs548860209 Chr.9: 34458864-34458864 on Build GRCh38
rs867008978 Chr.9: 34459053-34459054 on Build GRCh38
rs768605985 Chr.9: 34459076-34459076 on Build GRCh38
rs763848127 Chr.9: 34459014-34459014 on Build GRCh38
rs200245894 Chr.9: 34458965-34458965 on Build GRCh38
rs1008672553 Chr.9: 34458893-34458893 on Build GRCh38
rs914871099 Chr.9: 34459157-34459157 on Build GRCh38

View Full Product Details