Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34568818-34569324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTFR
Entrez Gene ID
1271
Gene Name
ciliary neurotrophic factor receptor
Gene Aliases
-
UniGene
Hs.129966
Gene Chromosome Location
Chr.9: 34551432-34590386 on Build GRCh38
Transcripts
NM_001207011.1 NM_001842.4 NM_147164.2 XM_017014260.1 XM_017014261.1 XM_017014262.1 XM_017014263.1 XM_017014264.1 XM_017014265.1 NR_024369.1

Gene Symbol
CNTFR-AS1
Entrez Gene ID
415056
Gene Name
CNTFR antisense RNA 1
Gene Aliases
-
UniGene
Hs.734229
Gene Chromosome Location
Chr.9: 34568012-34583072 on Build GRCh38
Transcripts
NM_001207011.1 NM_001842.4 NM_147164.2 XM_017014260.1 XM_017014261.1 XM_017014262.1 XM_017014263.1 XM_017014264.1 XM_017014265.1 NR_024369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6458431 c.51_53delCGC p.A18delA Chr.9: 34568929-34568931 on Build GRCh38
5677986 c.15C>G p.V5V Chr.9: 34568967-34568967 on Build GRCh38
6208498 c.84G>T p.Q28H Chr.9: 34568898-34568898 on Build GRCh38
4981013 c.4G>T p.A2S Chr.9: 34568978-34568978 on Build GRCh38
1201645 c.64G>A p.A22T Chr.9: 34568918-34568918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374939382 Chr.9: 34569261-34569261 on Build GRCh38
rs753791400 Chr.9: 34568918-34568918 on Build GRCh38
rs566399574 Chr.9: 34569236-34569236 on Build GRCh38
rs1009641324 Chr.9: 34568912-34568912 on Build GRCh38
rs9695558 Chr.9: 34569012-34569012 on Build GRCh38
rs756354569 Chr.9: 34568910-34568910 on Build GRCh38
rs12000687 Chr.9: 34569299-34569299 on Build GRCh38
rs745332685 Chr.9: 34568936-34568936 on Build GRCh38
rs533501969 Chr.9: 34568999-34568999 on Build GRCh38
rs772421469 Chr.9: 34568974-34568974 on Build GRCh38
rs549824300 Chr.9: 34569095-34569095 on Build GRCh38
rs926823617 Chr.9: 34569228-34569228 on Build GRCh38
rs375307592 Chr.9: 34568868-34568868 on Build GRCh38
rs193287854 Chr.9: 34568885-34568885 on Build GRCh38
rs200157404 Chr.9: 34568995-34568995 on Build GRCh38
rs143644881 Chr.9: 34568934-34568934 on Build GRCh38
rs941295356 Chr.9: 34568865-34568865 on Build GRCh38
rs557564185 Chr.9: 34569291-34569291 on Build GRCh38
rs578047061 Chr.9: 34568882-34568882 on Build GRCh38
rs1002224705 Chr.9: 34568841-34568841 on Build GRCh38
rs982886702 Chr.9: 34568863-34568863 on Build GRCh38
rs146101081 Chr.9: 34568937-34568937 on Build GRCh38
rs573694260 Chr.9: 34568928-34568928 on Build GRCh38
rs10814124 Chr.9: 34569124-34569124 on Build GRCh38
rs770533170 Chr.9: 34569287-34569287 on Build GRCh38
rs993570030 Chr.9: 34569028-34569028 on Build GRCh38
rs772041907 Chr.9: 34569019-34569019 on Build GRCh38
rs376688767 Chr.9: 34568930-34568930 on Build GRCh38
rs773479532 Chr.9: 34568985-34568985 on Build GRCh38
rs1017222476 Chr.9: 34569049-34569049 on Build GRCh38
rs752681172 Chr.9: 34568998-34568998 on Build GRCh38
rs1026474283 Chr.9: 34568964-34568964 on Build GRCh38
rs938269072 Chr.9: 34569237-34569237 on Build GRCh38
rs369912296 Chr.9: 34568987-34568987 on Build GRCh38
rs762063501 Chr.9: 34568997-34568997 on Build GRCh38
rs141701455 Chr.9: 34568916-34568916 on Build GRCh38
rs975767060 Chr.9: 34569107-34569107 on Build GRCh38
rs1039048558 Chr.9: 34569117-34569117 on Build GRCh38
rs112178786 Chr.9: 34569110-34569110 on Build GRCh38
rs745433423 Chr.9: 34569015-34569015 on Build GRCh38
rs780760519 Chr.9: 34568935-34568935 on Build GRCh38
rs933732260 Chr.9: 34568933-34568933 on Build GRCh38
rs1057149719 Chr.9: 34569258-34569258 on Build GRCh38
rs750613000 Chr.9: 34568899-34568899 on Build GRCh38
rs1025474720 Chr.9: 34568877-34568877 on Build GRCh38
rs979627973 Chr.9: 34569177-34569177 on Build GRCh38
rs150343950 Chr.9: 34568911-34568911 on Build GRCh38
rs369923771 Chr.9: 34568939-34568939 on Build GRCh38
rs767776514 Chr.9: 34568884-34568884 on Build GRCh38
rs778671795 Chr.9: 34568919-34568919 on Build GRCh38
rs200191904 Chr.9: 34568940-34568940 on Build GRCh38
rs560581051 Chr.9: 34568929-34568931 on Build GRCh38
rs756967352 Chr.9: 34569008-34569008 on Build GRCh38
rs779462299 Chr.9: 34569020-34569020 on Build GRCh38
rs187533508 Chr.9: 34568996-34568996 on Build GRCh38
rs751247932 Chr.9: 34569003-34569003 on Build GRCh38
rs774818916 Chr.9: 34568965-34568965 on Build GRCh38
rs200141364 Chr.9: 34568869-34568869 on Build GRCh38
rs548955868 Chr.9: 34569172-34569172 on Build GRCh38
rs982531227 Chr.9: 34568970-34568970 on Build GRCh38
rs946853519 Chr.9: 34569164-34569164 on Build GRCh38
rs41274859 Chr.9: 34568859-34568859 on Build GRCh38
rs184670508 Chr.9: 34568931-34568931 on Build GRCh38
rs370113389 Chr.9: 34569129-34569129 on Build GRCh38
rs762205617 Chr.9: 34568972-34568972 on Build GRCh38
rs763996882 Chr.9: 34569002-34569002 on Build GRCh38
rs372635857 Chr.9: 34568906-34568906 on Build GRCh38
rs774731062 Chr.9: 34568878-34568878 on Build GRCh38
rs748640401 Chr.9: 34569031-34569031 on Build GRCh38
rs746562774 Chr.9: 34568848-34568848 on Build GRCh38
rs913665964 Chr.9: 34569144-34569144 on Build GRCh38
rs370378077 Chr.9: 34568940-34568942 on Build GRCh38
rs540121615 Chr.9: 34569275-34569275 on Build GRCh38
rs1005398728 Chr.9: 34569044-34569044 on Build GRCh38
rs774698156 Chr.9: 34568928-34568929 on Build GRCh38
rs913715952 Chr.9: 34569062-34569062 on Build GRCh38
rs759481918 Chr.9: 34568997-34568997 on Build GRCh38
rs777525169 Chr.9: 34569037-34569037 on Build GRCh38
rs943309796 Chr.9: 34569113-34569113 on Build GRCh38
rs1026426816 Chr.9: 34569034-34569034 on Build GRCh38

View Full Product Details