Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34581009-34581504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTFR
Entrez Gene ID
1271
Gene Name
ciliary neurotrophic factor receptor
Gene Aliases
-
UniGene
Hs.129966
Gene Chromosome Location
Chr.9: 34551432-34590386 on Build GRCh38
Transcripts
NM_001207011.1 NM_001842.4 NM_147164.2 XM_017014260.1 XM_017014261.1 XM_017014262.1 XM_017014263.1 XM_017014264.1 XM_017014265.1 NR_024369.1

Gene Symbol
CNTFR-AS1
Entrez Gene ID
415056
Gene Name
CNTFR antisense RNA 1
Gene Aliases
-
UniGene
Hs.734229
Gene Chromosome Location
Chr.9: 34568012-34583072 on Build GRCh38
Transcripts
NM_001207011.1 NM_001842.4 NM_147164.2 XM_017014260.1 XM_017014261.1 XM_017014262.1 XM_017014263.1 XM_017014264.1 XM_017014265.1 NR_024369.1

SNP Detail

SNP ID Chromosome Location
rs557777741 Chr.9: 34581420-34581420 on Build GRCh38
rs528390581 Chr.9: 34581432-34581432 on Build GRCh38
rs971028514 Chr.9: 34581199-34581199 on Build GRCh38
rs977958805 Chr.9: 34581254-34581254 on Build GRCh38
rs552405144 Chr.9: 34581114-34581114 on Build GRCh38
rs1008159496 Chr.9: 34581055-34581055 on Build GRCh38
rs1032499080 Chr.9: 34581126-34581126 on Build GRCh38
rs570212313 Chr.9: 34581040-34581040 on Build GRCh38
rs991384128 Chr.9: 34581258-34581258 on Build GRCh38
rs570614889 Chr.9: 34581332-34581332 on Build GRCh38
rs748643520 Chr.9: 34581073-34581073 on Build GRCh38
rs977833243 Chr.9: 34581099-34581099 on Build GRCh38
rs907849649 Chr.9: 34581419-34581419 on Build GRCh38
rs889692850 Chr.9: 34581031-34581031 on Build GRCh38
rs536048705 Chr.9: 34581186-34581186 on Build GRCh38
rs185542414 Chr.9: 34581442-34581442 on Build GRCh38
rs922006108 Chr.9: 34581378-34581378 on Build GRCh38
rs917864479 Chr.9: 34581133-34581133 on Build GRCh38
rs534773388 Chr.9: 34581417-34581417 on Build GRCh38
rs566199383 Chr.9: 34581409-34581409 on Build GRCh38
rs775180166 Chr.9: 34581063-34581063 on Build GRCh38
rs990472639 Chr.9: 34581131-34581133 on Build GRCh38
rs899225807 Chr.9: 34581469-34581469 on Build GRCh38
rs959061876 Chr.9: 34581124-34581124 on Build GRCh38
rs549281967 Chr.9: 34581392-34581392 on Build GRCh38
rs916761188 Chr.9: 34581332-34581332 on Build GRCh38
rs949615212 Chr.9: 34581377-34581377 on Build GRCh38
rs923728780 Chr.9: 34581304-34581304 on Build GRCh38
rs957836073 Chr.9: 34581150-34581150 on Build GRCh38
rs966369073 Chr.9: 34581076-34581076 on Build GRCh38
rs746421416 Chr.9: 34581053-34581053 on Build GRCh38
rs1026734470 Chr.9: 34581120-34581120 on Build GRCh38
rs192670349 Chr.9: 34581039-34581039 on Build GRCh38
rs1037749066 Chr.9: 34581435-34581435 on Build GRCh38

View Full Product Details