Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34726122-34726626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM205A
Entrez Gene ID
259308
Gene Name
family with sequence similarity 205 member A
Gene Aliases
C9orf144B
UniGene
Hs.522093
Gene Chromosome Location
Chr.9: 34723053-34729538 on Build GRCh38
Transcripts
NM_001141917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991147 c.760T>C p.L254L Chr.9: 34726480-34726480 on Build GRCh38
4163812 c.706G>C p.E236Q Chr.9: 34726534-34726534 on Build GRCh38
4163810 c.1050T>C p.H350H Chr.9: 34726190-34726190 on Build GRCh38
5932393 c.751C>T p.Q251* Chr.9: 34726489-34726489 on Build GRCh38
2773592 c.1035T>C p.P345P Chr.9: 34726205-34726205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975757135 Chr.9: 34726433-34726433 on Build GRCh38
rs998580754 Chr.9: 34726368-34726368 on Build GRCh38
rs749613399 Chr.9: 34726364-34726364 on Build GRCh38
rs1055818857 Chr.9: 34726590-34726590 on Build GRCh38
rs2937382 Chr.9: 34726421-34726421 on Build GRCh38
rs563987580 Chr.9: 34726345-34726345 on Build GRCh38
rs776313462 Chr.9: 34726526-34726526 on Build GRCh38
rs766388235 Chr.9: 34726532-34726534 on Build GRCh38
rs7467715 Chr.9: 34726357-34726357 on Build GRCh38
rs76045500 Chr.9: 34726158-34726158 on Build GRCh38
rs537237239 Chr.9: 34726555-34726555 on Build GRCh38
rs521611 Chr.9: 34726190-34726190 on Build GRCh38
rs1013752548 Chr.9: 34726310-34726310 on Build GRCh38
rs781706268 Chr.9: 34726260-34726260 on Build GRCh38
rs938023309 Chr.9: 34726581-34726581 on Build GRCh38
rs1021098492 Chr.9: 34726313-34726313 on Build GRCh38
rs3895393 Chr.9: 34726562-34726562 on Build GRCh38
rs756544302 Chr.9: 34726304-34726304 on Build GRCh38
rs774320420 Chr.9: 34726164-34726164 on Build GRCh38
rs543688204 Chr.9: 34726302-34726302 on Build GRCh38
rs2988119 Chr.9: 34726567-34726567 on Build GRCh38
rs868804006 Chr.9: 34726213-34726213 on Build GRCh38
rs756633986 Chr.9: 34726578-34726578 on Build GRCh38
rs772268298 Chr.9: 34726431-34726431 on Build GRCh38
rs551061684 Chr.9: 34726487-34726487 on Build GRCh38
rs368271218 Chr.9: 34726432-34726432 on Build GRCh38
rs764847312 Chr.9: 34726528-34726528 on Build GRCh38
rs908361899 Chr.9: 34726527-34726527 on Build GRCh38
rs780574308 Chr.9: 34726597-34726597 on Build GRCh38
rs748195461 Chr.9: 34726596-34726596 on Build GRCh38
rs1038441771 Chr.9: 34726537-34726537 on Build GRCh38
rs145034991 Chr.9: 34726473-34726473 on Build GRCh38
rs758299649 Chr.9: 34726423-34726423 on Build GRCh38
rs761917296 Chr.9: 34726173-34726173 on Build GRCh38
rs879783584 Chr.9: 34726214-34726214 on Build GRCh38
rs763596841 Chr.9: 34726564-34726564 on Build GRCh38
rs521630 Chr.9: 34726185-34726185 on Build GRCh38
rs377672072 Chr.9: 34726549-34726549 on Build GRCh38
rs768796301 Chr.9: 34726388-34726388 on Build GRCh38
rs2988120 Chr.9: 34726605-34726605 on Build GRCh38
rs1810820 Chr.9: 34726527-34726527 on Build GRCh38
rs878949485 Chr.9: 34726424-34726424 on Build GRCh38
rs752188908 Chr.9: 34726239-34726239 on Build GRCh38
rs774605174 Chr.9: 34726392-34726392 on Build GRCh38
rs521574 Chr.9: 34726207-34726207 on Build GRCh38
rs746367419 Chr.9: 34726268-34726268 on Build GRCh38
rs757880347 Chr.9: 34726245-34726245 on Build GRCh38
rs112032167 Chr.9: 34726459-34726459 on Build GRCh38
rs564617921 Chr.9: 34726205-34726205 on Build GRCh38
rs921186085 Chr.9: 34726441-34726441 on Build GRCh38

View Full Product Details