Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35075201-35075711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
UniGene
Hs.591084
Gene Chromosome Location
Chr.9: 35073838-35080016 on Build GRCh38
Transcripts
NM_004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6295281 c.1341C>A p.Y447* Chr.9: 35075557-35075557 on Build GRCh38
1108585 c.1441G>A p.E481K Chr.9: 35075318-35075318 on Build GRCh38
260836 c.1474G>T p.E492* Chr.9: 35075285-35075285 on Build GRCh38
1108587 c.1351T>C p.W451R Chr.9: 35075547-35075547 on Build GRCh38
94270 c.1286C>T p.P429L Chr.9: 35075612-35075612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187315404 Chr.9: 35075456-35075456 on Build GRCh38
rs757442131 Chr.9: 35075552-35075552 on Build GRCh38
rs748730134 Chr.9: 35075682-35075682 on Build GRCh38
rs756069860 Chr.9: 35075544-35075544 on Build GRCh38
rs769402230 Chr.9: 35075490-35075490 on Build GRCh38
rs368631340 Chr.9: 35075375-35075375 on Build GRCh38
rs764116943 Chr.9: 35075545-35075545 on Build GRCh38
rs777880901 Chr.9: 35075441-35075441 on Build GRCh38
rs191508370 Chr.9: 35075570-35075570 on Build GRCh38
rs764032365 Chr.9: 35075621-35075621 on Build GRCh38
rs767398356 Chr.9: 35075373-35075374 on Build GRCh38
rs376961485 Chr.9: 35075444-35075444 on Build GRCh38
rs897901007 Chr.9: 35075573-35075573 on Build GRCh38
rs200456325 Chr.9: 35075566-35075566 on Build GRCh38
rs372341195 Chr.9: 35075369-35075369 on Build GRCh38
rs746164729 Chr.9: 35075558-35075558 on Build GRCh38
rs948298405 Chr.9: 35075420-35075420 on Build GRCh38
rs587778345 Chr.9: 35075532-35075532 on Build GRCh38
rs762631021 Chr.9: 35075499-35075499 on Build GRCh38
rs772794286 Chr.9: 35075587-35075587 on Build GRCh38
rs758665937 Chr.9: 35075650-35075650 on Build GRCh38
rs201884798 Chr.9: 35075306-35075306 on Build GRCh38
rs375682263 Chr.9: 35075297-35075297 on Build GRCh38
rs758259248 Chr.9: 35075246-35075246 on Build GRCh38
rs945983673 Chr.9: 35075645-35075645 on Build GRCh38
rs764992166 Chr.9: 35075239-35075239 on Build GRCh38
rs772460728 Chr.9: 35075318-35075318 on Build GRCh38
rs750585976 Chr.9: 35075639-35075639 on Build GRCh38
rs149616199 Chr.9: 35075278-35075278 on Build GRCh38
rs761391172 Chr.9: 35075341-35075341 on Build GRCh38
rs758656355 Chr.9: 35075560-35075560 on Build GRCh38
rs764902323 Chr.9: 35075347-35075347 on Build GRCh38
rs760537318 Chr.9: 35075619-35075619 on Build GRCh38
rs1041369619 Chr.9: 35075689-35075689 on Build GRCh38
rs765394242 Chr.9: 35075631-35075631 on Build GRCh38
rs772542885 Chr.9: 35075457-35075457 on Build GRCh38
rs927127258 Chr.9: 35075478-35075478 on Build GRCh38
rs754635557 Chr.9: 35075576-35075576 on Build GRCh38
rs752700756 Chr.9: 35075434-35075434 on Build GRCh38
rs753917194 Chr.9: 35075548-35075548 on Build GRCh38
rs371171892 Chr.9: 35075676-35075676 on Build GRCh38
rs765965051 Chr.9: 35075512-35075512 on Build GRCh38
rs757276792 Chr.9: 35075630-35075630 on Build GRCh38
rs886063896 Chr.9: 35075646-35075646 on Build GRCh38
rs767253119 Chr.9: 35075611-35075611 on Build GRCh38
rs371809395 Chr.9: 35075665-35075665 on Build GRCh38
rs753829462 Chr.9: 35075629-35075629 on Build GRCh38
rs759285011 Chr.9: 35075606-35075606 on Build GRCh38
rs778986343 Chr.9: 35075557-35075557 on Build GRCh38
rs756155858 Chr.9: 35075435-35075435 on Build GRCh38
rs776133588 Chr.9: 35075475-35075475 on Build GRCh38
rs904322133 Chr.9: 35075322-35075322 on Build GRCh38
rs758423821 Chr.9: 35075682-35075682 on Build GRCh38
rs767385265 Chr.9: 35075533-35075533 on Build GRCh38
rs183777537 Chr.9: 35075601-35075601 on Build GRCh38
rs985426777 Chr.9: 35075270-35075270 on Build GRCh38
rs889128525 Chr.9: 35075635-35075635 on Build GRCh38
rs748738986 Chr.9: 35075600-35075600 on Build GRCh38
rs769921972 Chr.9: 35075286-35075286 on Build GRCh38
rs1018027137 Chr.9: 35075285-35075285 on Build GRCh38
rs201070684 Chr.9: 35075453-35075453 on Build GRCh38
rs747392982 Chr.9: 35075661-35075661 on Build GRCh38
rs148808709 Chr.9: 35075531-35075531 on Build GRCh38
rs766157946 Chr.9: 35075468-35075468 on Build GRCh38
rs780502064 Chr.9: 35075660-35075660 on Build GRCh38
rs989531823 Chr.9: 35075396-35075396 on Build GRCh38
rs779750311 Chr.9: 35075255-35075255 on Build GRCh38
rs752882385 Chr.9: 35075679-35075680 on Build GRCh38
rs751287895 Chr.9: 35075526-35075526 on Build GRCh38
rs771305176 Chr.9: 35075302-35075302 on Build GRCh38
rs374096159 Chr.9: 35075272-35075272 on Build GRCh38
rs753317485 Chr.9: 35075235-35075235 on Build GRCh38
rs980822010 Chr.9: 35075470-35075470 on Build GRCh38
rs781764361 Chr.9: 35075679-35075679 on Build GRCh38
rs754889211 Chr.9: 35075417-35075417 on Build GRCh38
rs767426960 Chr.9: 35075264-35075264 on Build GRCh38
rs200078931 Chr.9: 35075419-35075419 on Build GRCh38
rs775293827 Chr.9: 35075612-35075612 on Build GRCh38
rs781269744 Chr.9: 35075276-35075276 on Build GRCh38
rs906662045 Chr.9: 35075485-35075485 on Build GRCh38
rs368664759 Chr.9: 35075362-35075362 on Build GRCh38
rs17881054 Chr.9: 35075610-35075610 on Build GRCh38
rs772883935 Chr.9: 35075492-35075492 on Build GRCh38
rs994955867 Chr.9: 35075597-35075597 on Build GRCh38
rs754927660 Chr.9: 35075321-35075321 on Build GRCh38
rs776146510 Chr.9: 35075319-35075319 on Build GRCh38
rs769293655 Chr.9: 35075578-35075578 on Build GRCh38
rs780612204 Chr.9: 35075563-35075563 on Build GRCh38
rs756902343 Chr.9: 35075236-35075236 on Build GRCh38
rs77152961 Chr.9: 35075305-35075305 on Build GRCh38
rs935110627 Chr.9: 35075379-35075379 on Build GRCh38
rs17882757 Chr.9: 35075348-35075348 on Build GRCh38
rs144369518 Chr.9: 35075476-35075476 on Build GRCh38
rs779268127 Chr.9: 35075454-35075454 on Build GRCh38
rs777969388 Chr.9: 35075294-35075294 on Build GRCh38

View Full Product Details