Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35561468-35561975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM166B
Entrez Gene ID
730112
Gene Name
family with sequence similarity 166 member B
Gene Aliases
-
UniGene
Hs.98959
Gene Chromosome Location
Chr.9: 35561830-35563899 on Build GRCh38
Transcripts
NM_001099951.3 NM_001164310.2 NM_001287238.1 NM_001287239.1 XM_011518028.2 NM_001135999.1 NM_014806.3 NR_052015.2 XM_006716896.2

Gene Symbol
RUSC2
Entrez Gene ID
9853
Gene Name
RUN and SH3 domain containing 2
Gene Aliases
Iporin
UniGene
Hs.493796
Gene Chromosome Location
Chr.9: 35487999-35561898 on Build GRCh38
Transcripts
NM_001099951.3 NM_001164310.2 NM_001287238.1 NM_001287239.1 XM_011518028.2 NM_001135999.1 NM_014806.3 NR_052015.2 XM_006716896.2

SNP Detail

SNP ID Chromosome Location
rs886501327 Chr.9: 35561841-35561844 on Build GRCh38
rs753808616 Chr.9: 35561923-35561923 on Build GRCh38
rs760590380 Chr.9: 35561921-35561921 on Build GRCh38
rs1005416548 Chr.9: 35561822-35561822 on Build GRCh38
rs4878620 Chr.9: 35561547-35561547 on Build GRCh38
rs1044383366 Chr.9: 35561561-35561561 on Build GRCh38
rs962653456 Chr.9: 35561873-35561874 on Build GRCh38
rs1018847242 Chr.9: 35561552-35561565 on Build GRCh38
rs1046419 Chr.9: 35561633-35561633 on Build GRCh38
rs974810221 Chr.9: 35561565-35561565 on Build GRCh38
rs771051127 Chr.9: 35561840-35561841 on Build GRCh38
rs767272643 Chr.9: 35561844-35561844 on Build GRCh38
rs992305682 Chr.9: 35561733-35561733 on Build GRCh38
rs781749892 Chr.9: 35561944-35561944 on Build GRCh38
rs1039215005 Chr.9: 35561805-35561805 on Build GRCh38
rs954770491 Chr.9: 35561882-35561882 on Build GRCh38
rs899751681 Chr.9: 35561830-35561830 on Build GRCh38
rs763323260 Chr.9: 35561935-35561936 on Build GRCh38
rs886926648 Chr.9: 35561804-35561804 on Build GRCh38
rs111468168 Chr.9: 35561631-35561631 on Build GRCh38
rs1016878968 Chr.9: 35561542-35561542 on Build GRCh38
rs759988433 Chr.9: 35561907-35561908 on Build GRCh38
rs1043361303 Chr.9: 35561767-35561767 on Build GRCh38
rs1046869957 Chr.9: 35561785-35561785 on Build GRCh38
rs960468238 Chr.9: 35561729-35561729 on Build GRCh38
rs913645173 Chr.9: 35561916-35561916 on Build GRCh38
rs565870013 Chr.9: 35561495-35561495 on Build GRCh38
rs575369646 Chr.9: 35561931-35561938 on Build GRCh38
rs904092353 Chr.9: 35561587-35561587 on Build GRCh38
rs757003933 Chr.9: 35561942-35561944 on Build GRCh38
rs3209520 Chr.9: 35561914-35561914 on Build GRCh38
rs1056172399 Chr.9: 35561511-35561511 on Build GRCh38
rs753292158 Chr.9: 35561931-35561932 on Build GRCh38
rs1055469835 Chr.9: 35561753-35561753 on Build GRCh38
rs143266743 Chr.9: 35561916-35561923 on Build GRCh38
rs1017281368 Chr.9: 35561680-35561680 on Build GRCh38
rs72725074 Chr.9: 35561770-35561770 on Build GRCh38
rs987570259 Chr.9: 35561594-35561594 on Build GRCh38
rs980607744 Chr.9: 35561718-35561718 on Build GRCh38
rs534899414 Chr.9: 35561496-35561496 on Build GRCh38
rs191910543 Chr.9: 35561681-35561681 on Build GRCh38
rs954799576 Chr.9: 35561582-35561582 on Build GRCh38
rs967942308 Chr.9: 35561943-35561943 on Build GRCh38
rs1046415 Chr.9: 35561630-35561630 on Build GRCh38
rs539409417 Chr.9: 35561768-35561768 on Build GRCh38
rs761835109 Chr.9: 35561927-35561927 on Build GRCh38
rs777189853 Chr.9: 35561849-35561850 on Build GRCh38
rs533641698 Chr.9: 35561810-35561813 on Build GRCh38
rs535684708 Chr.9: 35561909-35561909 on Build GRCh38
rs1028963033 Chr.9: 35561877-35561877 on Build GRCh38
rs926260171 Chr.9: 35561674-35561674 on Build GRCh38
rs996546473 Chr.9: 35561851-35561851 on Build GRCh38
rs752021036 Chr.9: 35561939-35561939 on Build GRCh38
rs751004227 Chr.9: 35561936-35561937 on Build GRCh38
rs755512065 Chr.9: 35561942-35561942 on Build GRCh38
rs758615504 Chr.9: 35561932-35561932 on Build GRCh38
rs149325726 Chr.9: 35561540-35561540 on Build GRCh38
rs901967689 Chr.9: 35561614-35561614 on Build GRCh38
rs368058292 Chr.9: 35561799-35561799 on Build GRCh38
rs984778850 Chr.9: 35561670-35561670 on Build GRCh38
rs979412133 Chr.9: 35561669-35561669 on Build GRCh38
rs117264391 Chr.9: 35561829-35561829 on Build GRCh38
rs950762532 Chr.9: 35561660-35561660 on Build GRCh38
rs1002103144 Chr.9: 35561878-35561879 on Build GRCh38
rs963698803 Chr.9: 35561559-35561559 on Build GRCh38
rs901998955 Chr.9: 35561828-35561828 on Build GRCh38
rs945057129 Chr.9: 35561766-35561766 on Build GRCh38
rs780453753 Chr.9: 35561935-35561935 on Build GRCh38
rs555903249 Chr.9: 35561922-35561922 on Build GRCh38
rs1033542475 Chr.9: 35561900-35561900 on Build GRCh38
rs1028994217 Chr.9: 35561570-35561570 on Build GRCh38
rs765250532 Chr.9: 35561931-35561931 on Build GRCh38
rs748684788 Chr.9: 35561946-35561946 on Build GRCh38
rs926440820 Chr.9: 35561725-35561725 on Build GRCh38
rs913622804 Chr.9: 35561765-35561765 on Build GRCh38
rs1017695666 Chr.9: 35561605-35561605 on Build GRCh38
rs775326648 Chr.9: 35561919-35561919 on Build GRCh38

View Full Product Details