Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35563683-35564192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM166B
Entrez Gene ID
730112
Gene Name
family with sequence similarity 166 member B
Gene Aliases
-
UniGene
Hs.98959
Gene Chromosome Location
Chr.9: 35561830-35563899 on Build GRCh38
Transcripts
NM_001099951.3 NM_001164310.2 NM_001287238.1 NM_001287239.1 XM_011518028.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6567678 c.26C>T p.P9L Chr.9: 35563803-35563803 on Build GRCh38
6567679 c.26C>T p.P9L Chr.9: 35563803-35563803 on Build GRCh38
7149451 c.33C>G p.L11L Chr.9: 35563796-35563796 on Build GRCh38
7149450 c.33C>G p.L11L Chr.9: 35563796-35563796 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs552950777 Chr.9: 35564042-35564042 on Build GRCh38
rs1032119043 Chr.9: 35563927-35563927 on Build GRCh38
rs538343471 Chr.9: 35563904-35563904 on Build GRCh38
rs928647190 Chr.9: 35564041-35564041 on Build GRCh38
rs560863714 Chr.9: 35564061-35564061 on Build GRCh38
rs551997998 Chr.9: 35563729-35563729 on Build GRCh38
rs1008202711 Chr.9: 35564006-35564006 on Build GRCh38
rs745850494 Chr.9: 35563897-35563897 on Build GRCh38
rs558189376 Chr.9: 35563932-35563932 on Build GRCh38
rs367907663 Chr.9: 35563837-35563837 on Build GRCh38
rs920071065 Chr.9: 35563879-35563879 on Build GRCh38
rs368657738 Chr.9: 35563742-35563742 on Build GRCh38
rs748125077 Chr.9: 35563819-35563819 on Build GRCh38
rs142036794 Chr.9: 35564119-35564119 on Build GRCh38
rs376031481 Chr.9: 35563741-35563741 on Build GRCh38
rs552669999 Chr.9: 35563844-35563844 on Build GRCh38
rs200737115 Chr.9: 35563758-35563758 on Build GRCh38
rs1049889606 Chr.9: 35564133-35564133 on Build GRCh38
rs767933442 Chr.9: 35563863-35563863 on Build GRCh38
rs776375446 Chr.9: 35563817-35563817 on Build GRCh38
rs774742852 Chr.9: 35563852-35563852 on Build GRCh38
rs753078266 Chr.9: 35563867-35563867 on Build GRCh38
rs981412502 Chr.9: 35563993-35563993 on Build GRCh38
rs536277575 Chr.9: 35563816-35563816 on Build GRCh38
rs570739099 Chr.9: 35563887-35563887 on Build GRCh38
rs893260774 Chr.9: 35563955-35563956 on Build GRCh38
rs201821532 Chr.9: 35564066-35564069 on Build GRCh38
rs750518328 Chr.9: 35563750-35563750 on Build GRCh38
rs759767470 Chr.9: 35563855-35563855 on Build GRCh38
rs567934395 Chr.9: 35563792-35563792 on Build GRCh38
rs758489662 Chr.9: 35564056-35564056 on Build GRCh38
rs372361136 Chr.9: 35563748-35563748 on Build GRCh38
rs934910697 Chr.9: 35564146-35564146 on Build GRCh38
rs911444276 Chr.9: 35564164-35564164 on Build GRCh38
rs756709500 Chr.9: 35563869-35563869 on Build GRCh38
rs971926521 Chr.9: 35564046-35564046 on Build GRCh38
rs1049961782 Chr.9: 35563888-35563888 on Build GRCh38
rs940535585 Chr.9: 35563791-35563791 on Build GRCh38
rs902235166 Chr.9: 35563924-35563924 on Build GRCh38
rs559484900 Chr.9: 35564140-35564140 on Build GRCh38
rs1040698852 Chr.9: 35563905-35563905 on Build GRCh38
rs999662804 Chr.9: 35563925-35563925 on Build GRCh38
rs1045370091 Chr.9: 35564111-35564111 on Build GRCh38
rs879167092 Chr.9: 35563980-35563980 on Build GRCh38
rs1054542909 Chr.9: 35563951-35563951 on Build GRCh38
rs1019284759 Chr.9: 35564038-35564038 on Build GRCh38
rs543036790 Chr.9: 35564125-35564125 on Build GRCh38
rs920059143 Chr.9: 35564101-35564101 on Build GRCh38
rs1007812179 Chr.9: 35563892-35563892 on Build GRCh38
rs1012125773 Chr.9: 35563974-35563974 on Build GRCh38
rs769824610 Chr.9: 35563966-35563966 on Build GRCh38
rs528626874 Chr.9: 35564157-35564157 on Build GRCh38
rs746768133 Chr.9: 35563806-35563806 on Build GRCh38
rs758103087 Chr.9: 35563789-35563789 on Build GRCh38
rs915634983 Chr.9: 35564064-35564064 on Build GRCh38
rs951800258 Chr.9: 35564060-35564060 on Build GRCh38
rs58086772 Chr.9: 35564061-35564064 on Build GRCh38
rs116514144 Chr.9: 35563889-35563889 on Build GRCh38
rs766745754 Chr.9: 35563847-35563847 on Build GRCh38
rs575098456 Chr.9: 35563981-35563981 on Build GRCh38
rs764712911 Chr.9: 35563875-35563875 on Build GRCh38
rs903396251 Chr.9: 35563942-35563942 on Build GRCh38
rs999436923 Chr.9: 35563943-35563943 on Build GRCh38
rs551301899 Chr.9: 35563761-35563761 on Build GRCh38
rs995984736 Chr.9: 35564073-35564086 on Build GRCh38
rs920424816 Chr.9: 35564060-35564063 on Build GRCh38
rs796107529 Chr.9: 35564065-35564068 on Build GRCh38
rs377093854 Chr.9: 35563749-35563749 on Build GRCh38
rs372175228 Chr.9: 35563840-35563840 on Build GRCh38

View Full Product Details