Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35674033-35674539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA9
Entrez Gene ID
768
Gene Name
carbonic anhydrase 9
Gene Aliases
CAIX, MN
UniGene
Hs.63287
Gene Chromosome Location
Chr.9: 35673859-35681159 on Build GRCh38
Transcripts
NM_001216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3907190 c.261G>T p.E87D Chr.9: 35674220-35674220 on Build GRCh38
1462241 c.306_308delAGA p.E105delE Chr.9: 35674265-35674267 on Build GRCh38
1331559 c.388C>A p.H130N Chr.9: 35674347-35674347 on Build GRCh38
3763913 c.248T>C p.L83P Chr.9: 35674207-35674207 on Build GRCh38
4668249 c.230C>A p.P77Q Chr.9: 35674189-35674189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898356516 Chr.9: 35674411-35674411 on Build GRCh38
rs576502164 Chr.9: 35674416-35674416 on Build GRCh38
rs770545982 Chr.9: 35674092-35674092 on Build GRCh38
rs747536059 Chr.9: 35674068-35674068 on Build GRCh38
rs768612217 Chr.9: 35674389-35674389 on Build GRCh38
rs566948142 Chr.9: 35674140-35674140 on Build GRCh38
rs763624956 Chr.9: 35674209-35674209 on Build GRCh38
rs936887789 Chr.9: 35674362-35674362 on Build GRCh38
rs933692915 Chr.9: 35674205-35674205 on Build GRCh38
rs111602815 Chr.9: 35674181-35674181 on Build GRCh38
rs775277745 Chr.9: 35674214-35674214 on Build GRCh38
rs149032230 Chr.9: 35674420-35674420 on Build GRCh38
rs894470198 Chr.9: 35674443-35674443 on Build GRCh38
rs764592232 Chr.9: 35674288-35674288 on Build GRCh38
rs771768132 Chr.9: 35674343-35674343 on Build GRCh38
rs776744031 Chr.9: 35674224-35674224 on Build GRCh38
rs774190884 Chr.9: 35674096-35674096 on Build GRCh38
rs775063591 Chr.9: 35674231-35674231 on Build GRCh38
rs764007278 Chr.9: 35674220-35674220 on Build GRCh38
rs762183510 Chr.9: 35674398-35674398 on Build GRCh38
rs374065818 Chr.9: 35674227-35674227 on Build GRCh38
rs945461951 Chr.9: 35674302-35674302 on Build GRCh38
rs762605788 Chr.9: 35674091-35674091 on Build GRCh38
rs200022684 Chr.9: 35674189-35674189 on Build GRCh38
rs536591247 Chr.9: 35674117-35674117 on Build GRCh38
rs371355808 Chr.9: 35674264-35674264 on Build GRCh38
rs1034748837 Chr.9: 35674078-35674078 on Build GRCh38
rs867995116 Chr.9: 35674176-35674176 on Build GRCh38
rs377058222 Chr.9: 35674142-35674142 on Build GRCh38
rs908302251 Chr.9: 35674228-35674228 on Build GRCh38
rs975715331 Chr.9: 35674180-35674180 on Build GRCh38
rs368118579 Chr.9: 35674257-35674257 on Build GRCh38
rs912676212 Chr.9: 35674097-35674097 on Build GRCh38
rs759048994 Chr.9: 35674167-35674167 on Build GRCh38
rs755506506 Chr.9: 35674279-35674279 on Build GRCh38
rs781591028 Chr.9: 35674282-35674282 on Build GRCh38
rs780592140 Chr.9: 35674158-35674158 on Build GRCh38
rs764177293 Chr.9: 35674130-35674130 on Build GRCh38
rs200651095 Chr.9: 35674084-35674084 on Build GRCh38
rs201260414 Chr.9: 35674194-35674211 on Build GRCh38
rs12553173 Chr.9: 35674104-35674104 on Build GRCh38
rs146966274 Chr.9: 35674207-35674207 on Build GRCh38
rs1055207058 Chr.9: 35674418-35674418 on Build GRCh38
rs201022974 Chr.9: 35674308-35674308 on Build GRCh38
rs868176243 Chr.9: 35674212-35674212 on Build GRCh38
rs765168649 Chr.9: 35674404-35674404 on Build GRCh38
rs773923872 Chr.9: 35674203-35674203 on Build GRCh38
rs777101594 Chr.9: 35674177-35674194 on Build GRCh38
rs113670057 Chr.9: 35674188-35674188 on Build GRCh38
rs548199907 Chr.9: 35674054-35674054 on Build GRCh38
rs753926184 Chr.9: 35674133-35674133 on Build GRCh38
rs752590786 Chr.9: 35674125-35674125 on Build GRCh38
rs761837635 Chr.9: 35674396-35674396 on Build GRCh38
rs757499361 Chr.9: 35674135-35674135 on Build GRCh38
rs867137318 Chr.9: 35674506-35674506 on Build GRCh38
rs371183397 Chr.9: 35674170-35674170 on Build GRCh38
rs1002510278 Chr.9: 35674414-35674414 on Build GRCh38
rs565650695 Chr.9: 35674062-35674062 on Build GRCh38
rs775198289 Chr.9: 35674103-35674103 on Build GRCh38
rs137897809 Chr.9: 35674211-35674211 on Build GRCh38
rs751238550 Chr.9: 35674212-35674228 on Build GRCh38
rs974976527 Chr.9: 35674233-35674233 on Build GRCh38
rs750638464 Chr.9: 35674245-35674245 on Build GRCh38
rs113042654 Chr.9: 35674194-35674194 on Build GRCh38
rs745465512 Chr.9: 35674312-35674312 on Build GRCh38
rs760184777 Chr.9: 35674184-35674184 on Build GRCh38
rs181213293 Chr.9: 35674301-35674301 on Build GRCh38
rs746690273 Chr.9: 35674234-35674234 on Build GRCh38
rs2071676 Chr.9: 35674056-35674056 on Build GRCh38
rs765644887 Chr.9: 35674190-35674225 on Build GRCh38
rs747450063 Chr.9: 35674164-35674164 on Build GRCh38
rs113003874 Chr.9: 35674105-35674105 on Build GRCh38
rs142602359 Chr.9: 35674299-35674299 on Build GRCh38
rs143777802 Chr.9: 35674143-35674143 on Build GRCh38
rs748814740 Chr.9: 35674192-35674192 on Build GRCh38
rs769385538 Chr.9: 35674083-35674083 on Build GRCh38
rs77984049 Chr.9: 35674350-35674350 on Build GRCh38
rs895424346 Chr.9: 35674436-35674436 on Build GRCh38
rs867759516 Chr.9: 35674453-35674453 on Build GRCh38
rs545510618 Chr.9: 35674483-35674483 on Build GRCh38
rs967501960 Chr.9: 35674105-35674105 on Build GRCh38
rs201318433 Chr.9: 35674261-35674261 on Build GRCh38
rs776379775 Chr.9: 35674391-35674391 on Build GRCh38
rs1014269055 Chr.9: 35674500-35674500 on Build GRCh38
rs746817775 Chr.9: 35674364-35674364 on Build GRCh38
rs10972555 Chr.9: 35674193-35674193 on Build GRCh38
rs762377640 Chr.9: 35674193-35674194 on Build GRCh38
rs112632019 Chr.9: 35674178-35674178 on Build GRCh38
rs760594473 Chr.9: 35674218-35674218 on Build GRCh38
rs368343205 Chr.9: 35674121-35674121 on Build GRCh38
rs771788431 Chr.9: 35674204-35674204 on Build GRCh38
rs554959947 Chr.9: 35674278-35674278 on Build GRCh38
rs760610911 Chr.9: 35674129-35674129 on Build GRCh38

View Full Product Details