Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35679743-35680243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA9
Entrez Gene ID
768
Gene Name
carbonic anhydrase 9
Gene Aliases
CAIX, MN
UniGene
Hs.63287
Gene Chromosome Location
Chr.9: 35673859-35681159 on Build GRCh38
Transcripts
NM_001216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295444 c.1210G>A p.V404I Chr.9: 35679998-35679998 on Build GRCh38
4668252 c.1109G>A p.R370Q Chr.9: 35679897-35679897 on Build GRCh38
3848473 c.1127G>A p.R376Q Chr.9: 35679915-35679915 on Build GRCh38
2774396 c.1134G>A p.T378T Chr.9: 35679922-35679922 on Build GRCh38
5469078 c.1085C>T p.T362I Chr.9: 35679873-35679873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761778575 Chr.9: 35679922-35679922 on Build GRCh38
rs140019091 Chr.9: 35679941-35679941 on Build GRCh38
rs762888024 Chr.9: 35680019-35680019 on Build GRCh38
rs770274384 Chr.9: 35679897-35679897 on Build GRCh38
rs781425479 Chr.9: 35679865-35679865 on Build GRCh38
rs774396470 Chr.9: 35680147-35680147 on Build GRCh38
rs764546729 Chr.9: 35680170-35680170 on Build GRCh38
rs766449363 Chr.9: 35679938-35679938 on Build GRCh38
rs959555340 Chr.9: 35680036-35680036 on Build GRCh38
rs779679584 Chr.9: 35680096-35680096 on Build GRCh38
rs751673884 Chr.9: 35679991-35679991 on Build GRCh38
rs574121097 Chr.9: 35679790-35679790 on Build GRCh38
rs927548454 Chr.9: 35679785-35679785 on Build GRCh38
rs749798307 Chr.9: 35680084-35680084 on Build GRCh38
rs778061487 Chr.9: 35679863-35679866 on Build GRCh38
rs200431510 Chr.9: 35680103-35680103 on Build GRCh38
rs746066001 Chr.9: 35680027-35680028 on Build GRCh38
rs950111013 Chr.9: 35680089-35680089 on Build GRCh38
rs765108061 Chr.9: 35680018-35680018 on Build GRCh38
rs992130160 Chr.9: 35680048-35680048 on Build GRCh38
rs565441140 Chr.9: 35679918-35679918 on Build GRCh38
rs370076702 Chr.9: 35680085-35680085 on Build GRCh38
rs199769254 Chr.9: 35680023-35680023 on Build GRCh38
rs982761174 Chr.9: 35680215-35680215 on Build GRCh38
rs748678761 Chr.9: 35679882-35679882 on Build GRCh38
rs369391332 Chr.9: 35680095-35680095 on Build GRCh38
rs778093331 Chr.9: 35679973-35679973 on Build GRCh38
rs963516962 Chr.9: 35680028-35680028 on Build GRCh38
rs762875688 Chr.9: 35680137-35680137 on Build GRCh38
rs781572936 Chr.9: 35680018-35680018 on Build GRCh38
rs372703008 Chr.9: 35680216-35680216 on Build GRCh38
rs575743710 Chr.9: 35679921-35679921 on Build GRCh38
rs543329798 Chr.9: 35679782-35679782 on Build GRCh38
rs768232317 Chr.9: 35680002-35680002 on Build GRCh38
rs761462108 Chr.9: 35680017-35680017 on Build GRCh38
rs945781812 Chr.9: 35679800-35679800 on Build GRCh38
rs1039569215 Chr.9: 35679916-35679916 on Build GRCh38
rs760967813 Chr.9: 35680169-35680169 on Build GRCh38
rs553434374 Chr.9: 35679862-35679862 on Build GRCh38
rs754293793 Chr.9: 35680177-35680177 on Build GRCh38
rs767667342 Chr.9: 35680158-35680158 on Build GRCh38
rs768231579 Chr.9: 35680100-35680100 on Build GRCh38
rs780197280 Chr.9: 35679861-35679861 on Build GRCh38
rs573552495 Chr.9: 35679863-35679863 on Build GRCh38
rs750378648 Chr.9: 35679928-35679928 on Build GRCh38
rs765132128 Chr.9: 35679925-35679925 on Build GRCh38
rs757345104 Chr.9: 35680017-35680018 on Build GRCh38
rs746654610 Chr.9: 35679999-35679999 on Build GRCh38
rs763934915 Chr.9: 35679919-35679919 on Build GRCh38
rs771642912 Chr.9: 35679984-35679984 on Build GRCh38
rs781619865 Chr.9: 35679946-35679946 on Build GRCh38
rs770082000 Chr.9: 35680096-35680098 on Build GRCh38
rs756394763 Chr.9: 35680070-35680070 on Build GRCh38
rs775245763 Chr.9: 35679915-35679915 on Build GRCh38
rs748506137 Chr.9: 35679969-35679969 on Build GRCh38
rs916944625 Chr.9: 35679796-35679796 on Build GRCh38
rs746566853 Chr.9: 35680099-35680099 on Build GRCh38
rs973614167 Chr.9: 35679769-35679776 on Build GRCh38
rs750508367 Chr.9: 35679804-35679804 on Build GRCh38
rs758429038 Chr.9: 35679935-35679935 on Build GRCh38
rs936960992 Chr.9: 35679791-35679791 on Build GRCh38
rs759485908 Chr.9: 35680151-35680151 on Build GRCh38
rs758593162 Chr.9: 35679813-35679813 on Build GRCh38
rs756622774 Chr.9: 35679971-35679971 on Build GRCh38
rs1057188442 Chr.9: 35679792-35679792 on Build GRCh38
rs373161931 Chr.9: 35679828-35679828 on Build GRCh38
rs778288486 Chr.9: 35680075-35680075 on Build GRCh38
rs752797845 Chr.9: 35680162-35680162 on Build GRCh38
rs561778043 Chr.9: 35680050-35680050 on Build GRCh38
rs999987305 Chr.9: 35679913-35679913 on Build GRCh38
rs529821355 Chr.9: 35680047-35680047 on Build GRCh38
rs143639449 Chr.9: 35680127-35680127 on Build GRCh38
rs202231122 Chr.9: 35679905-35679905 on Build GRCh38
rs752986687 Chr.9: 35680063-35680063 on Build GRCh38
rs747838263 Chr.9: 35680116-35680116 on Build GRCh38
rs766347301 Chr.9: 35680139-35680139 on Build GRCh38
rs1009637891 Chr.9: 35679948-35679948 on Build GRCh38
rs778855846 Chr.9: 35680203-35680203 on Build GRCh38
rs536633148 Chr.9: 35679803-35679803 on Build GRCh38
rs376967634 Chr.9: 35679983-35679983 on Build GRCh38
rs963548115 Chr.9: 35679768-35679768 on Build GRCh38
rs775194948 Chr.9: 35679994-35679994 on Build GRCh38
rs766435358 Chr.9: 35680020-35680020 on Build GRCh38
rs769415658 Chr.9: 35680126-35680126 on Build GRCh38
rs201537277 Chr.9: 35679914-35679914 on Build GRCh38
rs776104167 Chr.9: 35680109-35680109 on Build GRCh38
rs200795260 Chr.9: 35680007-35680007 on Build GRCh38

View Full Product Details