Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35680020-35680553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA9
Entrez Gene ID
768
Gene Name
carbonic anhydrase 9
Gene Aliases
CAIX, MN
UniGene
Hs.63287
Gene Chromosome Location
Chr.9: 35673859-35681159 on Build GRCh38
Transcripts
NM_001216.2

SNP Detail

SNP ID Chromosome Location
rs1004905260 Chr.9: 35680370-35680371 on Build GRCh38
rs3750432 Chr.9: 35680373-35680373 on Build GRCh38
rs935783344 Chr.9: 35680283-35680283 on Build GRCh38
rs139468542 Chr.9: 35680372-35680373 on Build GRCh38
rs774396470 Chr.9: 35680147-35680147 on Build GRCh38
rs764546729 Chr.9: 35680170-35680170 on Build GRCh38
rs779679584 Chr.9: 35680096-35680096 on Build GRCh38
rs903620911 Chr.9: 35680355-35680355 on Build GRCh38
rs921341066 Chr.9: 35680510-35680510 on Build GRCh38
rs68054422 Chr.9: 35680366-35680367 on Build GRCh38
rs925722729 Chr.9: 35680241-35680241 on Build GRCh38
rs796989156 Chr.9: 35680372-35680373 on Build GRCh38
rs749798307 Chr.9: 35680084-35680084 on Build GRCh38
rs950111013 Chr.9: 35680089-35680089 on Build GRCh38
rs200431510 Chr.9: 35680103-35680103 on Build GRCh38
rs958936145 Chr.9: 35680388-35680388 on Build GRCh38
rs950667080 Chr.9: 35680426-35680426 on Build GRCh38
rs1038553838 Chr.9: 35680279-35680279 on Build GRCh38
rs992130160 Chr.9: 35680048-35680048 on Build GRCh38
rs370076702 Chr.9: 35680085-35680085 on Build GRCh38
rs933893747 Chr.9: 35680341-35680341 on Build GRCh38
rs982761174 Chr.9: 35680215-35680215 on Build GRCh38
rs975166985 Chr.9: 35680507-35680507 on Build GRCh38
rs369391332 Chr.9: 35680095-35680095 on Build GRCh38
rs2789751 Chr.9: 35680361-35680361 on Build GRCh38
rs566883388 Chr.9: 35680499-35680499 on Build GRCh38
rs933844127 Chr.9: 35680516-35680516 on Build GRCh38
rs750307903 Chr.9: 35680309-35680309 on Build GRCh38
rs762875688 Chr.9: 35680137-35680137 on Build GRCh38
rs558975600 Chr.9: 35680505-35680505 on Build GRCh38
rs372703008 Chr.9: 35680216-35680216 on Build GRCh38
rs554696762 Chr.9: 35680467-35680467 on Build GRCh38
rs552873681 Chr.9: 35680364-35680365 on Build GRCh38
rs3750431 Chr.9: 35680340-35680340 on Build GRCh38
rs1024561580 Chr.9: 35680409-35680409 on Build GRCh38
rs993453478 Chr.9: 35680395-35680395 on Build GRCh38
rs760967813 Chr.9: 35680169-35680169 on Build GRCh38
rs974193738 Chr.9: 35680478-35680478 on Build GRCh38
rs754293793 Chr.9: 35680177-35680177 on Build GRCh38
rs768231579 Chr.9: 35680100-35680100 on Build GRCh38
rs767667342 Chr.9: 35680158-35680158 on Build GRCh38
rs567633858 Chr.9: 35680448-35680448 on Build GRCh38
rs977534500 Chr.9: 35680235-35680235 on Build GRCh38
rs544174737 Chr.9: 35680370-35680370 on Build GRCh38
rs967424041 Chr.9: 35680412-35680412 on Build GRCh38
rs539294553 Chr.9: 35680502-35680502 on Build GRCh38
rs756394763 Chr.9: 35680070-35680070 on Build GRCh38
rs770082000 Chr.9: 35680096-35680098 on Build GRCh38
rs757981392 Chr.9: 35680527-35680527 on Build GRCh38
rs774367226 Chr.9: 35680422-35680422 on Build GRCh38
rs746566853 Chr.9: 35680099-35680099 on Build GRCh38
rs992390610 Chr.9: 35680418-35680418 on Build GRCh38
rs559732144 Chr.9: 35680298-35680298 on Build GRCh38
rs533427123 Chr.9: 35680425-35680425 on Build GRCh38
rs759485908 Chr.9: 35680151-35680151 on Build GRCh38
rs899962652 Chr.9: 35680292-35680292 on Build GRCh38
rs778288486 Chr.9: 35680075-35680075 on Build GRCh38
rs532232846 Chr.9: 35680327-35680327 on Build GRCh38
rs752797845 Chr.9: 35680162-35680162 on Build GRCh38
rs561778043 Chr.9: 35680050-35680050 on Build GRCh38
rs529821355 Chr.9: 35680047-35680047 on Build GRCh38
rs143639449 Chr.9: 35680127-35680127 on Build GRCh38
rs752986687 Chr.9: 35680063-35680063 on Build GRCh38
rs747838263 Chr.9: 35680116-35680116 on Build GRCh38
rs766347301 Chr.9: 35680139-35680139 on Build GRCh38
rs1033389898 Chr.9: 35680372-35680372 on Build GRCh38
rs778855846 Chr.9: 35680203-35680203 on Build GRCh38
rs781313469 Chr.9: 35680313-35680313 on Build GRCh38
rs1051031717 Chr.9: 35680365-35680365 on Build GRCh38
rs982792409 Chr.9: 35680440-35680440 on Build GRCh38
rs769640260 Chr.9: 35680421-35680422 on Build GRCh38
rs890414754 Chr.9: 35680369-35680370 on Build GRCh38
rs3833699 Chr.9: 35680379-35680380 on Build GRCh38
rs769415658 Chr.9: 35680126-35680126 on Build GRCh38
rs139705549 Chr.9: 35680466-35680466 on Build GRCh38
rs1034512630 Chr.9: 35680373-35680373 on Build GRCh38
rs185638034 Chr.9: 35680371-35680371 on Build GRCh38
rs767508521 Chr.9: 35680226-35680226 on Build GRCh38
rs776104167 Chr.9: 35680109-35680109 on Build GRCh38

View Full Product Details