Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35816963-35817509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM221B
Entrez Gene ID
392307
Gene Name
family with sequence similarity 221 member B
Gene Aliases
C9orf128
UniGene
Hs.128075
Gene Chromosome Location
Chr.9: 35817017-35829148 on Build GRCh38
Transcripts
NM_001012446.3 NR_052026.1

SNP Detail

SNP ID Chromosome Location
rs967811414 Chr.9: 35817327-35817327 on Build GRCh38
rs578032276 Chr.9: 35816996-35816996 on Build GRCh38
rs887198308 Chr.9: 35817140-35817140 on Build GRCh38
rs1035615899 Chr.9: 35817170-35817172 on Build GRCh38
rs1003686426 Chr.9: 35817137-35817137 on Build GRCh38
rs955048821 Chr.9: 35817280-35817280 on Build GRCh38
rs75163289 Chr.9: 35817175-35817175 on Build GRCh38
rs895825062 Chr.9: 35817111-35817111 on Build GRCh38
rs1021962727 Chr.9: 35817200-35817200 on Build GRCh38
rs749736915 Chr.9: 35817176-35817176 on Build GRCh38
rs997072979 Chr.9: 35817236-35817236 on Build GRCh38
rs540383597 Chr.9: 35817070-35817070 on Build GRCh38
rs967746790 Chr.9: 35817207-35817207 on Build GRCh38
rs184876276 Chr.9: 35817425-35817425 on Build GRCh38
rs139434729 Chr.9: 35817292-35817292 on Build GRCh38
rs745949680 Chr.9: 35817013-35817013 on Build GRCh38
rs549659921 Chr.9: 35817078-35817078 on Build GRCh38
rs775856935 Chr.9: 35817278-35817278 on Build GRCh38
rs887219009 Chr.9: 35817440-35817440 on Build GRCh38
rs563689854 Chr.9: 35817032-35817032 on Build GRCh38
rs146746216 Chr.9: 35817273-35817273 on Build GRCh38
rs551982711 Chr.9: 35817188-35817188 on Build GRCh38
rs780615229 Chr.9: 35817096-35817096 on Build GRCh38
rs950071378 Chr.9: 35817435-35817435 on Build GRCh38
rs77741309 Chr.9: 35817346-35817346 on Build GRCh38
rs747119961 Chr.9: 35817347-35817347 on Build GRCh38
rs770860960 Chr.9: 35817450-35817450 on Build GRCh38
rs1021147823 Chr.9: 35817322-35817322 on Build GRCh38
rs567208888 Chr.9: 35817316-35817316 on Build GRCh38
rs1047038556 Chr.9: 35817436-35817436 on Build GRCh38
rs1006156232 Chr.9: 35817156-35817156 on Build GRCh38
rs571785466 Chr.9: 35817199-35817199 on Build GRCh38
rs1047625868 Chr.9: 35817129-35817129 on Build GRCh38
rs144206363 Chr.9: 35817185-35817185 on Build GRCh38
rs992086422 Chr.9: 35817363-35817363 on Build GRCh38
rs117092705 Chr.9: 35817027-35817027 on Build GRCh38
rs1017333331 Chr.9: 35817283-35817283 on Build GRCh38
rs965035357 Chr.9: 35817468-35817468 on Build GRCh38
rs772310812 Chr.9: 35817339-35817339 on Build GRCh38
rs964149607 Chr.9: 35817214-35817214 on Build GRCh38
rs941440411 Chr.9: 35817062-35817062 on Build GRCh38
rs1014232793 Chr.9: 35817114-35817114 on Build GRCh38
rs78660413 Chr.9: 35817174-35817174 on Build GRCh38
rs543148008 Chr.9: 35817211-35817211 on Build GRCh38
rs577283396 Chr.9: 35817180-35817180 on Build GRCh38
rs181239335 Chr.9: 35817345-35817345 on Build GRCh38
rs769065230 Chr.9: 35817195-35817195 on Build GRCh38
rs975914613 Chr.9: 35817326-35817326 on Build GRCh38

View Full Product Details