Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35828833-35829324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM221B
Entrez Gene ID
392307
Gene Name
family with sequence similarity 221 member B
Gene Aliases
C9orf128
UniGene
Hs.128075
Gene Chromosome Location
Chr.9: 35817017-35829148 on Build GRCh38
Transcripts
XM_006716768.3 NM_001042589.2 NM_016446.3 XM_011517910.2 XM_011517911.2 XM_017014806.1

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
XM_006716768.3 NM_001042589.2 NM_016446.3 XM_011517910.2 XM_011517911.2 XM_017014806.1

SNP Detail

SNP ID Chromosome Location
rs900695588 Chr.9: 35828943-35828943 on Build GRCh38
rs997577564 Chr.9: 35829282-35829282 on Build GRCh38
rs891934858 Chr.9: 35829289-35829289 on Build GRCh38
rs368931074 Chr.9: 35828911-35828911 on Build GRCh38
rs1039163102 Chr.9: 35829249-35829249 on Build GRCh38
rs1015928824 Chr.9: 35828878-35828878 on Build GRCh38
rs147413670 Chr.9: 35829065-35829065 on Build GRCh38
rs1010522922 Chr.9: 35829293-35829293 on Build GRCh38
rs993495950 Chr.9: 35829240-35829244 on Build GRCh38
rs938547449 Chr.9: 35829070-35829070 on Build GRCh38
rs555428629 Chr.9: 35829170-35829170 on Build GRCh38
rs1047742670 Chr.9: 35828885-35828885 on Build GRCh38
rs1054551500 Chr.9: 35829121-35829121 on Build GRCh38
rs766260299 Chr.9: 35829006-35829006 on Build GRCh38
rs1036923875 Chr.9: 35829197-35829197 on Build GRCh38
rs927030670 Chr.9: 35829068-35829068 on Build GRCh38
rs900600955 Chr.9: 35829266-35829266 on Build GRCh38
rs992633715 Chr.9: 35829080-35829080 on Build GRCh38
rs897064766 Chr.9: 35828860-35828860 on Build GRCh38
rs929867577 Chr.9: 35828868-35828868 on Build GRCh38
rs916119766 Chr.9: 35829015-35829015 on Build GRCh38
rs1051935556 Chr.9: 35829286-35829286 on Build GRCh38
rs1008026305 Chr.9: 35829193-35829193 on Build GRCh38
rs942005356 Chr.9: 35829239-35829239 on Build GRCh38
rs1049548370 Chr.9: 35829140-35829140 on Build GRCh38
rs968783950 Chr.9: 35829064-35829064 on Build GRCh38
rs909253064 Chr.9: 35829237-35829237 on Build GRCh38
rs1021300330 Chr.9: 35829062-35829062 on Build GRCh38
rs879284 Chr.9: 35829034-35829034 on Build GRCh38
rs539475737 Chr.9: 35829039-35829039 on Build GRCh38
rs918397737 Chr.9: 35829096-35829096 on Build GRCh38
rs932438512 Chr.9: 35829137-35829137 on Build GRCh38
rs952104586 Chr.9: 35829261-35829261 on Build GRCh38
rs372890105 Chr.9: 35829061-35829061 on Build GRCh38
rs534260100 Chr.9: 35828908-35828908 on Build GRCh38
rs568716263 Chr.9: 35828898-35828898 on Build GRCh38
rs1023061556 Chr.9: 35829274-35829274 on Build GRCh38
rs981656114 Chr.9: 35829048-35829048 on Build GRCh38
rs778332868 Chr.9: 35828971-35828971 on Build GRCh38
rs914315165 Chr.9: 35829136-35829136 on Build GRCh38
rs1028103181 Chr.9: 35829252-35829252 on Build GRCh38
rs929914246 Chr.9: 35829222-35829222 on Build GRCh38
rs1048780971 Chr.9: 35829229-35829229 on Build GRCh38
rs1030473501 Chr.9: 35829032-35829032 on Build GRCh38
rs113130018 Chr.9: 35829162-35829162 on Build GRCh38
rs571427341 Chr.9: 35828865-35828865 on Build GRCh38
rs897083855 Chr.9: 35829236-35829236 on Build GRCh38
rs149713059 Chr.9: 35828924-35828924 on Build GRCh38
rs991647351 Chr.9: 35829009-35829009 on Build GRCh38

View Full Product Details