Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36191041-36191554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLTA
Entrez Gene ID
1211
Gene Name
clathrin light chain A
Gene Aliases
LCA
UniGene
Hs.522114
Gene Chromosome Location
Chr.9: 36190856-36212062 on Build GRCh38
Transcripts
NM_001076677.2 NM_001184760.1 NM_001184761.1 NM_001184762.1 NM_001311203.1 NM_001311204.1 NM_001311205.1 NM_001311206.1 NM_001833.3 NM_007096.3 NR_132349.1 XM_017014257.1 XM_017014258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6239693 c.138C>T p.I46I Chr.9: 36191194-36191194 on Build GRCh38
1569271 c.96T>C p.A32A Chr.9: 36191152-36191152 on Build GRCh38
5450585 c.108G>T p.L36F Chr.9: 36191164-36191164 on Build GRCh38
1718431 c.17C>T p.P6L Chr.9: 36191073-36191073 on Build GRCh38
6569187 c.156C>G p.F52L Chr.9: 36191212-36191212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021257331 Chr.9: 36191077-36191077 on Build GRCh38
rs908765516 Chr.9: 36191192-36191192 on Build GRCh38
rs563812223 Chr.9: 36191194-36191194 on Build GRCh38
rs550984188 Chr.9: 36191415-36191415 on Build GRCh38
rs1055303135 Chr.9: 36191452-36191452 on Build GRCh38
rs778934164 Chr.9: 36191184-36191184 on Build GRCh38
rs768841179 Chr.9: 36191128-36191128 on Build GRCh38
rs529517129 Chr.9: 36191245-36191245 on Build GRCh38
rs763731631 Chr.9: 36191312-36191312 on Build GRCh38
rs757143986 Chr.9: 36191177-36191177 on Build GRCh38
rs1009373129 Chr.9: 36191419-36191419 on Build GRCh38
rs751085255 Chr.9: 36191148-36191148 on Build GRCh38
rs982963714 Chr.9: 36191515-36191515 on Build GRCh38
rs765757524 Chr.9: 36191248-36191248 on Build GRCh38
rs761150114 Chr.9: 36191131-36191131 on Build GRCh38
rs1001186388 Chr.9: 36191517-36191517 on Build GRCh38
rs896194161 Chr.9: 36191076-36191076 on Build GRCh38
rs544059900 Chr.9: 36191164-36191164 on Build GRCh38
rs1000817690 Chr.9: 36191319-36191319 on Build GRCh38
rs898690659 Chr.9: 36191268-36191268 on Build GRCh38
rs747057246 Chr.9: 36191097-36191097 on Build GRCh38
rs755773670 Chr.9: 36191161-36191161 on Build GRCh38
rs186683951 Chr.9: 36191284-36191284 on Build GRCh38
rs967673950 Chr.9: 36191088-36191088 on Build GRCh38
rs752194151 Chr.9: 36191295-36191295 on Build GRCh38
rs749236110 Chr.9: 36191107-36191107 on Build GRCh38
rs964552930 Chr.9: 36191333-36191333 on Build GRCh38
rs192843693 Chr.9: 36191357-36191357 on Build GRCh38
rs954957019 Chr.9: 36191064-36191064 on Build GRCh38
rs938127398 Chr.9: 36191451-36191451 on Build GRCh38
rs751064824 Chr.9: 36191061-36191061 on Build GRCh38
rs908182349 Chr.9: 36191341-36191341 on Build GRCh38
rs937526849 Chr.9: 36191254-36191254 on Build GRCh38
rs189945560 Chr.9: 36191493-36191493 on Build GRCh38
rs971191033 Chr.9: 36191503-36191503 on Build GRCh38
rs757244515 Chr.9: 36191077-36191088 on Build GRCh38
rs781518113 Chr.9: 36191076-36191077 on Build GRCh38
rs776865961 Chr.9: 36191232-36191232 on Build GRCh38
rs2274524 Chr.9: 36191152-36191152 on Build GRCh38
rs147208924 Chr.9: 36191337-36191337 on Build GRCh38
rs769438385 Chr.9: 36191094-36191094 on Build GRCh38
rs1032147307 Chr.9: 36191082-36191082 on Build GRCh38
rs899752988 Chr.9: 36191376-36191376 on Build GRCh38
rs1033328368 Chr.9: 36191075-36191075 on Build GRCh38
rs770311992 Chr.9: 36191459-36191460 on Build GRCh38
rs996674910 Chr.9: 36191377-36191377 on Build GRCh38
rs908571405 Chr.9: 36191388-36191388 on Build GRCh38
rs890554823 Chr.9: 36191417-36191417 on Build GRCh38
rs11555991 Chr.9: 36191170-36191170 on Build GRCh38
rs564423777 Chr.9: 36191122-36191122 on Build GRCh38
rs917648908 Chr.9: 36191456-36191456 on Build GRCh38
rs1038281082 Chr.9: 36191374-36191374 on Build GRCh38
rs1051335613 Chr.9: 36191408-36191408 on Build GRCh38
rs763421913 Chr.9: 36191279-36191279 on Build GRCh38
rs991652295 Chr.9: 36191280-36191280 on Build GRCh38
rs184854801 Chr.9: 36191444-36191444 on Build GRCh38
rs921260080 Chr.9: 36191126-36191126 on Build GRCh38
rs967811743 Chr.9: 36191459-36191459 on Build GRCh38
rs768918708 Chr.9: 36191227-36191227 on Build GRCh38
rs765709490 Chr.9: 36191139-36191139 on Build GRCh38
rs773671483 Chr.9: 36191267-36191267 on Build GRCh38
rs747962162 Chr.9: 36191078-36191078 on Build GRCh38
rs934025178 Chr.9: 36191149-36191149 on Build GRCh38
rs749101121 Chr.9: 36191173-36191173 on Build GRCh38
rs747244844 Chr.9: 36191213-36191213 on Build GRCh38
rs773721183 Chr.9: 36191436-36191436 on Build GRCh38
rs748446095 Chr.9: 36191308-36191308 on Build GRCh38
rs958902572 Chr.9: 36191484-36191484 on Build GRCh38
rs368746473 Chr.9: 36191120-36191120 on Build GRCh38
rs896245072 Chr.9: 36191298-36191298 on Build GRCh38
rs1000188470 Chr.9: 36191463-36191463 on Build GRCh38
rs991662343 Chr.9: 36191489-36191489 on Build GRCh38
rs1042433145 Chr.9: 36191501-36191501 on Build GRCh38
rs775679746 Chr.9: 36191201-36191201 on Build GRCh38
rs746057561 Chr.9: 36191084-36191104 on Build GRCh38
rs868012291 Chr.9: 36191187-36191187 on Build GRCh38
rs961913181 Chr.9: 36191409-36191409 on Build GRCh38
rs777030848 Chr.9: 36191134-36191134 on Build GRCh38
rs752390516 Chr.9: 36191155-36191155 on Build GRCh38
rs754632055 Chr.9: 36191071-36191071 on Build GRCh38
rs762295830 Chr.9: 36191135-36191135 on Build GRCh38
rs940300742 Chr.9: 36191224-36191224 on Build GRCh38
rs756606123 Chr.9: 36191089-36191100 on Build GRCh38
rs779488340 Chr.9: 36191060-36191060 on Build GRCh38
rs770230356 Chr.9: 36191416-36191416 on Build GRCh38
rs780767112 Chr.9: 36191074-36191074 on Build GRCh38
rs772237494 Chr.9: 36191197-36191197 on Build GRCh38
rs1013630593 Chr.9: 36191302-36191302 on Build GRCh38
rs1033299277 Chr.9: 36191481-36191481 on Build GRCh38
rs979651549 Chr.9: 36191081-36191081 on Build GRCh38
rs1032116018 Chr.9: 36191320-36191320 on Build GRCh38
rs902199274 Chr.9: 36191510-36191510 on Build GRCh38
rs775804475 Chr.9: 36191129-36191129 on Build GRCh38
rs770884133 Chr.9: 36191113-36191113 on Build GRCh38

View Full Product Details