Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36219705-36220112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNE
Entrez Gene ID
10020
Gene Name
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Gene Aliases
DMRV, GLCNE, IBM2, NM, Uae1
UniGene
Hs.5920
Gene Chromosome Location
Chr.9: 36214441-36277056 on Build GRCh38
Transcripts
NM_001128227.2 NM_001190383.1 NM_001190384.1 NM_001190388.1 NM_005476.5 XM_005251334.4 XM_017014167.1 XM_017014168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5359556 c.1746A>G p.G582G Chr.9: 36219908-36219908 on Build GRCh38
1108895 c.1771G>A p.A591T Chr.9: 36219883-36219883 on Build GRCh38
1108896 c.1768T>G p.Y590D Chr.9: 36219886-36219886 on Build GRCh38
4687454 c.1767C>T p.H589H Chr.9: 36219980-36219980 on Build GRCh38
4687456 c.1659C>T p.H553H Chr.9: 36219980-36219980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763576597 Chr.9: 36220018-36220018 on Build GRCh38
rs745697898 Chr.9: 36220048-36220048 on Build GRCh38
rs112061124 Chr.9: 36219867-36219867 on Build GRCh38
rs762009737 Chr.9: 36219979-36219979 on Build GRCh38
rs759533498 Chr.9: 36219810-36219810 on Build GRCh38
rs980752867 Chr.9: 36220031-36220031 on Build GRCh38
rs750749076 Chr.9: 36219859-36219859 on Build GRCh38
rs780659199 Chr.9: 36219875-36219875 on Build GRCh38
rs746919372 Chr.9: 36219946-36219946 on Build GRCh38
rs778579770 Chr.9: 36219922-36219922 on Build GRCh38
rs754048020 Chr.9: 36219843-36219843 on Build GRCh38
rs121908625 Chr.9: 36219927-36219927 on Build GRCh38
rs765440649 Chr.9: 36219988-36219988 on Build GRCh38
rs121908632 Chr.9: 36219940-36219940 on Build GRCh38
rs147768211 Chr.9: 36220016-36220016 on Build GRCh38
rs774180720 Chr.9: 36219809-36219809 on Build GRCh38
rs775558035 Chr.9: 36219807-36219810 on Build GRCh38
rs775369694 Chr.9: 36219945-36219945 on Build GRCh38
rs754313408 Chr.9: 36220003-36220003 on Build GRCh38
rs918276456 Chr.9: 36219796-36219796 on Build GRCh38
rs202043018 Chr.9: 36219805-36219805 on Build GRCh38
rs765580553 Chr.9: 36219857-36219857 on Build GRCh38
rs777310871 Chr.9: 36219887-36219887 on Build GRCh38
rs758233921 Chr.9: 36220052-36220052 on Build GRCh38
rs752286512 Chr.9: 36219883-36219883 on Build GRCh38
rs756465454 Chr.9: 36219821-36219821 on Build GRCh38
rs748949603 Chr.9: 36219894-36219894 on Build GRCh38
rs763368852 Chr.9: 36219992-36219992 on Build GRCh38
rs886063925 Chr.9: 36219827-36219827 on Build GRCh38
rs545449250 Chr.9: 36220081-36220081 on Build GRCh38
rs866840797 Chr.9: 36219931-36219931 on Build GRCh38
rs938477742 Chr.9: 36219737-36219737 on Build GRCh38
rs111302956 Chr.9: 36219884-36219884 on Build GRCh38
rs779465942 Chr.9: 36220073-36220073 on Build GRCh38
rs528294561 Chr.9: 36219820-36219820 on Build GRCh38
rs770676846 Chr.9: 36219806-36219806 on Build GRCh38
rs886044550 Chr.9: 36219925-36219925 on Build GRCh38
rs768667610 Chr.9: 36219965-36219965 on Build GRCh38
rs758985414 Chr.9: 36219869-36219869 on Build GRCh38
rs144931111 Chr.9: 36219980-36219980 on Build GRCh38
rs139442869 Chr.9: 36220027-36220027 on Build GRCh38
rs1021602666 Chr.9: 36220024-36220024 on Build GRCh38
rs756926638 Chr.9: 36219912-36219912 on Build GRCh38
rs776760528 Chr.9: 36219973-36219973 on Build GRCh38
rs1057517157 Chr.9: 36219873-36219873 on Build GRCh38
rs200212703 Chr.9: 36219990-36219990 on Build GRCh38
rs1056240614 Chr.9: 36220068-36220068 on Build GRCh38
rs547919665 Chr.9: 36219838-36219838 on Build GRCh38
rs375600956 Chr.9: 36220033-36220033 on Build GRCh38
rs1056794417 Chr.9: 36219778-36219778 on Build GRCh38
rs886044449 Chr.9: 36219968-36219968 on Build GRCh38
rs886043979 Chr.9: 36219984-36219984 on Build GRCh38
rs541458680 Chr.9: 36219842-36219842 on Build GRCh38
rs766818430 Chr.9: 36219994-36219994 on Build GRCh38
rs756876449 Chr.9: 36220028-36220028 on Build GRCh38
rs779802128 Chr.9: 36220063-36220063 on Build GRCh38
rs548994228 Chr.9: 36219789-36219789 on Build GRCh38
rs762796465 Chr.9: 36219939-36219941 on Build GRCh38
rs760700813 Chr.9: 36219834-36219834 on Build GRCh38
rs387906347 Chr.9: 36219856-36219856 on Build GRCh38
rs1009223371 Chr.9: 36220085-36220085 on Build GRCh38
rs372732220 Chr.9: 36219847-36219847 on Build GRCh38

View Full Product Details