Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37510851-37511394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
UniGene
Hs.709527
Gene Chromosome Location
Chr.9: 37509920-37576459 on Build GRCh38
Transcripts
NM_012166.2 XM_006716754.3 XM_017014618.1 XM_017014619.1

SNP Detail

SNP ID Chromosome Location
rs749647716 Chr.9: 37511214-37511214 on Build GRCh38
rs1024999700 Chr.9: 37511321-37511321 on Build GRCh38
rs936856827 Chr.9: 37511144-37511144 on Build GRCh38
rs952687521 Chr.9: 37511113-37511113 on Build GRCh38
rs112321666 Chr.9: 37511253-37511253 on Build GRCh38
rs142203924 Chr.9: 37511354-37511354 on Build GRCh38
rs892390353 Chr.9: 37510901-37510901 on Build GRCh38
rs936282854 Chr.9: 37511208-37511208 on Build GRCh38
rs1128206 Chr.9: 37510971-37510971 on Build GRCh38
rs932516441 Chr.9: 37511324-37511324 on Build GRCh38
rs901036240 Chr.9: 37510982-37510982 on Build GRCh38
rs773512980 Chr.9: 37511207-37511207 on Build GRCh38
rs993068811 Chr.9: 37511162-37511162 on Build GRCh38
rs959551888 Chr.9: 37511082-37511082 on Build GRCh38
rs1013892844 Chr.9: 37511320-37511320 on Build GRCh38
rs565834919 Chr.9: 37511053-37511053 on Build GRCh38
rs913605032 Chr.9: 37511277-37511277 on Build GRCh38
rs755139003 Chr.9: 37510943-37510943 on Build GRCh38
rs764169182 Chr.9: 37511242-37511242 on Build GRCh38
rs548065469 Chr.9: 37511161-37511161 on Build GRCh38
rs1041269468 Chr.9: 37510952-37510952 on Build GRCh38
rs978079247 Chr.9: 37511333-37511333 on Build GRCh38
rs113940083 Chr.9: 37511257-37511257 on Build GRCh38
rs980900361 Chr.9: 37511182-37511182 on Build GRCh38
rs1015534180 Chr.9: 37511371-37511371 on Build GRCh38
rs112483154 Chr.9: 37511260-37511260 on Build GRCh38
rs150012291 Chr.9: 37511076-37511076 on Build GRCh38
rs541310616 Chr.9: 37510875-37510875 on Build GRCh38
rs945264508 Chr.9: 37511294-37511294 on Build GRCh38
rs139334754 Chr.9: 37510889-37510889 on Build GRCh38
rs933532751 Chr.9: 37511178-37511178 on Build GRCh38
rs528114530 Chr.9: 37511154-37511154 on Build GRCh38
rs781282392 Chr.9: 37510986-37510986 on Build GRCh38
rs577721987 Chr.9: 37510983-37510983 on Build GRCh38
rs146527731 Chr.9: 37511334-37511334 on Build GRCh38
rs35012461 Chr.9: 37510955-37510956 on Build GRCh38
rs563359534 Chr.9: 37511009-37511009 on Build GRCh38
rs970269625 Chr.9: 37511173-37511173 on Build GRCh38
rs1050231771 Chr.9: 37511330-37511330 on Build GRCh38
rs1015586614 Chr.9: 37511138-37511138 on Build GRCh38
rs746593740 Chr.9: 37510908-37510908 on Build GRCh38
rs376778091 Chr.9: 37511266-37511266 on Build GRCh38
rs925345595 Chr.9: 37511129-37511129 on Build GRCh38
rs571059877 Chr.9: 37511225-37511225 on Build GRCh38
rs530034069 Chr.9: 37511200-37511200 on Build GRCh38
rs533003705 Chr.9: 37511265-37511265 on Build GRCh38
rs1003886560 Chr.9: 37511000-37511000 on Build GRCh38
rs192857350 Chr.9: 37511117-37511117 on Build GRCh38
rs1050795096 Chr.9: 37511181-37511181 on Build GRCh38
rs974945989 Chr.9: 37511371-37511374 on Build GRCh38
rs184498035 Chr.9: 37511319-37511319 on Build GRCh38
rs1033893431 Chr.9: 37511279-37511279 on Build GRCh38
rs188400538 Chr.9: 37511001-37511001 on Build GRCh38
rs996563802 Chr.9: 37510949-37510949 on Build GRCh38
rs917386451 Chr.9: 37511167-37511167 on Build GRCh38
rs1040991103 Chr.9: 37511299-37511299 on Build GRCh38
rs992175526 Chr.9: 37511083-37511083 on Build GRCh38
rs771224033 Chr.9: 37511226-37511226 on Build GRCh38
rs149115671 Chr.9: 37511347-37511347 on Build GRCh38
rs746875777 Chr.9: 37510916-37510916 on Build GRCh38
rs768064924 Chr.9: 37511008-37511008 on Build GRCh38
rs1051160959 Chr.9: 37510954-37510954 on Build GRCh38

View Full Product Details