Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37511375-37511873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
UniGene
Hs.709527
Gene Chromosome Location
Chr.9: 37509920-37576459 on Build GRCh38
Transcripts
NM_012166.2 XM_006716754.3 XM_017014618.1 XM_017014619.1

SNP Detail

SNP ID Chromosome Location
rs61739629 Chr.9: 37511811-37511811 on Build GRCh38
rs1033166979 Chr.9: 37511822-37511822 on Build GRCh38
rs1014263359 Chr.9: 37511744-37511744 on Build GRCh38
rs989259208 Chr.9: 37511585-37511585 on Build GRCh38
rs866078627 Chr.9: 37511419-37511419 on Build GRCh38
rs563186899 Chr.9: 37511464-37511465 on Build GRCh38
rs774847729 Chr.9: 37511478-37511478 on Build GRCh38
rs544694604 Chr.9: 37511796-37511796 on Build GRCh38
rs573536910 Chr.9: 37511662-37511662 on Build GRCh38
rs972381766 Chr.9: 37511654-37511654 on Build GRCh38
rs1013402318 Chr.9: 37511466-37511466 on Build GRCh38
rs958159512 Chr.9: 37511551-37511551 on Build GRCh38
rs769817187 Chr.9: 37511609-37511609 on Build GRCh38
rs577878508 Chr.9: 37511442-37511442 on Build GRCh38
rs902428985 Chr.9: 37511485-37511486 on Build GRCh38
rs913554200 Chr.9: 37511600-37511600 on Build GRCh38
rs919564325 Chr.9: 37511684-37511684 on Build GRCh38
rs980298442 Chr.9: 37511485-37511485 on Build GRCh38
rs1049570683 Chr.9: 37511714-37511714 on Build GRCh38
rs777445976 Chr.9: 37511711-37511711 on Build GRCh38
rs113552601 Chr.9: 37511488-37511488 on Build GRCh38
rs957767884 Chr.9: 37511579-37511579 on Build GRCh38
rs1036654486 Chr.9: 37511418-37511419 on Build GRCh38
rs112110586 Chr.9: 37511630-37511630 on Build GRCh38
rs897185232 Chr.9: 37511422-37511422 on Build GRCh38
rs564773381 Chr.9: 37511797-37511797 on Build GRCh38
rs368518608 Chr.9: 37511748-37511748 on Build GRCh38
rs543577755 Chr.9: 37511443-37511443 on Build GRCh38
rs35594223 Chr.9: 37511563-37511563 on Build GRCh38
rs189291284 Chr.9: 37511417-37511417 on Build GRCh38
rs868643891 Chr.9: 37511418-37511418 on Build GRCh38
rs543103712 Chr.9: 37511441-37511441 on Build GRCh38
rs1054981239 Chr.9: 37511446-37511446 on Build GRCh38
rs184723886 Chr.9: 37511842-37511842 on Build GRCh38
rs192340699 Chr.9: 37511721-37511721 on Build GRCh38
rs776185435 Chr.9: 37511815-37511815 on Build GRCh38
rs906112814 Chr.9: 37511799-37511799 on Build GRCh38
rs939725856 Chr.9: 37511395-37511395 on Build GRCh38
rs1046222963 Chr.9: 37511772-37511772 on Build GRCh38
rs1037095090 Chr.9: 37511729-37511729 on Build GRCh38
rs759999688 Chr.9: 37511621-37511621 on Build GRCh38
rs897133402 Chr.9: 37511733-37511733 on Build GRCh38
rs151195878 Chr.9: 37511720-37511720 on Build GRCh38
rs1029058393 Chr.9: 37511612-37511612 on Build GRCh38
rs999527837 Chr.9: 37511516-37511516 on Build GRCh38
rs528031970 Chr.9: 37511750-37511750 on Build GRCh38
rs922554128 Chr.9: 37511653-37511653 on Build GRCh38
rs894948239 Chr.9: 37511455-37511455 on Build GRCh38
rs902438934 Chr.9: 37511783-37511783 on Build GRCh38
rs1033635094 Chr.9: 37511564-37511564 on Build GRCh38
rs1032308866 Chr.9: 37511525-37511526 on Build GRCh38
rs763619507 Chr.9: 37511711-37511711 on Build GRCh38

View Full Product Details