Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37518041-37518535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
UniGene
Hs.709527
Gene Chromosome Location
Chr.9: 37509920-37576459 on Build GRCh38
Transcripts
NM_012166.2 XM_006716754.3 XM_017014618.1 XM_017014619.1

SNP Detail

SNP ID Chromosome Location
rs776126565 Chr.9: 37518378-37518378 on Build GRCh38
rs930869268 Chr.9: 37518179-37518179 on Build GRCh38
rs763777212 Chr.9: 37518225-37518225 on Build GRCh38
rs752828824 Chr.9: 37518402-37518402 on Build GRCh38
rs756561849 Chr.9: 37518404-37518404 on Build GRCh38
rs906239656 Chr.9: 37518268-37518268 on Build GRCh38
rs756042976 Chr.9: 37518152-37518152 on Build GRCh38
rs375374605 Chr.9: 37518208-37518208 on Build GRCh38
rs758539162 Chr.9: 37518273-37518273 on Build GRCh38
rs61742584 Chr.9: 37518233-37518233 on Build GRCh38
rs1011915033 Chr.9: 37518223-37518223 on Build GRCh38
rs776057817 Chr.9: 37518303-37518303 on Build GRCh38
rs574814970 Chr.9: 37518256-37518256 on Build GRCh38
rs770030505 Chr.9: 37518309-37518309 on Build GRCh38
rs201210928 Chr.9: 37518205-37518205 on Build GRCh38
rs751668500 Chr.9: 37518392-37518392 on Build GRCh38
rs114814730 Chr.9: 37518377-37518377 on Build GRCh38
rs760227299 Chr.9: 37518366-37518366 on Build GRCh38
rs780093422 Chr.9: 37518278-37518278 on Build GRCh38
rs747424055 Chr.9: 37518094-37518094 on Build GRCh38
rs578244469 Chr.9: 37518507-37518507 on Build GRCh38
rs772422694 Chr.9: 37518302-37518302 on Build GRCh38
rs201676458 Chr.9: 37518380-37518380 on Build GRCh38
rs749748138 Chr.9: 37518331-37518331 on Build GRCh38
rs755197163 Chr.9: 37518300-37518300 on Build GRCh38
rs754998938 Chr.9: 37518397-37518397 on Build GRCh38
rs762368519 Chr.9: 37518107-37518107 on Build GRCh38
rs769065168 Chr.9: 37518193-37518193 on Build GRCh38
rs368304635 Chr.9: 37518112-37518112 on Build GRCh38
rs778381633 Chr.9: 37518330-37518330 on Build GRCh38
rs761710834 Chr.9: 37518379-37518379 on Build GRCh38
rs766237092 Chr.9: 37518390-37518390 on Build GRCh38
rs767826451 Chr.9: 37518398-37518398 on Build GRCh38
rs758387108 Chr.9: 37518171-37518171 on Build GRCh38
rs753699828 Chr.9: 37518255-37518255 on Build GRCh38
rs771707966 Chr.9: 37518210-37518210 on Build GRCh38
rs868441133 Chr.9: 37518337-37518337 on Build GRCh38
rs535957692 Chr.9: 37518206-37518206 on Build GRCh38
rs767298585 Chr.9: 37518135-37518135 on Build GRCh38
rs866735793 Chr.9: 37518412-37518412 on Build GRCh38
rs765605119 Chr.9: 37518109-37518109 on Build GRCh38
rs192244773 Chr.9: 37518082-37518082 on Build GRCh38
rs567358595 Chr.9: 37518189-37518189 on Build GRCh38
rs763970145 Chr.9: 37518158-37518158 on Build GRCh38
rs367999710 Chr.9: 37518100-37518100 on Build GRCh38
rs76495281 Chr.9: 37518257-37518257 on Build GRCh38
rs745869979 Chr.9: 37518075-37518075 on Build GRCh38
rs748393774 Chr.9: 37518198-37518198 on Build GRCh38
rs750298328 Chr.9: 37518272-37518272 on Build GRCh38
rs749601452 Chr.9: 37518419-37518419 on Build GRCh38
rs749206853 Chr.9: 37518099-37518099 on Build GRCh38
rs369007874 Chr.9: 37518134-37518134 on Build GRCh38
rs761137598 Chr.9: 37518451-37518451 on Build GRCh38
rs956549460 Chr.9: 37518465-37518466 on Build GRCh38
rs931970911 Chr.9: 37518090-37518090 on Build GRCh38
rs370226873 Chr.9: 37518413-37518413 on Build GRCh38
rs757874663 Chr.9: 37518430-37518430 on Build GRCh38
rs777572957 Chr.9: 37518085-37518085 on Build GRCh38
rs573535073 Chr.9: 37518166-37518166 on Build GRCh38
rs1020655691 Chr.9: 37518062-37518062 on Build GRCh38
rs759881885 Chr.9: 37518290-37518290 on Build GRCh38
rs941494624 Chr.9: 37518174-37518174 on Build GRCh38
rs757227303 Chr.9: 37518162-37518162 on Build GRCh38
rs879128316 Chr.9: 37518393-37518393 on Build GRCh38
rs780296848 Chr.9: 37518181-37518181 on Build GRCh38
rs781549598 Chr.9: 37518194-37518194 on Build GRCh38
rs775149596 Chr.9: 37518363-37518363 on Build GRCh38
rs781228449 Chr.9: 37518301-37518301 on Build GRCh38
rs752372484 Chr.9: 37518137-37518137 on Build GRCh38
rs564742179 Chr.9: 37518409-37518409 on Build GRCh38
rs778677445 Chr.9: 37518165-37518165 on Build GRCh38
rs746567001 Chr.9: 37518472-37518472 on Build GRCh38
rs772371696 Chr.9: 37518080-37518080 on Build GRCh38
rs376743442 Chr.9: 37518336-37518336 on Build GRCh38
rs751580511 Chr.9: 37518284-37518284 on Build GRCh38
rs768977160 Chr.9: 37518098-37518098 on Build GRCh38
rs199866682 Chr.9: 37518133-37518133 on Build GRCh38
rs1007133914 Chr.9: 37518445-37518445 on Build GRCh38
rs370012357 Chr.9: 37518224-37518224 on Build GRCh38
rs775344192 Chr.9: 37518216-37518216 on Build GRCh38
rs779513862 Chr.9: 37518438-37518438 on Build GRCh38
rs753523555 Chr.9: 37518161-37518161 on Build GRCh38
rs143437639 Chr.9: 37518382-37518382 on Build GRCh38

View Full Product Details