Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111649359-111649905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC25
Entrez Gene ID
548645
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Gene Aliases
bA16L21.2.1
UniGene
Hs.534196
Gene Chromosome Location
Chr.9: 111631352-111654351 on Build GRCh38
Transcripts
NM_001015882.2 NR_037148.1 NM_004125.3

Gene Symbol
DNAJC25-GNG10
Entrez Gene ID
552891
Gene Name
DNAJC25-GNG10 readthrough
Gene Aliases
-
UniGene
Hs.534196
Gene Chromosome Location
Chr.9: 111631352-111670246 on Build GRCh38
Transcripts
NM_001015882.2 NR_037148.1 NM_004125.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3169976 c.799A>G p.I267V Chr.9: 111649762-111649762 on Build GRCh38
3169975 c.614G>A p.G205D Chr.9: 111649577-111649577 on Build GRCh38
7262939 c.594_595insA p.A201fs*16 Chr.9: 111649557-111649558 on Build GRCh38
260518 c.553C>T p.R185C Chr.9: 111649516-111649516 on Build GRCh38
7258528 c.770G>A p.C257Y Chr.9: 111649733-111649733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749289226 Chr.9: 111649760-111649760 on Build GRCh38
rs745714904 Chr.9: 111649735-111649735 on Build GRCh38
rs771511219 Chr.9: 111649780-111649780 on Build GRCh38
rs537603658 Chr.9: 111649501-111649501 on Build GRCh38
rs776791144 Chr.9: 111649503-111649503 on Build GRCh38
rs907133557 Chr.9: 111649706-111649706 on Build GRCh38
rs140621968 Chr.9: 111649809-111649809 on Build GRCh38
rs760951719 Chr.9: 111649500-111649500 on Build GRCh38
rs769980083 Chr.9: 111649618-111649618 on Build GRCh38
rs906357078 Chr.9: 111649762-111649762 on Build GRCh38
rs751963227 Chr.9: 111649576-111649576 on Build GRCh38
rs921977631 Chr.9: 111649541-111649541 on Build GRCh38
rs538717472 Chr.9: 111649429-111649429 on Build GRCh38
rs754682829 Chr.9: 111649525-111649525 on Build GRCh38
rs770085115 Chr.9: 111649756-111649756 on Build GRCh38
rs371744939 Chr.9: 111649636-111649636 on Build GRCh38
rs767868785 Chr.9: 111649794-111649794 on Build GRCh38
rs745598077 Chr.9: 111649606-111649606 on Build GRCh38
rs751987472 Chr.9: 111649655-111649655 on Build GRCh38
rs758620820 Chr.9: 111649863-111649864 on Build GRCh38
rs202015606 Chr.9: 111649840-111649840 on Build GRCh38
rs1002922095 Chr.9: 111649442-111649442 on Build GRCh38
rs7019332 Chr.9: 111649665-111649665 on Build GRCh38
rs764455550 Chr.9: 111649570-111649570 on Build GRCh38
rs766771850 Chr.9: 111649518-111649518 on Build GRCh38
rs753283385 Chr.9: 111649728-111649728 on Build GRCh38
rs995251346 Chr.9: 111649872-111649872 on Build GRCh38
rs777750152 Chr.9: 111649569-111649569 on Build GRCh38
rs376569149 Chr.9: 111649580-111649580 on Build GRCh38
rs763551475 Chr.9: 111649526-111649526 on Build GRCh38
rs375241958 Chr.9: 111649430-111649430 on Build GRCh38
rs748375928 Chr.9: 111649740-111649740 on Build GRCh38
rs746669881 Chr.9: 111649432-111649432 on Build GRCh38
rs762032954 Chr.9: 111649612-111649612 on Build GRCh38
rs773448980 Chr.9: 111649757-111649757 on Build GRCh38
rs756690189 Chr.9: 111649850-111649850 on Build GRCh38
rs768953133 Chr.9: 111649607-111649607 on Build GRCh38
rs752247206 Chr.9: 111649559-111649559 on Build GRCh38
rs755691509 Chr.9: 111649688-111649688 on Build GRCh38
rs930948272 Chr.9: 111649621-111649621 on Build GRCh38
rs535422079 Chr.9: 111649443-111649443 on Build GRCh38
rs1322259 Chr.9: 111649611-111649611 on Build GRCh38
rs753374082 Chr.9: 111649822-111649822 on Build GRCh38
rs936093747 Chr.9: 111649565-111649565 on Build GRCh38
rs552447394 Chr.9: 111649450-111649450 on Build GRCh38
rs750314561 Chr.9: 111649860-111649860 on Build GRCh38
rs755601452 Chr.9: 111649566-111649566 on Build GRCh38
rs368887646 Chr.9: 111649489-111649489 on Build GRCh38
rs983684481 Chr.9: 111649424-111649424 on Build GRCh38
rs374845005 Chr.9: 111649478-111649478 on Build GRCh38
rs760006244 Chr.9: 111649787-111649787 on Build GRCh38
rs765267335 Chr.9: 111649851-111649851 on Build GRCh38
rs370580899 Chr.9: 111649880-111649880 on Build GRCh38
rs766749038 Chr.9: 111649647-111649647 on Build GRCh38
rs780670002 Chr.9: 111649551-111649551 on Build GRCh38
rs372256683 Chr.9: 111649517-111649517 on Build GRCh38
rs894828413 Chr.9: 111649663-111649663 on Build GRCh38
rs750062063 Chr.9: 111649684-111649684 on Build GRCh38
rs753724007 Chr.9: 111649786-111649788 on Build GRCh38
rs770703441 Chr.9: 111649572-111649572 on Build GRCh38
rs193192759 Chr.9: 111649389-111649389 on Build GRCh38
rs776960637 Chr.9: 111649610-111649610 on Build GRCh38
rs757237830 Chr.9: 111649729-111649729 on Build GRCh38
rs55825206 Chr.9: 111649743-111649743 on Build GRCh38
rs778671787 Chr.9: 111649731-111649731 on Build GRCh38
rs758802430 Chr.9: 111649422-111649422 on Build GRCh38
rs750942878 Chr.9: 111649516-111649516 on Build GRCh38
rs754520350 Chr.9: 111649438-111649438 on Build GRCh38
rs771739888 Chr.9: 111649736-111649736 on Build GRCh38
rs781622335 Chr.9: 111649738-111649738 on Build GRCh38
rs868430633 Chr.9: 111649672-111649672 on Build GRCh38
rs773225801 Chr.9: 111649426-111649426 on Build GRCh38
rs576951424 Chr.9: 111649786-111649786 on Build GRCh38
rs763684985 Chr.9: 111649818-111649818 on Build GRCh38
rs554851029 Chr.9: 111649508-111649508 on Build GRCh38
rs769756835 Chr.9: 111649454-111649454 on Build GRCh38
rs760750830 Chr.9: 111649530-111649531 on Build GRCh38
rs751812581 Chr.9: 111649519-111649519 on Build GRCh38
rs766270759 Chr.9: 111649612-111649614 on Build GRCh38
rs1032205178 Chr.9: 111649476-111649476 on Build GRCh38
rs777266187 Chr.9: 111649698-111649698 on Build GRCh38
rs763529345 Chr.9: 111649642-111649642 on Build GRCh38
rs1055471412 Chr.9: 111649737-111649737 on Build GRCh38
rs780008929 Chr.9: 111649871-111649871 on Build GRCh38
rs534213065 Chr.9: 111649658-111649658 on Build GRCh38
rs371506049 Chr.9: 111649477-111649477 on Build GRCh38
rs775768214 Chr.9: 111649797-111649797 on Build GRCh38
rs773913181 Chr.9: 111649589-111649589 on Build GRCh38
rs758278806 Chr.9: 111649870-111649870 on Build GRCh38
rs367873033 Chr.9: 111649468-111649468 on Build GRCh38

View Full Product Details