Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111692015-111692512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
UniGene
Hs.428209
Gene Chromosome Location
Chr.9: 111686175-111795949 on Build GRCh38
Transcripts
NM_001080551.2 NM_173521.4 NR_109816.1 XM_006716974.2 XM_006716975.2 XM_011518302.2 XM_011518303.1 XM_011518306.1 XM_017014340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7283562 c.3290A>G p.D1097G Chr.9: 111692378-111692378 on Build GRCh38
4851668 c.3573C>G p.V1191V Chr.9: 111692095-111692095 on Build GRCh38
5091291 c.3520G>T p.E1174* Chr.9: 111692265-111692265 on Build GRCh38
7195090 c.3507_3510delTAAT p.N1170fs*1 Chr.9: 111692158-111692161 on Build GRCh38
7156364 c.3485G>C p.G1162A Chr.9: 111692183-111692183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899670078 Chr.9: 111692050-111692050 on Build GRCh38
rs776341458 Chr.9: 111692351-111692351 on Build GRCh38
rs537629803 Chr.9: 111692480-111692480 on Build GRCh38
rs771860306 Chr.9: 111692176-111692176 on Build GRCh38
rs745835560 Chr.9: 111692169-111692169 on Build GRCh38
rs755375331 Chr.9: 111692130-111692130 on Build GRCh38
rs1015562239 Chr.9: 111692232-111692232 on Build GRCh38
rs1042091303 Chr.9: 111692074-111692074 on Build GRCh38
rs1028449576 Chr.9: 111692313-111692313 on Build GRCh38
rs201575234 Chr.9: 111692311-111692311 on Build GRCh38
rs7869279 Chr.9: 111692264-111692264 on Build GRCh38
rs759510670 Chr.9: 111692370-111692370 on Build GRCh38
rs770557223 Chr.9: 111692157-111692157 on Build GRCh38
rs753541193 Chr.9: 111692288-111692288 on Build GRCh38
rs763087928 Chr.9: 111692219-111692219 on Build GRCh38
rs773411789 Chr.9: 111692207-111692207 on Build GRCh38
rs771039542 Chr.9: 111692200-111692201 on Build GRCh38
rs779947256 Chr.9: 111692306-111692306 on Build GRCh38
rs756530575 Chr.9: 111692269-111692269 on Build GRCh38
rs774422432 Chr.9: 111692471-111692471 on Build GRCh38
rs777633205 Chr.9: 111692287-111692287 on Build GRCh38
rs766485682 Chr.9: 111692222-111692222 on Build GRCh38
rs765089534 Chr.9: 111692306-111692307 on Build GRCh38
rs146868010 Chr.9: 111692203-111692203 on Build GRCh38
rs143924637 Chr.9: 111692182-111692182 on Build GRCh38
rs1013072159 Chr.9: 111692151-111692151 on Build GRCh38
rs773664316 Chr.9: 111692160-111692160 on Build GRCh38
rs748862510 Chr.9: 111692156-111692156 on Build GRCh38
rs758451439 Chr.9: 111692100-111692100 on Build GRCh38
rs751543959 Chr.9: 111692238-111692238 on Build GRCh38
rs767582583 Chr.9: 111692487-111692488 on Build GRCh38
rs148430343 Chr.9: 111692441-111692441 on Build GRCh38
rs12343237 Chr.9: 111692452-111692452 on Build GRCh38
rs185569270 Chr.9: 111692084-111692084 on Build GRCh38
rs745334756 Chr.9: 111692296-111692296 on Build GRCh38
rs368091767 Chr.9: 111692140-111692140 on Build GRCh38
rs1475110 Chr.9: 111692300-111692300 on Build GRCh38
rs768437310 Chr.9: 111692344-111692344 on Build GRCh38
rs752103420 Chr.9: 111692116-111692116 on Build GRCh38
rs145493933 Chr.9: 111692331-111692331 on Build GRCh38
rs771231552 Chr.9: 111692466-111692466 on Build GRCh38
rs753249231 Chr.9: 111692408-111692408 on Build GRCh38
rs371906607 Chr.9: 111692199-111692199 on Build GRCh38
rs762049578 Chr.9: 111692090-111692090 on Build GRCh38
rs200568507 Chr.9: 111692443-111692443 on Build GRCh38
rs771018313 Chr.9: 111692355-111692355 on Build GRCh38
rs762156009 Chr.9: 111692471-111692472 on Build GRCh38
rs764562944 Chr.9: 111692439-111692439 on Build GRCh38
rs751679362 Chr.9: 111692471-111692484 on Build GRCh38
rs186931900 Chr.9: 111692436-111692436 on Build GRCh38
rs568747748 Chr.9: 111692462-111692462 on Build GRCh38
rs145358586 Chr.9: 111692037-111692037 on Build GRCh38
rs1034902618 Chr.9: 111692163-111692163 on Build GRCh38
rs371940554 Chr.9: 111692060-111692060 on Build GRCh38
rs149179298 Chr.9: 111692152-111692152 on Build GRCh38
rs746023441 Chr.9: 111692472-111692472 on Build GRCh38
rs528797120 Chr.9: 111692383-111692383 on Build GRCh38
rs759208219 Chr.9: 111692299-111692306 on Build GRCh38
rs777543094 Chr.9: 111692154-111692154 on Build GRCh38
rs775388587 Chr.9: 111692180-111692180 on Build GRCh38
rs750545536 Chr.9: 111692097-111692097 on Build GRCh38
rs774527492 Chr.9: 111692360-111692360 on Build GRCh38
rs774329597 Chr.9: 111692041-111692041 on Build GRCh38
rs369777741 Chr.9: 111692373-111692373 on Build GRCh38
rs145278230 Chr.9: 111692290-111692290 on Build GRCh38
rs772046616 Chr.9: 111692477-111692477 on Build GRCh38
rs775093905 Chr.9: 111692088-111692088 on Build GRCh38
rs989323279 Chr.9: 111692420-111692420 on Build GRCh38
rs746953682 Chr.9: 111692453-111692453 on Build GRCh38
rs776727452 Chr.9: 111692249-111692249 on Build GRCh38
rs546584336 Chr.9: 111692444-111692444 on Build GRCh38
rs372656232 Chr.9: 111692149-111692149 on Build GRCh38
rs996634535 Chr.9: 111692070-111692070 on Build GRCh38
rs867018755 Chr.9: 111692135-111692135 on Build GRCh38
rs753159581 Chr.9: 111692265-111692265 on Build GRCh38
rs778512389 Chr.9: 111692292-111692292 on Build GRCh38
rs765405688 Chr.9: 111692092-111692092 on Build GRCh38
rs73656294 Chr.9: 111692455-111692455 on Build GRCh38
rs377103084 Chr.9: 111692352-111692352 on Build GRCh38
rs373919947 Chr.9: 111692115-111692115 on Build GRCh38
rs977126534 Chr.9: 111692324-111692324 on Build GRCh38
rs760120601 Chr.9: 111692260-111692260 on Build GRCh38

View Full Product Details