Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112687225-112687733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INIP
Entrez Gene ID
58493
Gene Name
INTS3 and NABP interacting protein
Gene Aliases
C9orf80, HSPC043, MISE, SOSSC, SSBIP1, hSSBIP1
UniGene
Hs.658575
Gene Chromosome Location
Chr.9: 112686506-112718107 on Build GRCh38
Transcripts
NM_021218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3675212 c.117C>A p.L39L Chr.9: 112687555-112687555 on Build GRCh38
7117098 c.256C>G p.Q86E Chr.9: 112687597-112687597 on Build GRCh38
7117099 c.75C>G p.L25L Chr.9: 112687597-112687597 on Build GRCh38
3675211 c.298C>A p.R100S Chr.9: 112687555-112687555 on Build GRCh38
6432330 c.229G>A p.A77T Chr.9: 112687624-112687624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769793637 Chr.9: 112687591-112687591 on Build GRCh38
rs750671358 Chr.9: 112687668-112687669 on Build GRCh38
rs959122411 Chr.9: 112687431-112687431 on Build GRCh38
rs748064382 Chr.9: 112687555-112687555 on Build GRCh38
rs555119345 Chr.9: 112687388-112687388 on Build GRCh38
rs765894291 Chr.9: 112687683-112687683 on Build GRCh38
rs200377309 Chr.9: 112687537-112687537 on Build GRCh38
rs758069479 Chr.9: 112687643-112687647 on Build GRCh38
rs752683964 Chr.9: 112687495-112687513 on Build GRCh38
rs891109421 Chr.9: 112687322-112687322 on Build GRCh38
rs537156612 Chr.9: 112687313-112687314 on Build GRCh38
rs754318083 Chr.9: 112687676-112687676 on Build GRCh38
rs758525818 Chr.9: 112687524-112687524 on Build GRCh38
rs746742474 Chr.9: 112687536-112687536 on Build GRCh38
rs111893760 Chr.9: 112687692-112687692 on Build GRCh38
rs918962622 Chr.9: 112687332-112687332 on Build GRCh38
rs764439079 Chr.9: 112687474-112687474 on Build GRCh38
rs757893139 Chr.9: 112687507-112687507 on Build GRCh38
rs943199679 Chr.9: 112687378-112687378 on Build GRCh38
rs764041408 Chr.9: 112687667-112687668 on Build GRCh38
rs1043554191 Chr.9: 112687691-112687691 on Build GRCh38
rs754406239 Chr.9: 112687483-112687483 on Build GRCh38
rs1035897539 Chr.9: 112687705-112687705 on Build GRCh38
rs369116300 Chr.9: 112687509-112687509 on Build GRCh38
rs537660052 Chr.9: 112687391-112687391 on Build GRCh38
rs1049766221 Chr.9: 112687306-112687306 on Build GRCh38
rs557640511 Chr.9: 112687402-112687402 on Build GRCh38
rs866617188 Chr.9: 112687265-112687265 on Build GRCh38
rs786969 Chr.9: 112687276-112687276 on Build GRCh38
rs781051198 Chr.9: 112687549-112687549 on Build GRCh38
rs1012394514 Chr.9: 112687460-112687460 on Build GRCh38
rs755323124 Chr.9: 112687344-112687344 on Build GRCh38
rs1008169998 Chr.9: 112687327-112687327 on Build GRCh38
rs768503746 Chr.9: 112687539-112687539 on Build GRCh38
rs1036357475 Chr.9: 112687249-112687249 on Build GRCh38
rs182168041 Chr.9: 112687487-112687487 on Build GRCh38
rs990297859 Chr.9: 112687674-112687674 on Build GRCh38
rs1038831511 Chr.9: 112687381-112687381 on Build GRCh38
rs762576946 Chr.9: 112687607-112687607 on Build GRCh38
rs755120988 Chr.9: 112687676-112687677 on Build GRCh38
rs372713136 Chr.9: 112687656-112687657 on Build GRCh38
rs780260531 Chr.9: 112687672-112687673 on Build GRCh38
rs770229733 Chr.9: 112687416-112687416 on Build GRCh38
rs772682472 Chr.9: 112687603-112687603 on Build GRCh38
rs756446451 Chr.9: 112687671-112687674 on Build GRCh38
rs79679349 Chr.9: 112687283-112687283 on Build GRCh38
rs750570977 Chr.9: 112687515-112687515 on Build GRCh38
rs761130344 Chr.9: 112687472-112687472 on Build GRCh38
rs759016094 Chr.9: 112687701-112687701 on Build GRCh38
rs771263218 Chr.9: 112687625-112687625 on Build GRCh38
rs764536415 Chr.9: 112687673-112687673 on Build GRCh38
rs190287731 Chr.9: 112687375-112687375 on Build GRCh38
rs774054401 Chr.9: 112687650-112687650 on Build GRCh38
rs754221331 Chr.9: 112687319-112687319 on Build GRCh38
rs766098900 Chr.9: 112687463-112687463 on Build GRCh38
rs760887356 Chr.9: 112687672-112687672 on Build GRCh38
rs753204371 Chr.9: 112687525-112687525 on Build GRCh38
rs752984201 Chr.9: 112687468-112687468 on Build GRCh38
rs373351991 Chr.9: 112687554-112687554 on Build GRCh38
rs960287173 Chr.9: 112687253-112687253 on Build GRCh38
rs950369991 Chr.9: 112687334-112687334 on Build GRCh38

View Full Product Details