Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112693719-112694219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INIP
Entrez Gene ID
58493
Gene Name
INTS3 and NABP interacting protein
Gene Aliases
C9orf80, HSPC043, MISE, SOSSC, SSBIP1, hSSBIP1
UniGene
Hs.658575
Gene Chromosome Location
Chr.9: 112686506-112718107 on Build GRCh38
Transcripts
NM_021218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4783469 c.24T>G p.N8K Chr.9: 112694145-112694145 on Build GRCh38
3653148 c.80G>C p.R27T Chr.9: 112694179-112694179 on Build GRCh38
1330793 c.49G>A p.A17T Chr.9: 112694210-112694210 on Build GRCh38
1459373 c.80G>T p.R27I Chr.9: 112694179-112694179 on Build GRCh38
1459372 c.114T>G p.N38K Chr.9: 112694145-112694145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046017345 Chr.9: 112693938-112693938 on Build GRCh38
rs374732730 Chr.9: 112694098-112694099 on Build GRCh38
rs1026073962 Chr.9: 112694044-112694044 on Build GRCh38
rs571895573 Chr.9: 112694107-112694107 on Build GRCh38
rs377334816 Chr.9: 112694074-112694083 on Build GRCh38
rs528906693 Chr.9: 112693794-112693794 on Build GRCh38
rs950558373 Chr.9: 112694061-112694061 on Build GRCh38
rs1057288399 Chr.9: 112694000-112694000 on Build GRCh38
rs985473563 Chr.9: 112694115-112694115 on Build GRCh38
rs947485873 Chr.9: 112693932-112693932 on Build GRCh38
rs1053211669 Chr.9: 112693834-112693834 on Build GRCh38
rs974661771 Chr.9: 112694118-112694118 on Build GRCh38
rs984999271 Chr.9: 112693758-112693758 on Build GRCh38
rs1013633546 Chr.9: 112694023-112694023 on Build GRCh38
rs193260710 Chr.9: 112694075-112694075 on Build GRCh38
rs771800380 Chr.9: 112694121-112694121 on Build GRCh38
rs907492599 Chr.9: 112693980-112693980 on Build GRCh38
rs747430063 Chr.9: 112693928-112693928 on Build GRCh38
rs1037295132 Chr.9: 112693859-112693859 on Build GRCh38
rs565650076 Chr.9: 112693990-112693990 on Build GRCh38
rs201179480 Chr.9: 112693873-112693873 on Build GRCh38
rs1001771818 Chr.9: 112693989-112693989 on Build GRCh38
rs985421309 Chr.9: 112693795-112693795 on Build GRCh38
rs147002098 Chr.9: 112694167-112694167 on Build GRCh38
rs745418556 Chr.9: 112694119-112694119 on Build GRCh38
rs906023684 Chr.9: 112693970-112693970 on Build GRCh38
rs765339312 Chr.9: 112694074-112694088 on Build GRCh38
rs931087606 Chr.9: 112693908-112693908 on Build GRCh38
rs913273547 Chr.9: 112693900-112693900 on Build GRCh38
rs954089117 Chr.9: 112694095-112694095 on Build GRCh38
rs190345522 Chr.9: 112693967-112693967 on Build GRCh38
rs964508603 Chr.9: 112694090-112694090 on Build GRCh38
rs950585116 Chr.9: 112693745-112693745 on Build GRCh38
rs199633346 Chr.9: 112694138-112694138 on Build GRCh38
rs750222793 Chr.9: 112694084-112694088 on Build GRCh38
rs1003109482 Chr.9: 112694004-112694004 on Build GRCh38
rs972665846 Chr.9: 112694144-112694144 on Build GRCh38
rs914591460 Chr.9: 112693748-112693748 on Build GRCh38
rs778147602 Chr.9: 112694104-112694107 on Build GRCh38
rs778088341 Chr.9: 112693831-112693831 on Build GRCh38
rs778451397 Chr.9: 112694109-112694109 on Build GRCh38
rs974643583 Chr.9: 112693802-112693802 on Build GRCh38
rs1011886106 Chr.9: 112693933-112693933 on Build GRCh38
rs909041326 Chr.9: 112693772-112693772 on Build GRCh38
rs143753591 Chr.9: 112694070-112694070 on Build GRCh38
rs775311670 Chr.9: 112694141-112694141 on Build GRCh38
rs748457885 Chr.9: 112694145-112694145 on Build GRCh38
rs567737828 Chr.9: 112694122-112694122 on Build GRCh38
rs923278437 Chr.9: 112693803-112693803 on Build GRCh38
rs1032296535 Chr.9: 112694016-112694016 on Build GRCh38
rs990807921 Chr.9: 112693947-112693947 on Build GRCh38
rs549018300 Chr.9: 112693968-112693968 on Build GRCh38
rs964645310 Chr.9: 112693782-112693782 on Build GRCh38
rs896173397 Chr.9: 112694020-112694020 on Build GRCh38

View Full Product Details