Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112694118-112694631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INIP
Entrez Gene ID
58493
Gene Name
INTS3 and NABP interacting protein
Gene Aliases
C9orf80, HSPC043, MISE, SOSSC, SSBIP1, hSSBIP1
UniGene
Hs.658575
Gene Chromosome Location
Chr.9: 112686506-112718107 on Build GRCh38
Transcripts
NM_021218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4783469 c.24T>G p.N8K Chr.9: 112694145-112694145 on Build GRCh38
3653148 c.80G>C p.R27T Chr.9: 112694179-112694179 on Build GRCh38
1330793 c.49G>A p.A17T Chr.9: 112694210-112694210 on Build GRCh38
1247024 c.26-3_26-2insT p.? Chr.9: 112694235-112694236 on Build GRCh38
5830937 c.26-3delT p.? Chr.9: 112694236-112694236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947792257 Chr.9: 112694208-112694208 on Build GRCh38
rs573949931 Chr.9: 112694310-112694310 on Build GRCh38
rs1020737302 Chr.9: 112694585-112694585 on Build GRCh38
rs770521210 Chr.9: 112694236-112694236 on Build GRCh38
rs1011767143 Chr.9: 112694403-112694403 on Build GRCh38
rs776296250 Chr.9: 112694235-112694236 on Build GRCh38
rs543675537 Chr.9: 112694442-112694442 on Build GRCh38
rs553390698 Chr.9: 112694322-112694322 on Build GRCh38
rs972665846 Chr.9: 112694144-112694144 on Build GRCh38
rs577191305 Chr.9: 112694405-112694405 on Build GRCh38
rs112741889 Chr.9: 112694264-112694265 on Build GRCh38
rs774472466 Chr.9: 112694246-112694246 on Build GRCh38
rs989247528 Chr.9: 112694255-112694255 on Build GRCh38
rs886100484 Chr.9: 112694392-112694392 on Build GRCh38
rs181657830 Chr.9: 112694606-112694606 on Build GRCh38
rs763512791 Chr.9: 112694202-112694202 on Build GRCh38
rs563924428 Chr.9: 112694417-112694417 on Build GRCh38
rs913213654 Chr.9: 112694285-112694285 on Build GRCh38
rs775278476 Chr.9: 112694278-112694278 on Build GRCh38
rs769358042 Chr.9: 112694269-112694270 on Build GRCh38
rs937469455 Chr.9: 112694334-112694334 on Build GRCh38
rs748457885 Chr.9: 112694145-112694145 on Build GRCh38
rs74461374 Chr.9: 112694317-112694317 on Build GRCh38
rs988741691 Chr.9: 112694555-112694555 on Build GRCh38
rs1006975603 Chr.9: 112694507-112694507 on Build GRCh38
rs1030152061 Chr.9: 112694531-112694531 on Build GRCh38
rs373316404 Chr.9: 112694383-112694383 on Build GRCh38
rs112645951 Chr.9: 112694240-112694240 on Build GRCh38
rs773507034 Chr.9: 112694198-112694198 on Build GRCh38
rs766839667 Chr.9: 112694205-112694205 on Build GRCh38
rs965412307 Chr.9: 112694569-112694569 on Build GRCh38
rs147002098 Chr.9: 112694167-112694167 on Build GRCh38
rs896026145 Chr.9: 112694358-112694358 on Build GRCh38
rs563439930 Chr.9: 112694494-112694494 on Build GRCh38
rs142604483 Chr.9: 112694421-112694421 on Build GRCh38
rs545126101 Chr.9: 112694344-112694344 on Build GRCh38
rs572990954 Chr.9: 112694329-112694329 on Build GRCh38
rs189013730 Chr.9: 112694274-112694274 on Build GRCh38
rs377288406 Chr.9: 112694271-112694272 on Build GRCh38
rs775476600 Chr.9: 112694367-112694367 on Build GRCh38
rs954811341 Chr.9: 112694547-112694547 on Build GRCh38
rs763247692 Chr.9: 112694473-112694473 on Build GRCh38

View Full Product Details