Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113178445-113178960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP15
Entrez Gene ID
23307
Gene Name
FK506 binding protein 15
Gene Aliases
FKBP133, KIAA0674, PPP1R76
UniGene
Hs.522351
Gene Chromosome Location
Chr.9: 113165036-113221361 on Build GRCh38
Transcripts
NM_015258.1 XM_006717018.2 XM_006717019.1 XM_017014552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3942878 c.1978A>T p.T660S Chr.9: 113178738-113178738 on Build GRCh38
3763543 c.2025A>G p.A675A Chr.9: 113178766-113178766 on Build GRCh38
3170510 c.2138C>G p.T713S Chr.9: 113178653-113178653 on Build GRCh38
6871552 c.2098G>A p.E700K Chr.9: 113178693-113178693 on Build GRCh38
5005850 c.2043G>C p.Q681H Chr.9: 113178748-113178748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374035252 Chr.9: 113178769-113178769 on Build GRCh38
rs1031232801 Chr.9: 113178741-113178741 on Build GRCh38
rs764577153 Chr.9: 113178653-113178653 on Build GRCh38
rs905230065 Chr.9: 113178755-113178755 on Build GRCh38
rs777030929 Chr.9: 113178767-113178767 on Build GRCh38
rs976695738 Chr.9: 113178536-113178536 on Build GRCh38
rs11790757 Chr.9: 113178787-113178787 on Build GRCh38
rs374047968 Chr.9: 113178608-113178608 on Build GRCh38
rs767788902 Chr.9: 113178837-113178837 on Build GRCh38
rs746697504 Chr.9: 113178745-113178745 on Build GRCh38
rs750502986 Chr.9: 113178672-113178672 on Build GRCh38
rs878866665 Chr.9: 113178893-113178893 on Build GRCh38
rs757852701 Chr.9: 113178651-113178651 on Build GRCh38
rs1025398380 Chr.9: 113178493-113178493 on Build GRCh38
rs370204594 Chr.9: 113178736-113178736 on Build GRCh38
rs1045148382 Chr.9: 113178922-113178922 on Build GRCh38
rs952537799 Chr.9: 113178554-113178554 on Build GRCh38
rs77435178 Chr.9: 113178547-113178547 on Build GRCh38
rs141462079 Chr.9: 113178751-113178751 on Build GRCh38
rs984355926 Chr.9: 113178873-113178873 on Build GRCh38
rs766465974 Chr.9: 113178813-113178813 on Build GRCh38
rs993676365 Chr.9: 113178776-113178776 on Build GRCh38
rs922536776 Chr.9: 113178918-113178918 on Build GRCh38
rs772761623 Chr.9: 113178634-113178634 on Build GRCh38
rs915347103 Chr.9: 113178714-113178714 on Build GRCh38
rs751790990 Chr.9: 113178709-113178709 on Build GRCh38
rs909749009 Chr.9: 113178881-113178881 on Build GRCh38
rs754770834 Chr.9: 113178581-113178581 on Build GRCh38
rs10118914 Chr.9: 113178864-113178864 on Build GRCh38
rs551544554 Chr.9: 113178858-113178858 on Build GRCh38
rs754715896 Chr.9: 113178715-113178715 on Build GRCh38
rs769661797 Chr.9: 113178621-113178621 on Build GRCh38
rs377595231 Chr.9: 113178775-113178775 on Build GRCh38
rs376621439 Chr.9: 113178754-113178754 on Build GRCh38
rs778846789 Chr.9: 113178474-113178474 on Build GRCh38
rs963718727 Chr.9: 113178897-113178897 on Build GRCh38
rs748151553 Chr.9: 113178595-113178595 on Build GRCh38
rs943929905 Chr.9: 113178525-113178525 on Build GRCh38
rs537672339 Chr.9: 113178903-113178903 on Build GRCh38
rs763398410 Chr.9: 113178618-113178618 on Build GRCh38
rs373476772 Chr.9: 113178788-113178788 on Build GRCh38
rs1028211483 Chr.9: 113178780-113178780 on Build GRCh38
rs774151809 Chr.9: 113178645-113178645 on Build GRCh38
rs371408703 Chr.9: 113178655-113178655 on Build GRCh38
rs762959561 Chr.9: 113178790-113178790 on Build GRCh38
rs200512984 Chr.9: 113178662-113178662 on Build GRCh38
rs774060142 Chr.9: 113178722-113178722 on Build GRCh38
rs545057011 Chr.9: 113178520-113178520 on Build GRCh38
rs778660528 Chr.9: 113178752-113178752 on Build GRCh38
rs751682332 Chr.9: 113178814-113178814 on Build GRCh38
rs1048439232 Chr.9: 113178614-113178614 on Build GRCh38
rs780936963 Chr.9: 113178723-113178723 on Build GRCh38
rs3810912 Chr.9: 113178766-113178766 on Build GRCh38
rs761256778 Chr.9: 113178835-113178836 on Build GRCh38
rs946814941 Chr.9: 113178727-113178727 on Build GRCh38
rs758470743 Chr.9: 113178688-113178688 on Build GRCh38
rs748766589 Chr.9: 113178747-113178747 on Build GRCh38
rs577461675 Chr.9: 113178643-113178643 on Build GRCh38
rs752487229 Chr.9: 113178732-113178732 on Build GRCh38
rs759734123 Chr.9: 113178836-113178836 on Build GRCh38
rs749700535 Chr.9: 113178855-113178855 on Build GRCh38
rs1021336265 Chr.9: 113178762-113178762 on Build GRCh38
rs367970889 Chr.9: 113178649-113178649 on Build GRCh38
rs573289215 Chr.9: 113178499-113178499 on Build GRCh38
rs147811705 Chr.9: 113178478-113178478 on Build GRCh38
rs766517438 Chr.9: 113178706-113178706 on Build GRCh38
rs993624293 Chr.9: 113178482-113178482 on Build GRCh38
rs918514946 Chr.9: 113178836-113178836 on Build GRCh38
rs763614342 Chr.9: 113178714-113178715 on Build GRCh38
rs12684370 Chr.9: 113178699-113178699 on Build GRCh38
rs976636657 Chr.9: 113178819-113178819 on Build GRCh38
rs765712264 Chr.9: 113178667-113178667 on Build GRCh38

View Full Product Details