Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113354231-113354620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSPRY
Entrez Gene ID
54836
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
UniGene
Hs.614517
Gene Chromosome Location
Chr.9: 113349532-113371233 on Build GRCh38
Transcripts
NM_001317943.1 NM_001317944.1 NM_017688.2 XM_006717149.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350859 c.300+8C>G p.? Chr.9: 113354346-113354346 on Build GRCh38
3320095 c.218A>T p.Q73L Chr.9: 113354256-113354256 on Build GRCh38
4834809 c.202-1G>A p.? Chr.9: 113354239-113354239 on Build GRCh38
3375038 c.208A>G p.I70V Chr.9: 113354246-113354246 on Build GRCh38
3952234 c.202-2A>G p.? Chr.9: 113354238-113354238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992095827 Chr.9: 113354395-113354395 on Build GRCh38
rs933535929 Chr.9: 113354470-113354470 on Build GRCh38
rs557164626 Chr.9: 113354308-113354308 on Build GRCh38
rs529103092 Chr.9: 113354486-113354486 on Build GRCh38
rs970141638 Chr.9: 113354340-113354340 on Build GRCh38
rs774157751 Chr.9: 113354291-113354291 on Build GRCh38
rs774139609 Chr.9: 113354374-113354374 on Build GRCh38
rs748770893 Chr.9: 113354358-113354358 on Build GRCh38
rs374063532 Chr.9: 113354372-113354372 on Build GRCh38
rs368306479 Chr.9: 113354316-113354316 on Build GRCh38
rs762404330 Chr.9: 113354288-113354288 on Build GRCh38
rs1053292835 Chr.9: 113354475-113354475 on Build GRCh38
rs573475250 Chr.9: 113354347-113354347 on Build GRCh38
rs10981763 Chr.9: 113354346-113354346 on Build GRCh38
rs940623912 Chr.9: 113354414-113354414 on Build GRCh38
rs772823188 Chr.9: 113354285-113354285 on Build GRCh38
rs764616023 Chr.9: 113354303-113354303 on Build GRCh38
rs201512287 Chr.9: 113354292-113354292 on Build GRCh38
rs74505973 Chr.9: 113354493-113354493 on Build GRCh38
rs781182938 Chr.9: 113354568-113354568 on Build GRCh38
rs377571038 Chr.9: 113354270-113354270 on Build GRCh38
rs371028609 Chr.9: 113354464-113354464 on Build GRCh38
rs753561751 Chr.9: 113354331-113354331 on Build GRCh38
rs769168556 Chr.9: 113354283-113354283 on Build GRCh38
rs561340099 Chr.9: 113354485-113354485 on Build GRCh38
rs1055844150 Chr.9: 113354477-113354477 on Build GRCh38
rs903411103 Chr.9: 113354492-113354492 on Build GRCh38
rs544732765 Chr.9: 113354478-113354478 on Build GRCh38
rs373291577 Chr.9: 113354305-113354305 on Build GRCh38
rs762490754 Chr.9: 113354309-113354309 on Build GRCh38
rs565262878 Chr.9: 113354438-113354438 on Build GRCh38
rs545120786 Chr.9: 113354425-113354425 on Build GRCh38
rs920460897 Chr.9: 113354453-113354453 on Build GRCh38
rs759317375 Chr.9: 113354384-113354384 on Build GRCh38
rs919208300 Chr.9: 113354404-113354404 on Build GRCh38
rs780185557 Chr.9: 113354317-113354317 on Build GRCh38
rs34944005 Chr.9: 113354501-113354502 on Build GRCh38
rs1054615370 Chr.9: 113354511-113354511 on Build GRCh38
rs1028708415 Chr.9: 113354586-113354586 on Build GRCh38
rs891806843 Chr.9: 113354311-113354311 on Build GRCh38
rs943145601 Chr.9: 113354444-113354444 on Build GRCh38
rs1011567154 Chr.9: 113354312-113354312 on Build GRCh38
rs768445660 Chr.9: 113354253-113354253 on Build GRCh38
rs923465885 Chr.9: 113354465-113354465 on Build GRCh38
rs987551579 Chr.9: 113354360-113354360 on Build GRCh38
rs984477297 Chr.9: 113354430-113354430 on Build GRCh38
rs182334591 Chr.9: 113354345-113354345 on Build GRCh38
rs900041300 Chr.9: 113354535-113354536 on Build GRCh38
rs575842489 Chr.9: 113354442-113354442 on Build GRCh38
rs907757866 Chr.9: 113354393-113354393 on Build GRCh38
rs747299077 Chr.9: 113354319-113354319 on Build GRCh38
rs950044977 Chr.9: 113354479-113354479 on Build GRCh38
rs754285338 Chr.9: 113354304-113354304 on Build GRCh38
rs776546213 Chr.9: 113354258-113354258 on Build GRCh38
rs905916285 Chr.9: 113354519-113354519 on Build GRCh38
rs754754848 Chr.9: 113354343-113354343 on Build GRCh38

View Full Product Details