Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114167586-114168078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
UniGene
Hs.494892
Gene Chromosome Location
Chr.9: 114154271-114312516 on Build GRCh38
Transcripts
NM_032888.3 XM_006717308.2 XM_011519138.2 XM_011519140.1 XM_011519142.2 XM_011519143.2 XM_011519144.2 XM_017015239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
357783 c.135T>C p.D45D Chr.9: 114167690-114167690 on Build GRCh38
3320847 c.462G>A p.G154G Chr.9: 114168017-114168017 on Build GRCh38
3320845 c.396C>T p.P132P Chr.9: 114167951-114167951 on Build GRCh38
1243168 c.236G>A p.R79Q Chr.9: 114167791-114167791 on Build GRCh38
3320844 c.353G>A p.R118H Chr.9: 114167908-114167908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768434921 Chr.9: 114167979-114167979 on Build GRCh38
rs750179344 Chr.9: 114167745-114167745 on Build GRCh38
rs780968064 Chr.9: 114167685-114167685 on Build GRCh38
rs150759476 Chr.9: 114167851-114167851 on Build GRCh38
rs761575685 Chr.9: 114167663-114167663 on Build GRCh38
rs141061686 Chr.9: 114167702-114167702 on Build GRCh38
rs779308507 Chr.9: 114167906-114167906 on Build GRCh38
rs143341894 Chr.9: 114168014-114168014 on Build GRCh38
rs766143475 Chr.9: 114167806-114167806 on Build GRCh38
rs761414386 Chr.9: 114167924-114167924 on Build GRCh38
rs2567707 Chr.9: 114167820-114167820 on Build GRCh38
rs770533328 Chr.9: 114168052-114168052 on Build GRCh38
rs367798942 Chr.9: 114167672-114167672 on Build GRCh38
rs79824663 Chr.9: 114167813-114167813 on Build GRCh38
rs769153208 Chr.9: 114167791-114167791 on Build GRCh38
rs777313693 Chr.9: 114167779-114167779 on Build GRCh38
rs775952838 Chr.9: 114167920-114167920 on Build GRCh38
rs377686014 Chr.9: 114167983-114167983 on Build GRCh38
rs754485502 Chr.9: 114167682-114167682 on Build GRCh38
rs371597072 Chr.9: 114167706-114167706 on Build GRCh38
rs747384879 Chr.9: 114167833-114167833 on Build GRCh38
rs926781124 Chr.9: 114167808-114167808 on Build GRCh38
rs201660579 Chr.9: 114167771-114167771 on Build GRCh38
rs748184039 Chr.9: 114168039-114168039 on Build GRCh38
rs750538488 Chr.9: 114168012-114168012 on Build GRCh38
rs111969026 Chr.9: 114167946-114167946 on Build GRCh38
rs147354366 Chr.9: 114167749-114167749 on Build GRCh38
rs575113764 Chr.9: 114167996-114167996 on Build GRCh38
rs759427839 Chr.9: 114167863-114167863 on Build GRCh38
rs754520598 Chr.9: 114167754-114167754 on Build GRCh38
rs753526489 Chr.9: 114167873-114167873 on Build GRCh38
rs149948860 Chr.9: 114167925-114167925 on Build GRCh38
rs755371196 Chr.9: 114167880-114167880 on Build GRCh38
rs190770493 Chr.9: 114167786-114167786 on Build GRCh38
rs766032017 Chr.9: 114167993-114167993 on Build GRCh38
rs143398547 Chr.9: 114167796-114167796 on Build GRCh38
rs760243902 Chr.9: 114167867-114167867 on Build GRCh38
rs749537461 Chr.9: 114167856-114167856 on Build GRCh38
rs746153745 Chr.9: 114167968-114167968 on Build GRCh38
rs544568291 Chr.9: 114167751-114167751 on Build GRCh38
rs746839506 Chr.9: 114167976-114167976 on Build GRCh38
rs751040547 Chr.9: 114167680-114167680 on Build GRCh38
rs754511035 Chr.9: 114167964-114167964 on Build GRCh38
rs893195062 Chr.9: 114167853-114167853 on Build GRCh38
rs1044459381 Chr.9: 114168015-114168015 on Build GRCh38
rs748820491 Chr.9: 114168051-114168051 on Build GRCh38
rs781683498 Chr.9: 114167841-114167841 on Build GRCh38
rs747530985 Chr.9: 114167790-114167790 on Build GRCh38
rs775896019 Chr.9: 114167726-114167726 on Build GRCh38
rs771225683 Chr.9: 114167997-114167997 on Build GRCh38
rs141549766 Chr.9: 114167707-114167707 on Build GRCh38
rs779873134 Chr.9: 114167675-114167675 on Build GRCh38
rs999300997 Chr.9: 114167624-114167624 on Build GRCh38
rs150242638 Chr.9: 114167725-114167725 on Build GRCh38
rs142192832 Chr.9: 114167831-114167831 on Build GRCh38
rs756213906 Chr.9: 114167888-114167888 on Build GRCh38
rs760780114 Chr.9: 114167728-114167728 on Build GRCh38
rs776665987 Chr.9: 114167980-114167980 on Build GRCh38
rs201917514 Chr.9: 114167819-114167819 on Build GRCh38
rs571516072 Chr.9: 114168018-114168018 on Build GRCh38
rs769725938 Chr.9: 114168043-114168043 on Build GRCh38
rs146793976 Chr.9: 114167952-114167952 on Build GRCh38
rs138977561 Chr.9: 114167732-114167732 on Build GRCh38
rs779654396 Chr.9: 114167742-114167743 on Build GRCh38
rs538733879 Chr.9: 114167981-114167981 on Build GRCh38
rs370978325 Chr.9: 114168003-114168003 on Build GRCh38
rs564819689 Chr.9: 114167735-114167735 on Build GRCh38
rs558626202 Chr.9: 114167982-114167982 on Build GRCh38
rs762687889 Chr.9: 114167747-114167747 on Build GRCh38
rs74930448 Chr.9: 114167812-114167812 on Build GRCh38
rs148339655 Chr.9: 114168038-114168038 on Build GRCh38
rs767204819 Chr.9: 114167768-114167768 on Build GRCh38
rs376638194 Chr.9: 114167644-114167644 on Build GRCh38
rs774343750 Chr.9: 114167807-114167807 on Build GRCh38
rs750124954 Chr.9: 114167927-114167927 on Build GRCh38
rs747870284 Chr.9: 114167699-114167699 on Build GRCh38
rs532200840 Chr.9: 114167652-114167652 on Build GRCh38
rs147026086 Chr.9: 114167960-114167960 on Build GRCh38
rs775586585 Chr.9: 114167660-114167660 on Build GRCh38
rs372265305 Chr.9: 114167919-114167919 on Build GRCh38
rs140539074 Chr.9: 114167954-114167954 on Build GRCh38
rs373943616 Chr.9: 114167661-114167661 on Build GRCh38
rs372211916 Chr.9: 114167804-114167804 on Build GRCh38
rs369477310 Chr.9: 114167972-114167972 on Build GRCh38
rs754848059 Chr.9: 114168029-114168029 on Build GRCh38
rs375672077 Chr.9: 114167858-114167858 on Build GRCh38
rs767523424 Chr.9: 114167864-114167864 on Build GRCh38
rs767189802 Chr.9: 114167617-114167617 on Build GRCh38
rs137868129 Chr.9: 114167785-114167785 on Build GRCh38
rs201093507 Chr.9: 114167877-114167877 on Build GRCh38
rs146589272 Chr.9: 114167963-114167963 on Build GRCh38
rs1041670066 Chr.9: 114167824-114167824 on Build GRCh38
rs764478171 Chr.9: 114168002-114168002 on Build GRCh38
rs763574320 Chr.9: 114167869-114167869 on Build GRCh38
rs774988270 Chr.9: 114167651-114167651 on Build GRCh38
rs772650006 Chr.9: 114168046-114168046 on Build GRCh38
rs750186045 Chr.9: 114167669-114167669 on Build GRCh38
rs762524310 Chr.9: 114167991-114167991 on Build GRCh38
rs753486760 Chr.9: 114167830-114167830 on Build GRCh38
rs369829082 Chr.9: 114167859-114167859 on Build GRCh38
rs200695070 Chr.9: 114167770-114167770 on Build GRCh38
rs771010276 Chr.9: 114167901-114167901 on Build GRCh38
rs776631973 Chr.9: 114167795-114167795 on Build GRCh38
rs373617346 Chr.9: 114167973-114167973 on Build GRCh38
rs745631704 Chr.9: 114167907-114167907 on Build GRCh38
rs778369339 Chr.9: 114167780-114167780 on Build GRCh38
rs772045340 Chr.9: 114167908-114167908 on Build GRCh38
rs773468575 Chr.9: 114167855-114167855 on Build GRCh38
rs903165829 Chr.9: 114167623-114167623 on Build GRCh38
rs377185109 Chr.9: 114167948-114167948 on Build GRCh38
rs765082448 Chr.9: 114167664-114167664 on Build GRCh38
rs368213196 Chr.9: 114167797-114167797 on Build GRCh38
rs765454716 Chr.9: 114167741-114167741 on Build GRCh38
rs1040369729 Chr.9: 114167610-114167610 on Build GRCh38
rs938819171 Chr.9: 114168054-114168054 on Build GRCh38
rs780214288 Chr.9: 114167832-114167832 on Build GRCh38
rs771469072 Chr.9: 114167649-114167649 on Build GRCh38
rs755425920 Chr.9: 114167835-114167835 on Build GRCh38
rs766068706 Chr.9: 114167959-114167959 on Build GRCh38
rs374714030 Chr.9: 114167897-114167897 on Build GRCh38
rs749400167 Chr.9: 114167900-114167900 on Build GRCh38
rs764962397 Chr.9: 114167878-114167878 on Build GRCh38
rs2567706 Chr.9: 114167914-114167914 on Build GRCh38
rs762785718 Chr.9: 114167805-114167805 on Build GRCh38
rs746746222 Chr.9: 114168019-114168019 on Build GRCh38
rs763774486 Chr.9: 114167826-114167826 on Build GRCh38
rs781648277 Chr.9: 114167886-114167886 on Build GRCh38
rs772465120 Chr.9: 114167654-114167654 on Build GRCh38
rs775558821 Chr.9: 114167909-114167909 on Build GRCh38
rs753062733 Chr.9: 114167951-114167951 on Build GRCh38
rs748194962 Chr.9: 114167850-114167850 on Build GRCh38
rs74339956 Chr.9: 114167733-114167733 on Build GRCh38
rs1023106800 Chr.9: 114167696-114167696 on Build GRCh38
rs774093023 Chr.9: 114168056-114168056 on Build GRCh38
rs757275050 Chr.9: 114168011-114168011 on Build GRCh38

View Full Product Details