Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 115141740-115142175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEC1
Entrez Gene ID
50514
Gene Name
deleted in esophageal cancer 1
Gene Aliases
CTS9
UniGene
Hs.148841
Gene Chromosome Location
Chr.9: 115141818-115402644 on Build GRCh38
Transcripts
NM_017418.2

SNP Detail

SNP ID Chromosome Location
rs1044263264 Chr.9: 115141848-115141848 on Build GRCh38
rs1017894482 Chr.9: 115141928-115141928 on Build GRCh38
rs371904610 Chr.9: 115142019-115142019 on Build GRCh38
rs973931512 Chr.9: 115141784-115141784 on Build GRCh38
rs966282640 Chr.9: 115141932-115141932 on Build GRCh38
rs573982844 Chr.9: 115141877-115141877 on Build GRCh38
rs145733377 Chr.9: 115142085-115142085 on Build GRCh38
rs911927771 Chr.9: 115142070-115142070 on Build GRCh38
rs563689597 Chr.9: 115141864-115141864 on Build GRCh38
rs560107688 Chr.9: 115141951-115141951 on Build GRCh38
rs931947387 Chr.9: 115141997-115141997 on Build GRCh38
rs896907067 Chr.9: 115141879-115141879 on Build GRCh38
rs944801612 Chr.9: 115142084-115142084 on Build GRCh38
rs894403918 Chr.9: 115142146-115142146 on Build GRCh38
rs781201929 Chr.9: 115141978-115141978 on Build GRCh38
rs115675257 Chr.9: 115141860-115141860 on Build GRCh38
rs113049916 Chr.9: 115141905-115141905 on Build GRCh38
rs200413477 Chr.9: 115141795-115141795 on Build GRCh38
rs963492069 Chr.9: 115141777-115141777 on Build GRCh38
rs552013084 Chr.9: 115141958-115141958 on Build GRCh38
rs927305119 Chr.9: 115141786-115141786 on Build GRCh38
rs914419177 Chr.9: 115141999-115141999 on Build GRCh38
rs979887039 Chr.9: 115142018-115142018 on Build GRCh38
rs930500730 Chr.9: 115142099-115142099 on Build GRCh38
rs989714061 Chr.9: 115141960-115141960 on Build GRCh38
rs914365329 Chr.9: 115141802-115141802 on Build GRCh38
rs368675638 Chr.9: 115141774-115141774 on Build GRCh38
rs1335331 Chr.9: 115141952-115141952 on Build GRCh38
rs995228545 Chr.9: 115141882-115141882 on Build GRCh38
rs997957941 Chr.9: 115141950-115141950 on Build GRCh38
rs772339857 Chr.9: 115141954-115141954 on Build GRCh38
rs887918960 Chr.9: 115141918-115141918 on Build GRCh38
rs897658898 Chr.9: 115142087-115142087 on Build GRCh38
rs1007721666 Chr.9: 115141923-115141923 on Build GRCh38
rs565769747 Chr.9: 115142098-115142098 on Build GRCh38
rs969893998 Chr.9: 115142016-115142016 on Build GRCh38
rs146233643 Chr.9: 115141786-115141787 on Build GRCh38
rs986170025 Chr.9: 115142036-115142036 on Build GRCh38
rs149772571 Chr.9: 115141810-115141810 on Build GRCh38
rs762826981 Chr.9: 115142044-115142044 on Build GRCh38
rs1036158594 Chr.9: 115141780-115141780 on Build GRCh38
rs1049436210 Chr.9: 115142139-115142140 on Build GRCh38
rs990225044 Chr.9: 115141971-115141971 on Build GRCh38
rs990394491 Chr.9: 115141783-115141783 on Build GRCh38
rs948656770 Chr.9: 115141821-115141821 on Build GRCh38
rs564780595 Chr.9: 115142015-115142015 on Build GRCh38
rs938369966 Chr.9: 115141862-115141862 on Build GRCh38

View Full Product Details