Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 115291649-115292170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEC1
Entrez Gene ID
50514
Gene Name
deleted in esophageal cancer 1
Gene Aliases
CTS9
UniGene
Hs.148841
Gene Chromosome Location
Chr.9: 115141818-115402644 on Build GRCh38
Transcripts
NM_017418.2

SNP Detail

SNP ID Chromosome Location
rs568173972 Chr.9: 115291887-115291887 on Build GRCh38
rs888701139 Chr.9: 115291749-115291749 on Build GRCh38
rs752406544 Chr.9: 115291681-115291681 on Build GRCh38
rs970372130 Chr.9: 115291805-115291805 on Build GRCh38
rs1030692222 Chr.9: 115292078-115292078 on Build GRCh38
rs532432860 Chr.9: 115292054-115292054 on Build GRCh38
rs956186030 Chr.9: 115291895-115291895 on Build GRCh38
rs560092641 Chr.9: 115292048-115292048 on Build GRCh38
rs887472127 Chr.9: 115291983-115291983 on Build GRCh38
rs569494783 Chr.9: 115292142-115292142 on Build GRCh38
rs538118763 Chr.9: 115291847-115291847 on Build GRCh38
rs569414567 Chr.9: 115291701-115291701 on Build GRCh38
rs1044971992 Chr.9: 115292004-115292004 on Build GRCh38
rs538094745 Chr.9: 115291711-115291711 on Build GRCh38
rs2285315 Chr.9: 115291899-115291899 on Build GRCh38
rs566042707 Chr.9: 115291847-115291847 on Build GRCh38
rs1003182234 Chr.9: 115292048-115292049 on Build GRCh38
rs969032551 Chr.9: 115291933-115291933 on Build GRCh38
rs1003693708 Chr.9: 115291670-115291670 on Build GRCh38
rs998370990 Chr.9: 115292117-115292117 on Build GRCh38
rs139260237 Chr.9: 115291982-115291982 on Build GRCh38
rs964267193 Chr.9: 115291843-115291843 on Build GRCh38
rs546063396 Chr.9: 115291937-115291937 on Build GRCh38
rs897462561 Chr.9: 115291679-115291679 on Build GRCh38
rs541663450 Chr.9: 115291975-115291975 on Build GRCh38
rs1003214987 Chr.9: 115291808-115291808 on Build GRCh38
rs2285316 Chr.9: 115291903-115291903 on Build GRCh38
rs540205715 Chr.9: 115291997-115291997 on Build GRCh38
rs766962084 Chr.9: 115291849-115291849 on Build GRCh38
rs576945004 Chr.9: 115291925-115291925 on Build GRCh38
rs980715610 Chr.9: 115291943-115291943 on Build GRCh38
rs772239240 Chr.9: 115291979-115291979 on Build GRCh38
rs552822856 Chr.9: 115292086-115292086 on Build GRCh38
rs926426038 Chr.9: 115291946-115291946 on Build GRCh38
rs1005798417 Chr.9: 115291797-115291797 on Build GRCh38
rs561005424 Chr.9: 115291978-115291978 on Build GRCh38
rs552620748 Chr.9: 115291676-115291676 on Build GRCh38
rs1056587912 Chr.9: 115291976-115291976 on Build GRCh38
rs939218442 Chr.9: 115291970-115291970 on Build GRCh38
rs1012315904 Chr.9: 115291753-115291753 on Build GRCh38
rs1018498450 Chr.9: 115292029-115292029 on Build GRCh38
rs187077847 Chr.9: 115291803-115291803 on Build GRCh38
rs556225518 Chr.9: 115291956-115291956 on Build GRCh38
rs899969798 Chr.9: 115292037-115292037 on Build GRCh38
rs576184089 Chr.9: 115291974-115291974 on Build GRCh38
rs906182772 Chr.9: 115292038-115292038 on Build GRCh38
rs777553820 Chr.9: 115291848-115291848 on Build GRCh38
rs767879193 Chr.9: 115291936-115291936 on Build GRCh38

View Full Product Details