Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120758431-120758940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW2
Entrez Gene ID
26190
Gene Name
F-box and WD repeat domain containing 2
Gene Aliases
FBW2, Fwd2, Md6
UniGene
Hs.494985
Gene Chromosome Location
Chr.9: 120756976-120793471 on Build GRCh38
Transcripts
NM_012164.3

SNP Detail

SNP ID Chromosome Location
rs1050922523 Chr.9: 120758751-120758751 on Build GRCh38
rs928097633 Chr.9: 120758611-120758611 on Build GRCh38
rs1042191764 Chr.9: 120758690-120758690 on Build GRCh38
rs368309742 Chr.9: 120758552-120758552 on Build GRCh38
rs959477297 Chr.9: 120758915-120758915 on Build GRCh38
rs921465615 Chr.9: 120758775-120758775 on Build GRCh38
rs938285278 Chr.9: 120758638-120758638 on Build GRCh38
rs973327277 Chr.9: 120758456-120758456 on Build GRCh38
rs34096879 Chr.9: 120758769-120758770 on Build GRCh38
rs569828501 Chr.9: 120758589-120758589 on Build GRCh38
rs1003744596 Chr.9: 120758892-120758892 on Build GRCh38
rs774673560 Chr.9: 120758686-120758686 on Build GRCh38
rs147854541 Chr.9: 120758772-120758772 on Build GRCh38
rs183475703 Chr.9: 120758510-120758510 on Build GRCh38
rs973536550 Chr.9: 120758576-120758576 on Build GRCh38
rs1047144427 Chr.9: 120758492-120758492 on Build GRCh38
rs982128382 Chr.9: 120758844-120758844 on Build GRCh38
rs748585857 Chr.9: 120758500-120758500 on Build GRCh38
rs541376206 Chr.9: 120758697-120758697 on Build GRCh38
rs950918406 Chr.9: 120758602-120758602 on Build GRCh38
rs1017615634 Chr.9: 120758553-120758553 on Build GRCh38
rs767483743 Chr.9: 120758798-120758798 on Build GRCh38
rs933739522 Chr.9: 120758907-120758907 on Build GRCh38
rs757111602 Chr.9: 120758835-120758835 on Build GRCh38
rs1003282802 Chr.9: 120758617-120758617 on Build GRCh38
rs928150000 Chr.9: 120758845-120758845 on Build GRCh38
rs920480415 Chr.9: 120758682-120758682 on Build GRCh38
rs953943668 Chr.9: 120758530-120758530 on Build GRCh38
rs902373424 Chr.9: 120758474-120758474 on Build GRCh38
rs145352296 Chr.9: 120758788-120758788 on Build GRCh38
rs1055756253 Chr.9: 120758649-120758649 on Build GRCh38
rs141516652 Chr.9: 120758776-120758776 on Build GRCh38
rs537892785 Chr.9: 120758540-120758540 on Build GRCh38
rs1032586652 Chr.9: 120758551-120758551 on Build GRCh38
rs187707130 Chr.9: 120758678-120758678 on Build GRCh38
rs753880789 Chr.9: 120758828-120758828 on Build GRCh38
rs763315469 Chr.9: 120758631-120758631 on Build GRCh38
rs1021102109 Chr.9: 120758467-120758467 on Build GRCh38
rs543594805 Chr.9: 120758855-120758855 on Build GRCh38
rs1041637402 Chr.9: 120758518-120758518 on Build GRCh38
rs946439530 Chr.9: 120758876-120758876 on Build GRCh38
rs531460123 Chr.9: 120758895-120758895 on Build GRCh38
rs773680000 Chr.9: 120758583-120758583 on Build GRCh38
rs35738692 Chr.9: 120758875-120758875 on Build GRCh38
rs982244231 Chr.9: 120758603-120758603 on Build GRCh38
rs73541843 Chr.9: 120758660-120758660 on Build GRCh38
rs1006824346 Chr.9: 120758771-120758771 on Build GRCh38
rs566357057 Chr.9: 120758499-120758499 on Build GRCh38
rs564429745 Chr.9: 120758577-120758577 on Build GRCh38
rs554700982 Chr.9: 120758567-120758567 on Build GRCh38
rs970436902 Chr.9: 120758615-120758615 on Build GRCh38
rs558986916 Chr.9: 120758673-120758673 on Build GRCh38
rs908535328 Chr.9: 120758763-120758763 on Build GRCh38
rs991113346 Chr.9: 120758873-120758873 on Build GRCh38
rs889593038 Chr.9: 120758769-120758769 on Build GRCh38
rs761255176 Chr.9: 120758632-120758632 on Build GRCh38
rs1042076531 Chr.9: 120758879-120758879 on Build GRCh38
rs997894599 Chr.9: 120758734-120758734 on Build GRCh38

View Full Product Details