Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120761541-120762041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW2
Entrez Gene ID
26190
Gene Name
F-box and WD repeat domain containing 2
Gene Aliases
FBW2, Fwd2, Md6
UniGene
Hs.494985
Gene Chromosome Location
Chr.9: 120756976-120793471 on Build GRCh38
Transcripts
NM_012164.3 XM_005251910.3 XM_005251913.2 XM_006717049.3 XM_006717051.2 XM_011518518.1 XM_017014615.1 XM_017014616.1 XM_017014617.1

SNP Detail

SNP ID Chromosome Location
rs114019566 Chr.9: 120761996-120761996 on Build GRCh38
rs1042773346 Chr.9: 120761784-120761784 on Build GRCh38
rs1036438076 Chr.9: 120761693-120761693 on Build GRCh38
rs77186830 Chr.9: 120761757-120761757 on Build GRCh38
rs767033742 Chr.9: 120761928-120761928 on Build GRCh38
rs1028754767 Chr.9: 120761939-120761939 on Build GRCh38
rs917481973 Chr.9: 120761772-120761772 on Build GRCh38
rs925577586 Chr.9: 120761835-120761835 on Build GRCh38
rs370972903 Chr.9: 120761627-120761627 on Build GRCh38
rs1007798023 Chr.9: 120761777-120761777 on Build GRCh38
rs891659495 Chr.9: 120761845-120761845 on Build GRCh38
rs375480278 Chr.9: 120761970-120761970 on Build GRCh38
rs544734840 Chr.9: 120762011-120762011 on Build GRCh38
rs900418902 Chr.9: 120761910-120761910 on Build GRCh38
rs1005615438 Chr.9: 120761984-120761984 on Build GRCh38
rs781636186 Chr.9: 120761757-120761757 on Build GRCh38
rs567849993 Chr.9: 120761606-120761606 on Build GRCh38
rs575209944 Chr.9: 120761717-120761717 on Build GRCh38
rs904182140 Chr.9: 120761793-120761793 on Build GRCh38
rs909197502 Chr.9: 120761659-120761659 on Build GRCh38
rs1028283092 Chr.9: 120761628-120761628 on Build GRCh38
rs1052835013 Chr.9: 120761842-120761842 on Build GRCh38
rs952805302 Chr.9: 120761632-120761632 on Build GRCh38
rs915795416 Chr.9: 120761584-120761584 on Build GRCh38
rs540865560 Chr.9: 120761840-120761840 on Build GRCh38
rs944527897 Chr.9: 120761858-120761858 on Build GRCh38
rs552740443 Chr.9: 120761594-120761594 on Build GRCh38
rs969970831 Chr.9: 120761894-120761894 on Build GRCh38
rs762924431 Chr.9: 120762008-120762008 on Build GRCh38
rs368309210 Chr.9: 120762015-120762015 on Build GRCh38
rs1031380157 Chr.9: 120761812-120761812 on Build GRCh38
rs942147066 Chr.9: 120761692-120761692 on Build GRCh38
rs768787142 Chr.9: 120761841-120761841 on Build GRCh38
rs999050775 Chr.9: 120761811-120761812 on Build GRCh38
rs997249384 Chr.9: 120761618-120761618 on Build GRCh38
rs535156742 Chr.9: 120761608-120761608 on Build GRCh38
rs959897286 Chr.9: 120761571-120761571 on Build GRCh38
rs961558489 Chr.9: 120762002-120762002 on Build GRCh38
rs144437174 Chr.9: 120761714-120761714 on Build GRCh38
rs189121277 Chr.9: 120761831-120761831 on Build GRCh38
rs558427985 Chr.9: 120761790-120761790 on Build GRCh38
rs776658701 Chr.9: 120761911-120761911 on Build GRCh38
rs972568270 Chr.9: 120761976-120761976 on Build GRCh38
rs1040328071 Chr.9: 120761876-120761876 on Build GRCh38
rs984139597 Chr.9: 120761703-120761703 on Build GRCh38
rs993156682 Chr.9: 120761572-120761572 on Build GRCh38
rs948454524 Chr.9: 120761812-120761812 on Build GRCh38

View Full Product Details