Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120763414-120763949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW2
Entrez Gene ID
26190
Gene Name
F-box and WD repeat domain containing 2
Gene Aliases
FBW2, Fwd2, Md6
UniGene
Hs.494985
Gene Chromosome Location
Chr.9: 120756976-120793471 on Build GRCh38
Transcripts
NM_012164.3 XM_005251910.3 XM_005251913.2 XM_006717049.3 XM_006717051.2 XM_011518518.1 XM_017014615.1 XM_017014616.1 XM_017014617.1

SNP Detail

SNP ID Chromosome Location
rs142306466 Chr.9: 120763873-120763873 on Build GRCh38
rs1001745568 Chr.9: 120763633-120763633 on Build GRCh38
rs191217762 Chr.9: 120763888-120763888 on Build GRCh38
rs186061129 Chr.9: 120763872-120763872 on Build GRCh38
rs542563560 Chr.9: 120763840-120763840 on Build GRCh38
rs538472533 Chr.9: 120763599-120763599 on Build GRCh38
rs1039638603 Chr.9: 120763465-120763465 on Build GRCh38
rs149146460 Chr.9: 120763850-120763850 on Build GRCh38
rs1005950872 Chr.9: 120763595-120763595 on Build GRCh38
rs1054556678 Chr.9: 120763549-120763549 on Build GRCh38
rs1055989981 Chr.9: 120763868-120763868 on Build GRCh38
rs981070565 Chr.9: 120763734-120763734 on Build GRCh38
rs1024929925 Chr.9: 120763626-120763626 on Build GRCh38
rs572116185 Chr.9: 120763810-120763810 on Build GRCh38
rs745804865 Chr.9: 120763899-120763899 on Build GRCh38
rs374012383 Chr.9: 120763759-120763759 on Build GRCh38
rs75016429 Chr.9: 120763479-120763479 on Build GRCh38
rs914580296 Chr.9: 120763799-120763799 on Build GRCh38
rs374489270 Chr.9: 120763680-120763680 on Build GRCh38
rs962366986 Chr.9: 120763642-120763642 on Build GRCh38
rs902096179 Chr.9: 120763518-120763518 on Build GRCh38
rs900504496 Chr.9: 120763504-120763504 on Build GRCh38
rs186247564 Chr.9: 120763552-120763552 on Build GRCh38
rs1025766575 Chr.9: 120763875-120763875 on Build GRCh38
rs879645035 Chr.9: 120763867-120763867 on Build GRCh38
rs1015840480 Chr.9: 120763885-120763885 on Build GRCh38
rs925160404 Chr.9: 120763753-120763753 on Build GRCh38
rs986558074 Chr.9: 120763710-120763710 on Build GRCh38
rs751776978 Chr.9: 120763542-120763542 on Build GRCh38
rs927300871 Chr.9: 120763751-120763751 on Build GRCh38
rs902121302 Chr.9: 120763835-120763835 on Build GRCh38
rs3750496 Chr.9: 120763892-120763892 on Build GRCh38
rs578068982 Chr.9: 120763706-120763706 on Build GRCh38
rs577189306 Chr.9: 120763480-120763480 on Build GRCh38
rs540886301 Chr.9: 120763627-120763627 on Build GRCh38
rs146822476 Chr.9: 120763889-120763889 on Build GRCh38
rs1034530497 Chr.9: 120763634-120763634 on Build GRCh38
rs190656220 Chr.9: 120763715-120763715 on Build GRCh38
rs891901686 Chr.9: 120763556-120763556 on Build GRCh38
rs1010310585 Chr.9: 120763571-120763571 on Build GRCh38
rs980603529 Chr.9: 120763920-120763920 on Build GRCh38
rs535178276 Chr.9: 120763682-120763682 on Build GRCh38
rs556465686 Chr.9: 120763697-120763697 on Build GRCh38
rs367836642 Chr.9: 120763670-120763670 on Build GRCh38
rs1015954984 Chr.9: 120763616-120763616 on Build GRCh38
rs933484245 Chr.9: 120763849-120763849 on Build GRCh38
rs907389580 Chr.9: 120763836-120763836 on Build GRCh38
rs888759873 Chr.9: 120763572-120763572 on Build GRCh38
rs990266489 Chr.9: 120763790-120763790 on Build GRCh38
rs1054894657 Chr.9: 120763782-120763782 on Build GRCh38
rs182890353 Chr.9: 120763801-120763801 on Build GRCh38
rs1042400823 Chr.9: 120763450-120763450 on Build GRCh38

View Full Product Details