Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121107521-121108035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTRL
Entrez Gene ID
11064
Gene Name
centriolin
Gene Aliases
CEP1, CEP110, FAN, bA165P4.1
UniGene
Hs.653263
Gene Chromosome Location
Chr.9: 121074778-121177610 on Build GRCh38
Transcripts
NM_007018.4 XM_005251679.3 XM_006716940.2 XM_006716944.3 XM_011518166.1 XM_011518167.1 XM_011518168.1 XM_011518169.2 XM_011518170.1 XM_011518172.2 XM_017014223.1 XM_017014224.1 XM_017014225.1 XM_017014227.1 XM_017014228.1 XM_017014229.1 XM_017014230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5175126 c.876A>T p.K292N Chr.9: 121107869-121107869 on Build GRCh38
1104736 c.894G>T p.E298D Chr.9: 121107887-121107887 on Build GRCh38
7361876 c.959G>A p.S320N Chr.9: 121107952-121107952 on Build GRCh38
486997 c.913A>T p.K305* Chr.9: 121107906-121107906 on Build GRCh38
6992534 c.994A>C p.N332H Chr.9: 121107987-121107987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756150242 Chr.9: 121107875-121107875 on Build GRCh38
rs947917646 Chr.9: 121107760-121107760 on Build GRCh38
rs147793533 Chr.9: 121107993-121107993 on Build GRCh38
rs936097754 Chr.9: 121107881-121107881 on Build GRCh38
rs766225655 Chr.9: 121107949-121107950 on Build GRCh38
rs767149339 Chr.9: 121107772-121107775 on Build GRCh38
rs762182765 Chr.9: 121107965-121107965 on Build GRCh38
rs747969832 Chr.9: 121107898-121107898 on Build GRCh38
rs761142043 Chr.9: 121107812-121107812 on Build GRCh38
rs62578443 Chr.9: 121107857-121107857 on Build GRCh38
rs766753998 Chr.9: 121107996-121107996 on Build GRCh38
rs527516067 Chr.9: 121107562-121107562 on Build GRCh38
rs765886661 Chr.9: 121107807-121107807 on Build GRCh38
rs747938207 Chr.9: 121107756-121107756 on Build GRCh38
rs781044359 Chr.9: 121107890-121107890 on Build GRCh38
rs1043520902 Chr.9: 121107848-121107848 on Build GRCh38
rs920373858 Chr.9: 121107554-121107554 on Build GRCh38
rs569197854 Chr.9: 121107833-121107833 on Build GRCh38
rs780309800 Chr.9: 121107870-121107870 on Build GRCh38
rs754940616 Chr.9: 121107879-121107879 on Build GRCh38
rs960078277 Chr.9: 121107751-121107751 on Build GRCh38
rs866669429 Chr.9: 121107697-121107697 on Build GRCh38
rs188753500 Chr.9: 121107924-121107924 on Build GRCh38
rs764657555 Chr.9: 121107962-121107962 on Build GRCh38
rs747122850 Chr.9: 121107873-121107873 on Build GRCh38
rs765206732 Chr.9: 121107689-121107689 on Build GRCh38
rs758506759 Chr.9: 121107867-121107867 on Build GRCh38
rs769479358 Chr.9: 121107904-121107904 on Build GRCh38
rs754217349 Chr.9: 121107840-121107840 on Build GRCh38
rs777013192 Chr.9: 121107963-121107963 on Build GRCh38
rs907550327 Chr.9: 121107601-121107601 on Build GRCh38
rs144350374 Chr.9: 121107952-121107952 on Build GRCh38
rs769470414 Chr.9: 121107782-121107782 on Build GRCh38
rs757630026 Chr.9: 121107842-121107842 on Build GRCh38
rs1021652007 Chr.9: 121107614-121107614 on Build GRCh38
rs867570634 Chr.9: 121107841-121107841 on Build GRCh38
rs77588859 Chr.9: 121107611-121107611 on Build GRCh38
rs141957736 Chr.9: 121107946-121107946 on Build GRCh38
rs111522049 Chr.9: 121107667-121107667 on Build GRCh38
rs977123533 Chr.9: 121107642-121107642 on Build GRCh38
rs916187232 Chr.9: 121107806-121107806 on Build GRCh38
rs750627071 Chr.9: 121107866-121107866 on Build GRCh38
rs969241886 Chr.9: 121107649-121107649 on Build GRCh38
rs367816310 Chr.9: 121107936-121107936 on Build GRCh38
rs547623052 Chr.9: 121107714-121107715 on Build GRCh38
rs550962825 Chr.9: 121107668-121107668 on Build GRCh38
rs549264644 Chr.9: 121107573-121107573 on Build GRCh38
rs372623233 Chr.9: 121107844-121107844 on Build GRCh38
rs750317819 Chr.9: 121107831-121107832 on Build GRCh38
rs953856406 Chr.9: 121107581-121107581 on Build GRCh38
rs776853842 Chr.9: 121107614-121107620 on Build GRCh38
rs758661060 Chr.9: 121107991-121107991 on Build GRCh38
rs924569695 Chr.9: 121107726-121107726 on Build GRCh38
rs10985157 Chr.9: 121107558-121107558 on Build GRCh38
rs759104568 Chr.9: 121107961-121107961 on Build GRCh38
rs1019227104 Chr.9: 121107969-121107969 on Build GRCh38
rs762381514 Chr.9: 121107799-121107799 on Build GRCh38
rs983178306 Chr.9: 121107593-121107593 on Build GRCh38
rs772805917 Chr.9: 121107793-121107793 on Build GRCh38
rs765545921 Chr.9: 121107980-121107980 on Build GRCh38

View Full Product Details