Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123370017-123370519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRB2
Entrez Gene ID
286204
Gene Name
crumbs 2, cell polarity complex component
Gene Aliases
FSGS9, VMCKD
UniGene
Hs.568340
Gene Chromosome Location
Chr.9: 123354065-123380335 on Build GRCh38
Transcripts
NM_173689.6 NR_104603.1 XM_005251934.2 XM_011518556.2 XM_011518557.2 XM_011518558.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3324239 c.1138T>C p.Y380H Chr.9: 123370191-123370191 on Build GRCh38
4796683 c.1160C>T p.T387I Chr.9: 123370213-123370213 on Build GRCh38
3926147 c.1257C>T p.V419V Chr.9: 123370310-123370310 on Build GRCh38
3324244 c.1439C>A p.T480N Chr.9: 123370492-123370492 on Build GRCh38
4796684 c.1160C>T p.T387I Chr.9: 123370213-123370213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55641439 Chr.9: 123370289-123370289 on Build GRCh38
rs775167408 Chr.9: 123370320-123370320 on Build GRCh38
rs139824767 Chr.9: 123370165-123370165 on Build GRCh38
rs779153310 Chr.9: 123370464-123370464 on Build GRCh38
rs771740605 Chr.9: 123370319-123370319 on Build GRCh38
rs7034379 Chr.9: 123370050-123370050 on Build GRCh38
rs757794643 Chr.9: 123370164-123370164 on Build GRCh38
rs756082545 Chr.9: 123370074-123370074 on Build GRCh38
rs143130521 Chr.9: 123370226-123370226 on Build GRCh38
rs947136180 Chr.9: 123370156-123370156 on Build GRCh38
rs140903037 Chr.9: 123370419-123370419 on Build GRCh38
rs546304271 Chr.9: 123370150-123370150 on Build GRCh38
rs374319931 Chr.9: 123370160-123370160 on Build GRCh38
rs775605972 Chr.9: 123370384-123370384 on Build GRCh38
rs755453438 Chr.9: 123370345-123370345 on Build GRCh38
rs146271715 Chr.9: 123370482-123370482 on Build GRCh38
rs759396169 Chr.9: 123370305-123370305 on Build GRCh38
rs762487019 Chr.9: 123370425-123370425 on Build GRCh38
rs537794968 Chr.9: 123370271-123370271 on Build GRCh38
rs762973665 Chr.9: 123370224-123370224 on Build GRCh38
rs772240881 Chr.9: 123370475-123370475 on Build GRCh38
rs753596312 Chr.9: 123370333-123370333 on Build GRCh38
rs756607676 Chr.9: 123370450-123370450 on Build GRCh38
rs768633459 Chr.9: 123370404-123370404 on Build GRCh38
rs754767713 Chr.9: 123370155-123370155 on Build GRCh38
rs1056808771 Chr.9: 123370394-123370394 on Build GRCh38
rs142345530 Chr.9: 123370481-123370481 on Build GRCh38
rs778871934 Chr.9: 123370370-123370370 on Build GRCh38
rs767283156 Chr.9: 123370246-123370246 on Build GRCh38
rs1019759525 Chr.9: 123370228-123370228 on Build GRCh38
rs770973228 Chr.9: 123370298-123370298 on Build GRCh38
rs755838160 Chr.9: 123370258-123370258 on Build GRCh38
rs761566722 Chr.9: 123370420-123370420 on Build GRCh38
rs769475728 Chr.9: 123370071-123370071 on Build GRCh38
rs200850549 Chr.9: 123370463-123370463 on Build GRCh38
rs940616444 Chr.9: 123370391-123370391 on Build GRCh38
rs371574543 Chr.9: 123370293-123370293 on Build GRCh38
rs746675919 Chr.9: 123370382-123370382 on Build GRCh38
rs373818132 Chr.9: 123370207-123370207 on Build GRCh38
rs749886718 Chr.9: 123370342-123370342 on Build GRCh38
rs763787486 Chr.9: 123370259-123370259 on Build GRCh38
rs575296054 Chr.9: 123370486-123370486 on Build GRCh38
rs374856766 Chr.9: 123370116-123370116 on Build GRCh38
rs554777053 Chr.9: 123370227-123370227 on Build GRCh38
rs778114990 Chr.9: 123370457-123370457 on Build GRCh38
rs937013342 Chr.9: 123370199-123370199 on Build GRCh38
rs756959702 Chr.9: 123370335-123370335 on Build GRCh38
rs771116975 Chr.9: 123370366-123370366 on Build GRCh38
rs926932466 Chr.9: 123370166-123370166 on Build GRCh38
rs765777464 Chr.9: 123370430-123370430 on Build GRCh38
rs746444086 Chr.9: 123370085-123370085 on Build GRCh38
rs763947687 Chr.9: 123370326-123370326 on Build GRCh38
rs368041718 Chr.9: 123370357-123370357 on Build GRCh38
rs745907346 Chr.9: 123370372-123370372 on Build GRCh38
rs781689148 Chr.9: 123370346-123370346 on Build GRCh38
rs770027817 Chr.9: 123370283-123370283 on Build GRCh38
rs759502135 Chr.9: 123370234-123370234 on Build GRCh38
rs1030620327 Chr.9: 123370472-123370472 on Build GRCh38
rs146203704 Chr.9: 123370423-123370423 on Build GRCh38
rs138891622 Chr.9: 123370449-123370449 on Build GRCh38
rs7021276 Chr.9: 123370095-123370095 on Build GRCh38
rs573438878 Chr.9: 123370479-123370479 on Build GRCh38
rs34178628 Chr.9: 123370309-123370310 on Build GRCh38
rs773294592 Chr.9: 123370219-123370219 on Build GRCh38
rs151308177 Chr.9: 123370145-123370145 on Build GRCh38
rs200968041 Chr.9: 123370172-123370172 on Build GRCh38
rs199500173 Chr.9: 123370471-123370471 on Build GRCh38
rs567786763 Chr.9: 123370205-123370205 on Build GRCh38
rs202196753 Chr.9: 123370192-123370192 on Build GRCh38
rs747522263 Chr.9: 123370170-123370170 on Build GRCh38
rs865783695 Chr.9: 123370468-123370468 on Build GRCh38
rs777846457 Chr.9: 123370144-123370144 on Build GRCh38
rs754143283 Chr.9: 123370461-123370461 on Build GRCh38
rs752469529 Chr.9: 123370255-123370255 on Build GRCh38
rs34802652 Chr.9: 123370270-123370270 on Build GRCh38
rs764886749 Chr.9: 123370422-123370422 on Build GRCh38
rs1018249210 Chr.9: 123370075-123370075 on Build GRCh38
rs781415118 Chr.9: 123370275-123370275 on Build GRCh38
rs748454767 Chr.9: 123370282-123370282 on Build GRCh38
rs199630307 Chr.9: 123370299-123370299 on Build GRCh38
rs570429818 Chr.9: 123370127-123370127 on Build GRCh38
rs150981911 Chr.9: 123370352-123370352 on Build GRCh38
rs373209085 Chr.9: 123370076-123370076 on Build GRCh38
rs748544542 Chr.9: 123370350-123370350 on Build GRCh38
rs191927047 Chr.9: 123370115-123370115 on Build GRCh38
rs760664924 Chr.9: 123370389-123370389 on Build GRCh38
rs777045044 Chr.9: 123370181-123370181 on Build GRCh38
rs750498594 Chr.9: 123370140-123370140 on Build GRCh38
rs145948620 Chr.9: 123370163-123370163 on Build GRCh38
rs760895610 Chr.9: 123370112-123370112 on Build GRCh38
rs150487212 Chr.9: 123370351-123370351 on Build GRCh38
rs923205937 Chr.9: 123370104-123370104 on Build GRCh38
rs753426684 Chr.9: 123370268-123370268 on Build GRCh38
rs370371819 Chr.9: 123370101-123370101 on Build GRCh38
rs747803718 Chr.9: 123370068-123370068 on Build GRCh38
rs766249014 Chr.9: 123370146-123370146 on Build GRCh38
rs777423121 Chr.9: 123370161-123370161 on Build GRCh38
rs577180539 Chr.9: 123370338-123370338 on Build GRCh38

View Full Product Details