Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13619075-13619588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL6
Entrez Gene ID
25975
Gene Name
EGF like domain multiple 6
Gene Aliases
MAEG, W80
UniGene
Hs.12844
Gene Chromosome Location
Chr.X: 13569575-13633575 on Build GRCh38
Transcripts
NM_001167890.1 NM_015507.3

SNP Detail

SNP ID Chromosome Location
rs762152150 Chr.X: 13619195-13619195 on Build GRCh38
rs139523577 Chr.X: 13619187-13619187 on Build GRCh38
rs180779900 Chr.X: 13619387-13619387 on Build GRCh38
rs755901786 Chr.X: 13619182-13619182 on Build GRCh38
rs189826048 Chr.X: 13619345-13619345 on Build GRCh38
rs751895906 Chr.X: 13619269-13619269 on Build GRCh38
rs1051623615 Chr.X: 13619458-13619458 on Build GRCh38
rs755664274 Chr.X: 13619286-13619286 on Build GRCh38
rs910576160 Chr.X: 13619454-13619454 on Build GRCh38
rs952779691 Chr.X: 13619104-13619104 on Build GRCh38
rs200583207 Chr.X: 13619184-13619184 on Build GRCh38
rs200143589 Chr.X: 13619192-13619192 on Build GRCh38
rs752035678 Chr.X: 13619134-13619134 on Build GRCh38
rs769053415 Chr.X: 13619462-13619462 on Build GRCh38
rs2370238 Chr.X: 13619331-13619331 on Build GRCh38
rs776071203 Chr.X: 13619351-13619351 on Build GRCh38
rs35842085 Chr.X: 13619193-13619193 on Build GRCh38
rs993805300 Chr.X: 13619449-13619449 on Build GRCh38
rs897916978 Chr.X: 13619546-13619546 on Build GRCh38
rs1050844009 Chr.X: 13619431-13619431 on Build GRCh38
rs879126413 Chr.X: 13619346-13619346 on Build GRCh38
rs141600459 Chr.X: 13619163-13619163 on Build GRCh38
rs770872193 Chr.X: 13619191-13619191 on Build GRCh38
rs1052715895 Chr.X: 13619522-13619522 on Build GRCh38
rs956822500 Chr.X: 13619343-13619343 on Build GRCh38
rs185164006 Chr.X: 13619101-13619101 on Build GRCh38
rs766712775 Chr.X: 13619128-13619128 on Build GRCh38
rs761849411 Chr.X: 13619226-13619226 on Build GRCh38
rs374821548 Chr.X: 13619308-13619308 on Build GRCh38
rs780584224 Chr.X: 13619149-13619149 on Build GRCh38
rs777369933 Chr.X: 13619293-13619293 on Build GRCh38
rs763119301 Chr.X: 13619246-13619246 on Build GRCh38
rs770047447 Chr.X: 13619238-13619238 on Build GRCh38
rs755443919 Chr.X: 13619135-13619135 on Build GRCh38
rs775526980 Chr.X: 13619129-13619129 on Build GRCh38
rs1037757481 Chr.X: 13619503-13619503 on Build GRCh38
rs760021622 Chr.X: 13619276-13619276 on Build GRCh38
rs923492448 Chr.X: 13619382-13619382 on Build GRCh38
rs968857634 Chr.X: 13619283-13619283 on Build GRCh38
rs745991336 Chr.X: 13619327-13619327 on Build GRCh38
rs185629793 Chr.X: 13619463-13619463 on Build GRCh38
rs766654733 Chr.X: 13619260-13619260 on Build GRCh38
rs776641020 Chr.X: 13619218-13619218 on Build GRCh38
rs773440608 Chr.X: 13619243-13619243 on Build GRCh38
rs752298920 Chr.X: 13619285-13619285 on Build GRCh38
rs1022660008 Chr.X: 13619279-13619279 on Build GRCh38
rs1012968679 Chr.X: 13619265-13619265 on Build GRCh38
rs768068306 Chr.X: 13619282-13619282 on Build GRCh38
rs765371497 Chr.X: 13619359-13619359 on Build GRCh38
rs897772497 Chr.X: 13619433-13619433 on Build GRCh38

View Full Product Details