Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14006994-14007540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN8
Entrez Gene ID
54960
Gene Name
gem nuclear organelle associated protein 8
Gene Aliases
FAM51A1
UniGene
Hs.592237
Gene Chromosome Location
Chr.X: 14002494-14030087 on Build GRCh38
Transcripts
NM_001042479.1 NM_001042480.1 NM_017856.2 XM_017029617.1 XM_017029618.1

SNP Detail

SNP ID Chromosome Location
rs754989805 Chr.X: 14007120-14007123 on Build GRCh38
rs1023512 Chr.X: 14007262-14007262 on Build GRCh38
rs1038658067 Chr.X: 14007252-14007252 on Build GRCh38
rs895938029 Chr.X: 14007403-14007403 on Build GRCh38
rs772618819 Chr.X: 14007480-14007480 on Build GRCh38
rs746298044 Chr.X: 14007409-14007409 on Build GRCh38
rs972358662 Chr.X: 14007221-14007221 on Build GRCh38
rs937069787 Chr.X: 14007154-14007154 on Build GRCh38
rs918266746 Chr.X: 14007239-14007239 on Build GRCh38
rs184081579 Chr.X: 14007263-14007263 on Build GRCh38
rs991646691 Chr.X: 14007229-14007229 on Build GRCh38
rs941136011 Chr.X: 14007210-14007210 on Build GRCh38
rs533473076 Chr.X: 14007190-14007190 on Build GRCh38
rs1057380688 Chr.X: 14007400-14007400 on Build GRCh38
rs748016477 Chr.X: 14007334-14007334 on Build GRCh38
rs953908723 Chr.X: 14007157-14007157 on Build GRCh38
rs188245775 Chr.X: 14007024-14007024 on Build GRCh38
rs928393879 Chr.X: 14007137-14007137 on Build GRCh38
rs941680063 Chr.X: 14007241-14007241 on Build GRCh38
rs891297613 Chr.X: 14007469-14007469 on Build GRCh38
rs867327397 Chr.X: 14007408-14007408 on Build GRCh38
rs1007009105 Chr.X: 14007489-14007489 on Build GRCh38
rs181381742 Chr.X: 14007096-14007096 on Build GRCh38
rs775871103 Chr.X: 14007488-14007488 on Build GRCh38
rs1054742 Chr.X: 14007067-14007067 on Build GRCh38
rs904590414 Chr.X: 14007455-14007455 on Build GRCh38
rs1023511 Chr.X: 14007324-14007324 on Build GRCh38
rs929537312 Chr.X: 14007310-14007310 on Build GRCh38
rs985731346 Chr.X: 14007165-14007165 on Build GRCh38
rs774665841 Chr.X: 14007100-14007100 on Build GRCh38

View Full Product Details