Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15244311-15244812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB9
Entrez Gene ID
140462
Gene Name
ankyrin repeat and SOCS box containing 9
Gene Aliases
-
UniGene
Hs.19404
Gene Chromosome Location
Chr.X: 15243987-15270476 on Build GRCh38
Transcripts
NM_001031739.2 NM_001168530.1 NM_001168531.1 NM_024087.2 XM_005274446.1 XM_011545458.1 XM_011545460.1 XM_017029283.1 XM_017029284.1 XM_017029285.1 XM_017029286.1

SNP Detail

SNP ID Chromosome Location
rs746540788 Chr.X: 15244644-15244644 on Build GRCh38
rs779841209 Chr.X: 15244517-15244517 on Build GRCh38
rs755155910 Chr.X: 15244740-15244740 on Build GRCh38
rs754386551 Chr.X: 15244479-15244479 on Build GRCh38
rs368074940 Chr.X: 15244662-15244662 on Build GRCh38
rs776824911 Chr.X: 15244607-15244607 on Build GRCh38
rs750013144 Chr.X: 15244648-15244648 on Build GRCh38
rs936210950 Chr.X: 15244353-15244353 on Build GRCh38
rs765918969 Chr.X: 15244657-15244657 on Build GRCh38
rs758175142 Chr.X: 15244514-15244514 on Build GRCh38
rs759028989 Chr.X: 15244617-15244617 on Build GRCh38
rs762423617 Chr.X: 15244612-15244612 on Build GRCh38
rs770377450 Chr.X: 15244399-15244399 on Build GRCh38
rs34974581 Chr.X: 15244539-15244540 on Build GRCh38
rs906655122 Chr.X: 15244342-15244342 on Build GRCh38
rs201067436 Chr.X: 15244454-15244454 on Build GRCh38
rs751268734 Chr.X: 15244661-15244661 on Build GRCh38
rs149241104 Chr.X: 15244548-15244548 on Build GRCh38
rs765640626 Chr.X: 15244634-15244634 on Build GRCh38
rs780790876 Chr.X: 15244539-15244539 on Build GRCh38
rs371063369 Chr.X: 15244475-15244475 on Build GRCh38
rs778823012 Chr.X: 15244768-15244768 on Build GRCh38
rs762550083 Chr.X: 15244653-15244653 on Build GRCh38
rs768778112 Chr.X: 15244431-15244431 on Build GRCh38
rs780652980 Chr.X: 15244693-15244693 on Build GRCh38
rs78887440 Chr.X: 15244691-15244691 on Build GRCh38
rs374177117 Chr.X: 15244547-15244547 on Build GRCh38
rs764837952 Chr.X: 15244497-15244497 on Build GRCh38
rs762181579 Chr.X: 15244493-15244493 on Build GRCh38
rs927258206 Chr.X: 15244731-15244731 on Build GRCh38
rs772248162 Chr.X: 15244473-15244473 on Build GRCh38
rs761011529 Chr.X: 15244478-15244478 on Build GRCh38
rs760754250 Chr.X: 15244592-15244592 on Build GRCh38
rs112382242 Chr.X: 15244691-15244692 on Build GRCh38
rs913470142 Chr.X: 15244772-15244772 on Build GRCh38
rs890161077 Chr.X: 15244419-15244419 on Build GRCh38
rs769090437 Chr.X: 15244604-15244604 on Build GRCh38
rs754805342 Chr.X: 15244527-15244527 on Build GRCh38
rs964363990 Chr.X: 15244484-15244484 on Build GRCh38
rs1010810603 Chr.X: 15244406-15244406 on Build GRCh38
rs749749849 Chr.X: 15244722-15244722 on Build GRCh38
rs1047483144 Chr.X: 15244398-15244398 on Build GRCh38
rs766917472 Chr.X: 15244692-15244693 on Build GRCh38
rs761932422 Chr.X: 15244463-15244463 on Build GRCh38
rs776007989 Chr.X: 15244571-15244571 on Build GRCh38
rs749923006 Chr.X: 15244513-15244513 on Build GRCh38
rs774246796 Chr.X: 15244457-15244457 on Build GRCh38
rs771890468 Chr.X: 15244518-15244518 on Build GRCh38
rs760625112 Chr.X: 15244560-15244560 on Build GRCh38
rs376203367 Chr.X: 15244746-15244746 on Build GRCh38
rs28628283 Chr.X: 15244692-15244692 on Build GRCh38
rs766263309 Chr.X: 15244562-15244562 on Build GRCh38
rs923802006 Chr.X: 15244699-15244700 on Build GRCh38
rs144457529 Chr.X: 15244550-15244550 on Build GRCh38
rs879004698 Chr.X: 15244757-15244757 on Build GRCh38
rs112232773 Chr.X: 15244409-15244409 on Build GRCh38
rs184687335 Chr.X: 15244423-15244423 on Build GRCh38
rs35732224 Chr.X: 15244703-15244704 on Build GRCh38
rs746428099 Chr.X: 15244354-15244354 on Build GRCh38

View Full Product Details