Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15282722-15283225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB11
Entrez Gene ID
140456
Gene Name
ankyrin repeat and SOCS box containing 11
Gene Aliases
-
UniGene
Hs.352183
Gene Chromosome Location
Chr.X: 15281697-15315624 on Build GRCh38
Transcripts
NM_001012428.2 NM_001201583.1 NM_080873.2

SNP Detail

SNP ID Chromosome Location
rs891613776 Chr.X: 15282953-15282953 on Build GRCh38
rs542026463 Chr.X: 15282979-15282980 on Build GRCh38
rs73635074 Chr.X: 15282840-15282840 on Build GRCh38
rs749132047 Chr.X: 15283164-15283164 on Build GRCh38
rs917028539 Chr.X: 15282814-15282814 on Build GRCh38
rs200099511 Chr.X: 15283016-15283017 on Build GRCh38
rs888932886 Chr.X: 15282858-15282859 on Build GRCh38
rs765331480 Chr.X: 15282774-15282774 on Build GRCh38
rs1036407997 Chr.X: 15283183-15283184 on Build GRCh38
rs1028805220 Chr.X: 15282798-15282799 on Build GRCh38
rs956850644 Chr.X: 15283007-15283007 on Build GRCh38
rs763443756 Chr.X: 15282966-15282967 on Build GRCh38
rs893657034 Chr.X: 15282978-15283001 on Build GRCh38
rs867237680 Chr.X: 15282992-15282992 on Build GRCh38
rs891187038 Chr.X: 15282989-15282989 on Build GRCh38
rs940457780 Chr.X: 15283181-15283181 on Build GRCh38
rs113376517 Chr.X: 15282976-15282977 on Build GRCh38
rs992375175 Chr.X: 15283152-15283152 on Build GRCh38
rs532564335 Chr.X: 15283201-15283201 on Build GRCh38
rs200897958 Chr.X: 15282955-15282956 on Build GRCh38
rs935927074 Chr.X: 15282849-15282849 on Build GRCh38
rs1025802404 Chr.X: 15282775-15282775 on Build GRCh38
rs1018242158 Chr.X: 15282995-15282995 on Build GRCh38
rs201885038 Chr.X: 15282978-15282979 on Build GRCh38
rs988508833 Chr.X: 15283008-15283008 on Build GRCh38
rs189973709 Chr.X: 15283004-15283004 on Build GRCh38
rs757032478 Chr.X: 15283169-15283169 on Build GRCh38
rs751253375 Chr.X: 15282968-15282968 on Build GRCh38
rs995770123 Chr.X: 15283106-15283106 on Build GRCh38
rs192223431 Chr.X: 15283045-15283045 on Build GRCh38
rs1007387671 Chr.X: 15282991-15282991 on Build GRCh38
rs756807071 Chr.X: 15282975-15282975 on Build GRCh38
rs1026338115 Chr.X: 15283131-15283131 on Build GRCh38
rs994768148 Chr.X: 15282787-15282787 on Build GRCh38
rs780871030 Chr.X: 15282976-15282976 on Build GRCh38
rs949792789 Chr.X: 15282821-15282821 on Build GRCh38
rs754227640 Chr.X: 15282977-15282977 on Build GRCh38
rs367631030 Chr.X: 15282800-15282800 on Build GRCh38
rs1022260509 Chr.X: 15283015-15283016 on Build GRCh38
rs979754018 Chr.X: 15282829-15282829 on Build GRCh38
rs1008758909 Chr.X: 15282902-15282903 on Build GRCh38
rs1054821945 Chr.X: 15282906-15282906 on Build GRCh38
rs868555189 Chr.X: 15282973-15282973 on Build GRCh38
rs755326107 Chr.X: 15282978-15282978 on Build GRCh38
rs945914267 Chr.X: 15282971-15282971 on Build GRCh38
rs899045460 Chr.X: 15282776-15282776 on Build GRCh38
rs779409414 Chr.X: 15282987-15282987 on Build GRCh38
rs926902548 Chr.X: 15283103-15283106 on Build GRCh38
rs983179844 Chr.X: 15282811-15282811 on Build GRCh38
rs762564369 Chr.X: 15282966-15282966 on Build GRCh38

View Full Product Details