Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18165140-18165647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND2
Entrez Gene ID
139105
Gene Name
BEN domain containing 2
Gene Aliases
CXorf20
UniGene
Hs.403802
Gene Chromosome Location
Chr.X: 18162931-18220904 on Build GRCh38
Transcripts
NM_153346.4 XM_011545447.1 XM_017029266.1

SNP Detail

SNP ID Chromosome Location
rs199844789 Chr.X: 18165245-18165245 on Build GRCh38
rs1054730559 Chr.X: 18165542-18165542 on Build GRCh38
rs918220399 Chr.X: 18165436-18165436 on Build GRCh38
rs962586325 Chr.X: 18165389-18165389 on Build GRCh38
rs1001790059 Chr.X: 18165423-18165424 on Build GRCh38
rs755616014 Chr.X: 18165516-18165516 on Build GRCh38
rs1018104311 Chr.X: 18165381-18165381 on Build GRCh38
rs759074673 Chr.X: 18165451-18165451 on Build GRCh38
rs150252640 Chr.X: 18165162-18165162 on Build GRCh38
rs1009812372 Chr.X: 18165279-18165279 on Build GRCh38
rs910653630 Chr.X: 18165605-18165605 on Build GRCh38
rs974047099 Chr.X: 18165395-18165395 on Build GRCh38
rs191033083 Chr.X: 18165531-18165531 on Build GRCh38
rs930645871 Chr.X: 18165593-18165593 on Build GRCh38
rs766441115 Chr.X: 18165259-18165259 on Build GRCh38
rs765324205 Chr.X: 18165228-18165228 on Build GRCh38
rs188974579 Chr.X: 18165332-18165332 on Build GRCh38
rs752337949 Chr.X: 18165491-18165491 on Build GRCh38
rs753759791 Chr.X: 18165577-18165577 on Build GRCh38
rs763200842 Chr.X: 18165255-18165255 on Build GRCh38
rs182419596 Chr.X: 18165435-18165435 on Build GRCh38
rs772108408 Chr.X: 18165180-18165180 on Build GRCh38
rs762470241 Chr.X: 18165383-18165383 on Build GRCh38
rs774047430 Chr.X: 18165163-18165163 on Build GRCh38
rs1033401680 Chr.X: 18165452-18165452 on Build GRCh38
rs1044565140 Chr.X: 18165523-18165523 on Build GRCh38
rs917681437 Chr.X: 18165472-18165472 on Build GRCh38
rs1015420168 Chr.X: 18165501-18165501 on Build GRCh38
rs758967965 Chr.X: 18165532-18165532 on Build GRCh38
rs947824545 Chr.X: 18165497-18165497 on Build GRCh38
rs145864716 Chr.X: 18165175-18165175 on Build GRCh38
rs992279648 Chr.X: 18165466-18165466 on Build GRCh38
rs903530809 Chr.X: 18165422-18165422 on Build GRCh38
rs186163955 Chr.X: 18165227-18165227 on Build GRCh38
rs775094003 Chr.X: 18165186-18165186 on Build GRCh38
rs944797998 Chr.X: 18165609-18165609 on Build GRCh38
rs1021954756 Chr.X: 18165418-18165418 on Build GRCh38
rs973990004 Chr.X: 18165539-18165539 on Build GRCh38
rs986108787 Chr.X: 18165598-18165598 on Build GRCh38
rs765641688 Chr.X: 18165407-18165407 on Build GRCh38
rs187393749 Chr.X: 18165465-18165465 on Build GRCh38

View Full Product Details