Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19915483-19915987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf23
Entrez Gene ID
256643
Gene Name
chromosome X open reading frame 23
Gene Aliases
-
UniGene
Hs.28896
Gene Chromosome Location
Chr.X: 19912860-19991040 on Build GRCh38
Transcripts
NM_198279.3 XM_005274473.3 XM_005274474.3 XM_005274475.3 XM_011545475.2 XM_011545476.2 XM_011545477.2 XM_011545478.2 XM_011545479.2 XM_011545482.2 XM_017029390.1 XM_017029391.1

SNP Detail

SNP ID Chromosome Location
rs892097471 Chr.X: 19915516-19915516 on Build GRCh38
rs755822989 Chr.X: 19915926-19915926 on Build GRCh38
rs886151045 Chr.X: 19915575-19915575 on Build GRCh38
rs1044686418 Chr.X: 19915640-19915640 on Build GRCh38
rs997365296 Chr.X: 19915960-19915960 on Build GRCh38
rs914539572 Chr.X: 19915772-19915772 on Build GRCh38
rs907133097 Chr.X: 19915541-19915541 on Build GRCh38
rs1007501138 Chr.X: 19915560-19915560 on Build GRCh38
rs986926099 Chr.X: 19915673-19915673 on Build GRCh38
rs965938228 Chr.X: 19915569-19915569 on Build GRCh38
rs909445724 Chr.X: 19915607-19915607 on Build GRCh38
rs573801528 Chr.X: 19915608-19915608 on Build GRCh38
rs183170596 Chr.X: 19915817-19915817 on Build GRCh38
rs1044804787 Chr.X: 19915836-19915836 on Build GRCh38
rs555515608 Chr.X: 19915527-19915527 on Build GRCh38
rs73443591 Chr.X: 19915540-19915540 on Build GRCh38
rs892234964 Chr.X: 19915732-19915732 on Build GRCh38
rs186878255 Chr.X: 19915602-19915602 on Build GRCh38
rs950684645 Chr.X: 19915594-19915594 on Build GRCh38
rs878880106 Chr.X: 19915618-19915618 on Build GRCh38
rs928793245 Chr.X: 19915641-19915641 on Build GRCh38
rs903552561 Chr.X: 19915792-19915792 on Build GRCh38
rs967978001 Chr.X: 19915547-19915547 on Build GRCh38
rs1043396343 Chr.X: 19915767-19915767 on Build GRCh38
rs893169121 Chr.X: 19915518-19915518 on Build GRCh38
rs964439742 Chr.X: 19915610-19915610 on Build GRCh38
rs1017558748 Chr.X: 19915875-19915875 on Build GRCh38
rs781452237 Chr.X: 19915847-19915847 on Build GRCh38
rs919350350 Chr.X: 19915577-19915577 on Build GRCh38
rs767015597 Chr.X: 19915574-19915574 on Build GRCh38
rs182265771 Chr.X: 19915599-19915599 on Build GRCh38
rs955614317 Chr.X: 19915655-19915655 on Build GRCh38
rs541507074 Chr.X: 19915656-19915656 on Build GRCh38
rs1056583298 Chr.X: 19915725-19915725 on Build GRCh38
rs948630616 Chr.X: 19915523-19915523 on Build GRCh38
rs766963952 Chr.X: 19915752-19915752 on Build GRCh38
rs1044343481 Chr.X: 19915528-19915528 on Build GRCh38
rs148020057 Chr.X: 19915565-19915565 on Build GRCh38
rs191770669 Chr.X: 19915754-19915754 on Build GRCh38
rs1009245863 Chr.X: 19915536-19915536 on Build GRCh38
rs928521721 Chr.X: 19915920-19915920 on Build GRCh38
rs924811923 Chr.X: 19915727-19915727 on Build GRCh38

View Full Product Details