Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19929478-19929990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf23
Entrez Gene ID
256643
Gene Name
chromosome X open reading frame 23
Gene Aliases
-
UniGene
Hs.28896
Gene Chromosome Location
Chr.X: 19912860-19991040 on Build GRCh38
Transcripts
NM_198279.3 XM_005274473.3 XM_005274474.3 XM_005274475.3 XM_011545475.2 XM_011545476.2 XM_011545477.2 XM_011545478.2 XM_011545479.2 XM_011545482.2 XM_017029390.1 XM_017029391.1

SNP Detail

SNP ID Chromosome Location
rs991206959 Chr.X: 19929771-19929771 on Build GRCh38
rs878915016 Chr.X: 19929671-19929671 on Build GRCh38
rs765199560 Chr.X: 19929895-19929895 on Build GRCh38
rs749152983 Chr.X: 19929849-19929849 on Build GRCh38
rs750873055 Chr.X: 19929798-19929798 on Build GRCh38
rs747587175 Chr.X: 19929690-19929690 on Build GRCh38
rs748219446 Chr.X: 19929824-19929824 on Build GRCh38
rs774005570 Chr.X: 19929855-19929855 on Build GRCh38
rs780469602 Chr.X: 19929669-19929669 on Build GRCh38
rs183432077 Chr.X: 19929640-19929640 on Build GRCh38
rs867571111 Chr.X: 19929897-19929897 on Build GRCh38
rs763353881 Chr.X: 19929909-19929909 on Build GRCh38
rs932698015 Chr.X: 19929579-19929579 on Build GRCh38
rs73443601 Chr.X: 19929758-19929758 on Build GRCh38
rs947568123 Chr.X: 19929754-19929754 on Build GRCh38
rs761549140 Chr.X: 19929950-19929950 on Build GRCh38
rs368213137 Chr.X: 19929815-19929815 on Build GRCh38
rs769089735 Chr.X: 19929701-19929701 on Build GRCh38
rs912800223 Chr.X: 19929565-19929565 on Build GRCh38
rs370505066 Chr.X: 19929820-19929820 on Build GRCh38
rs1054210114 Chr.X: 19929559-19929559 on Build GRCh38
rs770057089 Chr.X: 19929949-19929950 on Build GRCh38
rs770969944 Chr.X: 19929850-19929850 on Build GRCh38
rs752010720 Chr.X: 19929943-19929943 on Build GRCh38
rs759348070 Chr.X: 19929856-19929856 on Build GRCh38
rs774882064 Chr.X: 19929577-19929577 on Build GRCh38
rs891100533 Chr.X: 19929636-19929636 on Build GRCh38
rs373927242 Chr.X: 19929876-19929876 on Build GRCh38
rs766849440 Chr.X: 19929914-19929914 on Build GRCh38
rs754128138 Chr.X: 19929762-19929762 on Build GRCh38
rs780275831 Chr.X: 19929819-19929819 on Build GRCh38
rs761918425 Chr.X: 19929779-19929779 on Build GRCh38
rs771998295 Chr.X: 19929858-19929858 on Build GRCh38
rs747824743 Chr.X: 19929834-19929834 on Build GRCh38
rs773407149 Chr.X: 19929906-19929906 on Build GRCh38
rs780753038 Chr.X: 19929830-19929830 on Build GRCh38
rs765439223 Chr.X: 19929792-19929792 on Build GRCh38
rs1043097236 Chr.X: 19929823-19929823 on Build GRCh38
rs760805572 Chr.X: 19929743-19929743 on Build GRCh38
rs1027155230 Chr.X: 19929757-19929757 on Build GRCh38
rs142247466 Chr.X: 19929838-19929838 on Build GRCh38
rs934624303 Chr.X: 19929556-19929556 on Build GRCh38
rs759943589 Chr.X: 19929679-19929679 on Build GRCh38
rs145759402 Chr.X: 19929605-19929605 on Build GRCh38
rs891991094 Chr.X: 19929732-19929732 on Build GRCh38
rs150826975 Chr.X: 19929870-19929870 on Build GRCh38
rs73631376 Chr.X: 19929649-19929649 on Build GRCh38
rs777395420 Chr.X: 19929843-19929843 on Build GRCh38

View Full Product Details