Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 20138480-20138992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AX
Entrez Gene ID
1964
Gene Name
eukaryotic translation initiation factor 1A, X-linked
Gene Aliases
EIF1A, EIF1AP1, EIF4C, eIF-1A, eIF-4C
UniGene
Hs.522590
Gene Chromosome Location
Chr.X: 20124518-20141848 on Build GRCh38
Transcripts
NM_001412.3

SNP Detail

SNP ID Chromosome Location
rs769115973 Chr.X: 20138764-20138764 on Build GRCh38
rs939699417 Chr.X: 20138758-20138758 on Build GRCh38
rs770135230 Chr.X: 20138963-20138963 on Build GRCh38
rs779464943 Chr.X: 20138875-20138875 on Build GRCh38
rs762878162 Chr.X: 20138510-20138510 on Build GRCh38
rs868028391 Chr.X: 20138620-20138620 on Build GRCh38
rs145529347 Chr.X: 20138524-20138524 on Build GRCh38
rs750320713 Chr.X: 20138506-20138506 on Build GRCh38
rs929062284 Chr.X: 20138894-20138894 on Build GRCh38
rs771871926 Chr.X: 20138637-20138641 on Build GRCh38
rs753933670 Chr.X: 20138639-20138641 on Build GRCh38
rs1021478012 Chr.X: 20138947-20138947 on Build GRCh38
rs915852290 Chr.X: 20138781-20138781 on Build GRCh38
rs771410912 Chr.X: 20138654-20138654 on Build GRCh38
rs1051317720 Chr.X: 20138747-20138747 on Build GRCh38
rs933485508 Chr.X: 20138743-20138743 on Build GRCh38
rs943588265 Chr.X: 20138527-20138527 on Build GRCh38
rs377249043 Chr.X: 20138517-20138517 on Build GRCh38
rs747413383 Chr.X: 20138757-20138757 on Build GRCh38
rs777058573 Chr.X: 20138841-20138841 on Build GRCh38
rs762136651 Chr.X: 20138510-20138511 on Build GRCh38
rs977463451 Chr.X: 20138711-20138711 on Build GRCh38
rs778122765 Chr.X: 20138573-20138573 on Build GRCh38
rs1002162141 Chr.X: 20138866-20138866 on Build GRCh38
rs756778021 Chr.X: 20138518-20138518 on Build GRCh38
rs1033578683 Chr.X: 20138874-20138874 on Build GRCh38
rs767914931 Chr.X: 20138632-20138633 on Build GRCh38
rs1048766617 Chr.X: 20138902-20138902 on Build GRCh38
rs745408269 Chr.X: 20138926-20138926 on Build GRCh38
rs923479779 Chr.X: 20138629-20138629 on Build GRCh38
rs1032336139 Chr.X: 20138965-20138965 on Build GRCh38
rs377327969 Chr.X: 20138644-20138644 on Build GRCh38
rs967472465 Chr.X: 20138599-20138599 on Build GRCh38
rs778488157 Chr.X: 20138520-20138520 on Build GRCh38
rs887517861 Chr.X: 20138914-20138914 on Build GRCh38
rs779812636 Chr.X: 20138659-20138662 on Build GRCh38
rs778986847 Chr.X: 20138641-20138641 on Build GRCh38
rs200817041 Chr.X: 20138582-20138582 on Build GRCh38
rs762374729 Chr.X: 20138915-20138915 on Build GRCh38
rs760888515 Chr.X: 20138633-20138636 on Build GRCh38
rs757280897 Chr.X: 20138611-20138611 on Build GRCh38
rs966782263 Chr.X: 20138948-20138948 on Build GRCh38
rs773378025 Chr.X: 20138633-20138633 on Build GRCh38
rs751362563 Chr.X: 20138521-20138521 on Build GRCh38

View Full Product Details