Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23703293-23703793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT9
Entrez Gene ID
23597
Gene Name
acyl-CoA thioesterase 9
Gene Aliases
ACATE2, CGI-16, MT-ACT48, MTACT48
UniGene
Hs.298885
Gene Chromosome Location
Chr.X: 23701055-23743290 on Build GRCh38
Transcripts
NM_001033583.2 NM_001037171.1 XM_005274472.2

SNP Detail

SNP ID Chromosome Location
rs779365368 Chr.X: 23703350-23703353 on Build GRCh38
rs920516313 Chr.X: 23703511-23703511 on Build GRCh38
rs1021766749 Chr.X: 23703746-23703746 on Build GRCh38
rs776573491 Chr.X: 23703609-23703609 on Build GRCh38
rs989302780 Chr.X: 23703729-23703729 on Build GRCh38
rs141484914 Chr.X: 23703327-23703330 on Build GRCh38
rs1021903527 Chr.X: 23703366-23703366 on Build GRCh38
rs935179010 Chr.X: 23703684-23703684 on Build GRCh38
rs142239349 Chr.X: 23703619-23703619 on Build GRCh38
rs1009477634 Chr.X: 23703378-23703378 on Build GRCh38
rs768843183 Chr.X: 23703598-23703598 on Build GRCh38
rs970851800 Chr.X: 23703638-23703638 on Build GRCh38
rs1026629686 Chr.X: 23703471-23703471 on Build GRCh38
rs990886960 Chr.X: 23703631-23703631 on Build GRCh38
rs747044948 Chr.X: 23703561-23703561 on Build GRCh38
rs746258463 Chr.X: 23703525-23703525 on Build GRCh38
rs890529338 Chr.X: 23703564-23703564 on Build GRCh38
rs758797456 Chr.X: 23703540-23703540 on Build GRCh38
rs868463128 Chr.X: 23703336-23703336 on Build GRCh38
rs952498592 Chr.X: 23703479-23703479 on Build GRCh38
rs1034051703 Chr.X: 23703445-23703445 on Build GRCh38
rs893778662 Chr.X: 23703541-23703541 on Build GRCh38
rs940551067 Chr.X: 23703472-23703472 on Build GRCh38
rs1023332354 Chr.X: 23703633-23703633 on Build GRCh38
rs770023790 Chr.X: 23703672-23703672 on Build GRCh38
rs931867791 Chr.X: 23703516-23703516 on Build GRCh38
rs949201048 Chr.X: 23703519-23703519 on Build GRCh38
rs935209839 Chr.X: 23703532-23703532 on Build GRCh38
rs1028311157 Chr.X: 23703611-23703611 on Build GRCh38
rs915139783 Chr.X: 23703730-23703730 on Build GRCh38
rs748309317 Chr.X: 23703610-23703610 on Build GRCh38
rs373915620 Chr.X: 23703644-23703644 on Build GRCh38
rs890568230 Chr.X: 23703376-23703376 on Build GRCh38
rs923753543 Chr.X: 23703656-23703656 on Build GRCh38
rs189980983 Chr.X: 23703677-23703677 on Build GRCh38
rs893801157 Chr.X: 23703351-23703351 on Build GRCh38
rs1029504960 Chr.X: 23703372-23703372 on Build GRCh38
rs959973308 Chr.X: 23703468-23703468 on Build GRCh38
rs760154363 Chr.X: 23703593-23703594 on Build GRCh38
rs891850997 Chr.X: 23703683-23703683 on Build GRCh38
rs747188790 Chr.X: 23703379-23703379 on Build GRCh38
rs923782582 Chr.X: 23703530-23703530 on Build GRCh38
rs776726490 Chr.X: 23703615-23703615 on Build GRCh38
rs1010412018 Chr.X: 23703742-23703742 on Build GRCh38
rs1015062739 Chr.X: 23703593-23703593 on Build GRCh38
rs763337670 Chr.X: 23703319-23703320 on Build GRCh38
rs979771527 Chr.X: 23703437-23703437 on Build GRCh38
rs956259121 Chr.X: 23703355-23703355 on Build GRCh38
rs185290220 Chr.X: 23703542-23703542 on Build GRCh38
rs973655335 Chr.X: 23703470-23703470 on Build GRCh38
rs113264734 Chr.X: 23703725-23703725 on Build GRCh38
rs866333166 Chr.X: 23703337-23703337 on Build GRCh38
rs139470002 Chr.X: 23703325-23703328 on Build GRCh38
rs962255988 Chr.X: 23703401-23703401 on Build GRCh38
rs990532190 Chr.X: 23703484-23703484 on Build GRCh38
rs780157156 Chr.X: 23703362-23703362 on Build GRCh38
rs556204870 Chr.X: 23703336-23703339 on Build GRCh38
rs1008994160 Chr.X: 23703565-23703565 on Build GRCh38
rs916296127 Chr.X: 23703514-23703514 on Build GRCh38
rs1053648203 Chr.X: 23703539-23703539 on Build GRCh38

View Full Product Details