Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24057329-24057841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2S3
Entrez Gene ID
1968
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Gene Aliases
EIF2, EIF2G, EIF2gamma, eIF-2gA
UniGene
Hs.539684
Gene Chromosome Location
Chr.X: 24054948-24078810 on Build GRCh38
Transcripts
NM_001415.3

SNP Detail

SNP ID Chromosome Location
rs868450802 Chr.X: 24057484-24057484 on Build GRCh38
rs753319082 Chr.X: 24057569-24057569 on Build GRCh38
rs760993238 Chr.X: 24057798-24057798 on Build GRCh38
rs752231864 Chr.X: 24057389-24057389 on Build GRCh38
rs759957027 Chr.X: 24057580-24057580 on Build GRCh38
rs757232445 Chr.X: 24057452-24057452 on Build GRCh38
rs772508324 Chr.X: 24057771-24057771 on Build GRCh38
rs746321081 Chr.X: 24057766-24057766 on Build GRCh38
rs765236944 Chr.X: 24057655-24057655 on Build GRCh38
rs143766658 Chr.X: 24057710-24057710 on Build GRCh38
rs749439547 Chr.X: 24057737-24057737 on Build GRCh38
rs751699394 Chr.X: 24057576-24057577 on Build GRCh38
rs778139079 Chr.X: 24057714-24057714 on Build GRCh38
rs999090964 Chr.X: 24057760-24057760 on Build GRCh38
rs891539671 Chr.X: 24057385-24057385 on Build GRCh38
rs753384185 Chr.X: 24057639-24057639 on Build GRCh38
rs767256429 Chr.X: 24057384-24057384 on Build GRCh38
rs753790674 Chr.X: 24057443-24057443 on Build GRCh38
rs748345014 Chr.X: 24057554-24057554 on Build GRCh38
rs1057515578 Chr.X: 24057695-24057695 on Build GRCh38
rs760269170 Chr.X: 24057612-24057612 on Build GRCh38
rs761305657 Chr.X: 24057651-24057651 on Build GRCh38
rs755435237 Chr.X: 24057815-24057815 on Build GRCh38
rs758073633 Chr.X: 24057536-24057536 on Build GRCh38
rs368009248 Chr.X: 24057357-24057357 on Build GRCh38
rs772258826 Chr.X: 24057596-24057596 on Build GRCh38
rs376678844 Chr.X: 24057576-24057576 on Build GRCh38
rs770016776 Chr.X: 24057558-24057558 on Build GRCh38
rs763550804 Chr.X: 24057411-24057411 on Build GRCh38
rs911791849 Chr.X: 24057401-24057401 on Build GRCh38
rs779477417 Chr.X: 24057757-24057757 on Build GRCh38
rs761857770 Chr.X: 24057611-24057612 on Build GRCh38
rs745737387 Chr.X: 24057468-24057468 on Build GRCh38
rs372984450 Chr.X: 24057601-24057601 on Build GRCh38
rs764179777 Chr.X: 24057561-24057565 on Build GRCh38
rs781485564 Chr.X: 24057539-24057539 on Build GRCh38
rs758248150 Chr.X: 24057684-24057684 on Build GRCh38
rs766361482 Chr.X: 24057613-24057614 on Build GRCh38
rs376468211 Chr.X: 24057381-24057381 on Build GRCh38
rs186942094 Chr.X: 24057616-24057616 on Build GRCh38
rs374957172 Chr.X: 24057795-24057795 on Build GRCh38
rs747044374 Chr.X: 24057775-24057775 on Build GRCh38
rs369734267 Chr.X: 24057577-24057577 on Build GRCh38
rs779668646 Chr.X: 24057699-24057699 on Build GRCh38
rs373670584 Chr.X: 24057575-24057575 on Build GRCh38
rs1052544228 Chr.X: 24057811-24057811 on Build GRCh38
rs373616711 Chr.X: 24057728-24057728 on Build GRCh38
rs972456777 Chr.X: 24057415-24057415 on Build GRCh38
rs16997659 Chr.X: 24057745-24057745 on Build GRCh38
rs764041264 Chr.X: 24057383-24057383 on Build GRCh38
rs376466811 Chr.X: 24057779-24057779 on Build GRCh38
rs370164270 Chr.X: 24057677-24057677 on Build GRCh38
rs755625866 Chr.X: 24057408-24057408 on Build GRCh38
rs778681778 Chr.X: 24057458-24057458 on Build GRCh38
rs750096753 Chr.X: 24057813-24057813 on Build GRCh38
rs556362722 Chr.X: 24057713-24057713 on Build GRCh38
rs140083602 Chr.X: 24057704-24057704 on Build GRCh38

View Full Product Details